Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 42276778-42277277 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCG1
Entrez Gene ID
9619
Gene Name
ATP binding cassette subfamily G member 1
Gene Aliases
ABC8, WHITE1
UniGene
Hs.124649
Gene Chromosome Location
Chr.21: 42199689-42304389 on Build GRCh38
Transcripts
NM_004915.3 NM_016818.2 NM_207174.1 NM_207627.1 NM_207628.1 NM_207629.1 XM_011529806.1 XM_011529807.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4777022 c.577G>A p.E193K Chr.21: 42276928-42276928 on Build GRCh38
149235 c.544-6C>T p.? Chr.21: 42276889-42276889 on Build GRCh38
6997006 c.543G>A p.S181S Chr.21: 42276900-42276900 on Build GRCh38
6997008 c.549G>A p.S183S Chr.21: 42276900-42276900 on Build GRCh38
5526083 c.581G>A p.R194K Chr.21: 42276938-42276938 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs890155030 Chr.21: 42277150-42277150 on Build GRCh38
rs879226979 Chr.21: 42276847-42276847 on Build GRCh38
rs915151098 Chr.21: 42276986-42276986 on Build GRCh38
rs916054047 Chr.21: 42277120-42277120 on Build GRCh38
rs927975989 Chr.21: 42277003-42277003 on Build GRCh38
rs534238684 Chr.21: 42277152-42277152 on Build GRCh38
rs201964021 Chr.21: 42276992-42276992 on Build GRCh38
rs567232191 Chr.21: 42276839-42276839 on Build GRCh38
rs910096542 Chr.21: 42277140-42277140 on Build GRCh38
rs56191607 Chr.21: 42277008-42277008 on Build GRCh38
rs541066993 Chr.21: 42277234-42277234 on Build GRCh38
rs968858231 Chr.21: 42276817-42276817 on Build GRCh38
rs987211956 Chr.21: 42276908-42276908 on Build GRCh38
rs964943930 Chr.21: 42276956-42276956 on Build GRCh38
rs984799635 Chr.21: 42277140-42277142 on Build GRCh38
rs111977731 Chr.21: 42277182-42277182 on Build GRCh38
rs1011172273 Chr.21: 42277191-42277191 on Build GRCh38
rs777669875 Chr.21: 42276969-42276969 on Build GRCh38
rs377637926 Chr.21: 42276890-42276890 on Build GRCh38
rs2234718 Chr.21: 42277085-42277085 on Build GRCh38
rs56145149 Chr.21: 42276889-42276889 on Build GRCh38
rs565742021 Chr.21: 42277016-42277016 on Build GRCh38
rs780192631 Chr.21: 42277251-42277251 on Build GRCh38
rs777999545 Chr.21: 42277173-42277173 on Build GRCh38
rs933708800 Chr.21: 42277186-42277186 on Build GRCh38
rs879662244 Chr.21: 42277245-42277245 on Build GRCh38
rs148465364 Chr.21: 42277000-42277000 on Build GRCh38
rs1017384850 Chr.21: 42276942-42276942 on Build GRCh38
rs758096166 Chr.21: 42276939-42276939 on Build GRCh38
rs776802807 Chr.21: 42276854-42276854 on Build GRCh38
rs375659857 Chr.21: 42277020-42277020 on Build GRCh38
rs941532888 Chr.21: 42277019-42277019 on Build GRCh38
rs545554853 Chr.21: 42277153-42277153 on Build GRCh38
rs945375240 Chr.21: 42276802-42276802 on Build GRCh38
rs527705683 Chr.21: 42276813-42276813 on Build GRCh38
rs370673215 Chr.21: 42276984-42276984 on Build GRCh38
rs753275307 Chr.21: 42276900-42276900 on Build GRCh38
rs534606653 Chr.21: 42277017-42277017 on Build GRCh38
rs1038587117 Chr.21: 42277069-42277069 on Build GRCh38
rs1047371860 Chr.21: 42277122-42277122 on Build GRCh38
rs774731367 Chr.21: 42276863-42276863 on Build GRCh38
rs373480801 Chr.21: 42276985-42276985 on Build GRCh38
rs1022190578 Chr.21: 42277214-42277214 on Build GRCh38
rs369825449 Chr.21: 42276855-42276855 on Build GRCh38
rs1042441873 Chr.21: 42276812-42276812 on Build GRCh38
rs112352275 Chr.21: 42277070-42277070 on Build GRCh38
rs138056067 Chr.21: 42276899-42276899 on Build GRCh38
rs188000057 Chr.21: 42276826-42276826 on Build GRCh38
rs759147987 Chr.21: 42276902-42276902 on Build GRCh38
rs142441719 Chr.21: 42276923-42276923 on Build GRCh38
rs150977124 Chr.21: 42277170-42277170 on Build GRCh38
rs752755711 Chr.21: 42277087-42277087 on Build GRCh38
rs763206321 Chr.21: 42276859-42276859 on Build GRCh38
rs191949270 Chr.21: 42276840-42276840 on Build GRCh38
rs373825683 Chr.21: 42276827-42276827 on Build GRCh38
rs771023413 Chr.21: 42276852-42276852 on Build GRCh38
rs998771541 Chr.21: 42277028-42277028 on Build GRCh38
rs556748233 Chr.21: 42277149-42277149 on Build GRCh38
rs752440795 Chr.21: 42276938-42276938 on Build GRCh38

View Full Product Details