Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24440491-24440983 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADORA2A
Entrez Gene ID
135
Gene Name
adenosine A2a receptor
Gene Aliases
A2aR, ADORA2, RDC8
UniGene
Hs.197029
Gene Chromosome Location
Chr.22: 24423597-24442360 on Build GRCh38
Transcripts
NM_000675.5 NM_001278497.1 NM_001278498.1 NM_001278499.1 NM_001278500.1 NR_103543.1 NR_103544.1 NR_028483.2 NR_028484.2 NR_103546.1

Gene Symbol
ADORA2A-AS1
Entrez Gene ID
646023
Gene Name
ADORA2A antisense RNA 1
Gene Aliases
C22orf45
UniGene
Hs.632761
Gene Chromosome Location
Chr.22: 24429206-24495074 on Build GRCh38
Transcripts
NM_000675.5 NM_001278497.1 NM_001278498.1 NM_001278499.1 NM_001278500.1 NR_103543.1 NR_103544.1 NR_028483.2 NR_028484.2 NR_103546.1

Gene Symbol
SPECC1L-ADORA2A
Entrez Gene ID
101730217
Gene Name
SPECC1L-ADORA2A readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.474384
Gene Chromosome Location
Chr.22: 24270817-24442360 on Build GRCh38
Transcripts
NM_000675.5 NM_001278497.1 NM_001278498.1 NM_001278499.1 NM_001278500.1 NR_103543.1 NR_103544.1 NR_028483.2 NR_028484.2 NR_103546.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3552642 c.604C>T p.L202L Chr.22: 24440854-24440854 on Build GRCh38
1180121 c.481G>A p.E161K Chr.22: 24440731-24440731 on Build GRCh38
6595319 c.421C>T p.L141L Chr.22: 24440671-24440671 on Build GRCh38
5072921 c.377C>T p.A126V Chr.22: 24440627-24440627 on Build GRCh38
6595320 c.603C>A p.F201L Chr.22: 24440853-24440853 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866947347 Chr.22: 24440603-24440603 on Build GRCh38
rs199949453 Chr.22: 24440697-24440697 on Build GRCh38
rs200755653 Chr.22: 24440847-24440847 on Build GRCh38
rs746275437 Chr.22: 24440846-24440846 on Build GRCh38
rs753620615 Chr.22: 24440894-24440894 on Build GRCh38
rs141083636 Chr.22: 24440724-24440724 on Build GRCh38
rs201891365 Chr.22: 24440884-24440884 on Build GRCh38
rs141732109 Chr.22: 24440785-24440785 on Build GRCh38
rs372335618 Chr.22: 24440722-24440722 on Build GRCh38
rs766290851 Chr.22: 24440886-24440886 on Build GRCh38
rs201235831 Chr.22: 24440760-24440760 on Build GRCh38
rs968262442 Chr.22: 24440546-24440546 on Build GRCh38
rs751273195 Chr.22: 24440662-24440663 on Build GRCh38
rs771114928 Chr.22: 24440703-24440703 on Build GRCh38
rs199600702 Chr.22: 24440601-24440601 on Build GRCh38
rs750210199 Chr.22: 24440766-24440766 on Build GRCh38
rs773766284 Chr.22: 24440864-24440864 on Build GRCh38
rs199726273 Chr.22: 24440740-24440740 on Build GRCh38
rs138506386 Chr.22: 24440516-24440516 on Build GRCh38
rs779140681 Chr.22: 24440595-24440595 on Build GRCh38
rs879098425 Chr.22: 24440680-24440680 on Build GRCh38
rs200932591 Chr.22: 24440548-24440548 on Build GRCh38
rs765688222 Chr.22: 24440638-24440638 on Build GRCh38
rs767230391 Chr.22: 24440670-24440670 on Build GRCh38
rs757441319 Chr.22: 24440825-24440825 on Build GRCh38
rs771184124 Chr.22: 24440616-24440616 on Build GRCh38
rs150249680 Chr.22: 24440728-24440728 on Build GRCh38
rs200674969 Chr.22: 24440901-24440901 on Build GRCh38
rs747473116 Chr.22: 24440611-24440611 on Build GRCh38
rs199725333 Chr.22: 24440900-24440900 on Build GRCh38
rs771672458 Chr.22: 24440867-24440867 on Build GRCh38
rs780091269 Chr.22: 24440688-24440688 on Build GRCh38
rs115000790 Chr.22: 24440542-24440542 on Build GRCh38
rs202012742 Chr.22: 24440607-24440607 on Build GRCh38
rs756976329 Chr.22: 24440532-24440532 on Build GRCh38
rs201680338 Chr.22: 24440834-24440834 on Build GRCh38
rs763584556 Chr.22: 24440731-24440731 on Build GRCh38
rs755952858 Chr.22: 24440770-24440770 on Build GRCh38
rs201432461 Chr.22: 24440646-24440646 on Build GRCh38
rs199921896 Chr.22: 24440931-24440931 on Build GRCh38
rs757325519 Chr.22: 24440909-24440909 on Build GRCh38
rs149141630 Chr.22: 24440949-24440949 on Build GRCh38
rs8192446 Chr.22: 24440682-24440682 on Build GRCh38
rs747066875 Chr.22: 24440806-24440806 on Build GRCh38
rs201149976 Chr.22: 24440859-24440859 on Build GRCh38
rs753812201 Chr.22: 24440789-24440789 on Build GRCh38
rs778113831 Chr.22: 24440693-24440693 on Build GRCh38
rs939477540 Chr.22: 24440908-24440908 on Build GRCh38
rs780405269 Chr.22: 24440925-24440925 on Build GRCh38
rs748198311 Chr.22: 24440596-24440596 on Build GRCh38
rs369324965 Chr.22: 24440738-24440738 on Build GRCh38
rs146452856 Chr.22: 24440641-24440641 on Build GRCh38
rs758431179 Chr.22: 24440716-24440716 on Build GRCh38
rs139833983 Chr.22: 24440655-24440655 on Build GRCh38
rs5760421 Chr.22: 24440880-24440880 on Build GRCh38
rs769141421 Chr.22: 24440862-24440862 on Build GRCh38
rs201332411 Chr.22: 24440672-24440672 on Build GRCh38
rs772993000 Chr.22: 24440874-24440874 on Build GRCh38
rs201346032 Chr.22: 24440915-24440915 on Build GRCh38
rs991139776 Chr.22: 24440769-24440769 on Build GRCh38
rs781339364 Chr.22: 24440914-24440914 on Build GRCh38
rs780599770 Chr.22: 24440858-24440858 on Build GRCh38
rs199767901 Chr.22: 24440845-24440845 on Build GRCh38
rs942698715 Chr.22: 24440606-24440606 on Build GRCh38
rs761339165 Chr.22: 24440866-24440866 on Build GRCh38
rs375627708 Chr.22: 24440727-24440727 on Build GRCh38
rs147072171 Chr.22: 24440798-24440798 on Build GRCh38
rs868830392 Chr.22: 24440723-24440723 on Build GRCh38
rs763780831 Chr.22: 24440652-24440652 on Build GRCh38
rs895401205 Chr.22: 24440511-24440511 on Build GRCh38
rs757137296 Chr.22: 24440656-24440656 on Build GRCh38
rs138352666 Chr.22: 24440934-24440934 on Build GRCh38
rs780504541 Chr.22: 24440837-24440838 on Build GRCh38
rs747653711 Chr.22: 24440935-24440935 on Build GRCh38
rs200688325 Chr.22: 24440602-24440602 on Build GRCh38
rs202036955 Chr.22: 24440793-24440793 on Build GRCh38
rs769798091 Chr.22: 24440577-24440577 on Build GRCh38
rs200446971 Chr.22: 24440800-24440800 on Build GRCh38
rs1056353441 Chr.22: 24440916-24440916 on Build GRCh38
rs745782306 Chr.22: 24440566-24440566 on Build GRCh38
rs377038706 Chr.22: 24440763-24440763 on Build GRCh38
rs78720419 Chr.22: 24440689-24440689 on Build GRCh38
rs202128195 Chr.22: 24440736-24440736 on Build GRCh38
rs776995788 Chr.22: 24440618-24440618 on Build GRCh38
rs80266635 Chr.22: 24440961-24440961 on Build GRCh38
rs77393092 Chr.22: 24440743-24440743 on Build GRCh38
rs201329533 Chr.22: 24440552-24440552 on Build GRCh38
rs756917857 Chr.22: 24440770-24440771 on Build GRCh38
rs200176420 Chr.22: 24440624-24440624 on Build GRCh38

View Full Product Details