Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 89259859-89260408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBP5
Entrez Gene ID
115362
Gene Name
guanylate binding protein 5
Gene Aliases
GBP-5
UniGene
Hs.513726
Gene Chromosome Location
Chr.1: 89258950-89272861 on Build GRCh38
Transcripts
NM_001134486.2 NM_052942.3

SNP Detail

SNP ID Chromosome Location
rs187030262 Chr.1: 89260005-89260005 on Build GRCh38
rs1003795757 Chr.1: 89260355-89260355 on Build GRCh38
rs777805177 Chr.1: 89259913-89259913 on Build GRCh38
rs670164 Chr.1: 89260084-89260084 on Build GRCh38
rs1018137505 Chr.1: 89259895-89259895 on Build GRCh38
rs963567048 Chr.1: 89259910-89259910 on Build GRCh38
rs1009505407 Chr.1: 89260112-89260112 on Build GRCh38
rs983341700 Chr.1: 89259974-89259974 on Build GRCh38
rs994891017 Chr.1: 89260187-89260187 on Build GRCh38
rs551664321 Chr.1: 89259888-89259889 on Build GRCh38
rs999082212 Chr.1: 89260073-89260073 on Build GRCh38
rs553761591 Chr.1: 89260093-89260093 on Build GRCh38
rs544238363 Chr.1: 89260306-89260306 on Build GRCh38
rs779401185 Chr.1: 89260295-89260295 on Build GRCh38
rs550668678 Chr.1: 89260359-89260359 on Build GRCh38
rs375400624 Chr.1: 89260162-89260162 on Build GRCh38
rs946478673 Chr.1: 89260027-89260027 on Build GRCh38
rs1027673150 Chr.1: 89260197-89260197 on Build GRCh38
rs1043382131 Chr.1: 89260064-89260064 on Build GRCh38
rs1031291496 Chr.1: 89260358-89260358 on Build GRCh38
rs1004588941 Chr.1: 89259885-89259885 on Build GRCh38
rs114461675 Chr.1: 89259994-89259994 on Build GRCh38
rs947221868 Chr.1: 89260085-89260085 on Build GRCh38
rs670184 Chr.1: 89260102-89260102 on Build GRCh38
rs573936912 Chr.1: 89260240-89260240 on Build GRCh38
rs573667453 Chr.1: 89260203-89260203 on Build GRCh38
rs542777749 Chr.1: 89260315-89260315 on Build GRCh38
rs746164247 Chr.1: 89260340-89260340 on Build GRCh38
rs950613826 Chr.1: 89259924-89259924 on Build GRCh38
rs1012854710 Chr.1: 89259972-89259972 on Build GRCh38
rs1043433327 Chr.1: 89260345-89260345 on Build GRCh38
rs916355033 Chr.1: 89260020-89260020 on Build GRCh38
rs927792569 Chr.1: 89259986-89259986 on Build GRCh38
rs890697208 Chr.1: 89260103-89260103 on Build GRCh38
rs147190134 Chr.1: 89260146-89260146 on Build GRCh38
rs902149353 Chr.1: 89260069-89260069 on Build GRCh38
rs916382730 Chr.1: 89260310-89260310 on Build GRCh38
rs1012012320 Chr.1: 89260296-89260296 on Build GRCh38
rs555748783 Chr.1: 89260149-89260149 on Build GRCh38
rs34523100 Chr.1: 89260066-89260066 on Build GRCh38
rs753946099 Chr.1: 89260008-89260008 on Build GRCh38
rs927889075 Chr.1: 89260293-89260293 on Build GRCh38
rs112340058 Chr.1: 89260294-89260294 on Build GRCh38
rs983437954 Chr.1: 89260267-89260267 on Build GRCh38
rs1053271590 Chr.1: 89260089-89260089 on Build GRCh38
rs556648186 Chr.1: 89260342-89260345 on Build GRCh38
rs921778653 Chr.1: 89260130-89260130 on Build GRCh38
rs565577 Chr.1: 89259899-89259899 on Build GRCh38
rs556091095 Chr.1: 89259897-89259897 on Build GRCh38
rs752956173 Chr.1: 89259889-89259889 on Build GRCh38
rs572979597 Chr.1: 89260114-89260114 on Build GRCh38

View Full Product Details