Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 92842835-92843351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM69A
Entrez Gene ID
388650
Gene Name
family with sequence similarity 69 member A
Gene Aliases
-
UniGene
Hs.180946
Gene Chromosome Location
Chr.1: 92832729-92961522 on Build GRCh38
Transcripts
NM_001006605.4 NM_001252269.1 NM_001252270.1 NM_001252271.1 NM_001252273.1

SNP Detail

SNP ID Chromosome Location
rs762574479 Chr.1: 92843270-92843270 on Build GRCh38
rs1003035741 Chr.1: 92842923-92842923 on Build GRCh38
rs767634071 Chr.1: 92843173-92843174 on Build GRCh38
rs945682714 Chr.1: 92843265-92843266 on Build GRCh38
rs1028121436 Chr.1: 92843096-92843096 on Build GRCh38
rs747170840 Chr.1: 92843004-92843004 on Build GRCh38
rs779230408 Chr.1: 92842927-92842927 on Build GRCh38
rs962479583 Chr.1: 92843022-92843022 on Build GRCh38
rs994034645 Chr.1: 92843319-92843319 on Build GRCh38
rs1030400782 Chr.1: 92843144-92843144 on Build GRCh38
rs2244496 Chr.1: 92843296-92843296 on Build GRCh38
rs758732647 Chr.1: 92842967-92842967 on Build GRCh38
rs1041522014 Chr.1: 92843267-92843267 on Build GRCh38
rs979212593 Chr.1: 92843161-92843161 on Build GRCh38
rs554970138 Chr.1: 92843145-92843145 on Build GRCh38
rs993590760 Chr.1: 92843072-92843072 on Build GRCh38
rs889343850 Chr.1: 92842998-92842998 on Build GRCh38
rs770390297 Chr.1: 92843027-92843028 on Build GRCh38
rs924985033 Chr.1: 92843199-92843199 on Build GRCh38
rs570577395 Chr.1: 92843002-92843002 on Build GRCh38
rs1016230196 Chr.1: 92843003-92843003 on Build GRCh38
rs72726483 Chr.1: 92843132-92843132 on Build GRCh38
rs1040978846 Chr.1: 92843283-92843283 on Build GRCh38
rs1055975693 Chr.1: 92843307-92843307 on Build GRCh38
rs913730731 Chr.1: 92843143-92843143 on Build GRCh38
rs542645972 Chr.1: 92843074-92843077 on Build GRCh38
rs141134097 Chr.1: 92842861-92842861 on Build GRCh38
rs755731722 Chr.1: 92843278-92843278 on Build GRCh38
rs574753905 Chr.1: 92843311-92843313 on Build GRCh38
rs932318141 Chr.1: 92843222-92843222 on Build GRCh38
rs1041472199 Chr.1: 92842874-92842874 on Build GRCh38
rs932322532 Chr.1: 92843223-92843223 on Build GRCh38
rs866804790 Chr.1: 92843269-92843269 on Build GRCh38
rs879386529 Chr.1: 92842994-92842994 on Build GRCh38
rs79082115 Chr.1: 92843311-92843311 on Build GRCh38
rs889395893 Chr.1: 92843266-92843266 on Build GRCh38
rs990045870 Chr.1: 92843261-92843261 on Build GRCh38
rs750704049 Chr.1: 92842957-92842957 on Build GRCh38
rs572722206 Chr.1: 92843101-92843101 on Build GRCh38
rs960597540 Chr.1: 92843136-92843136 on Build GRCh38
rs747812914 Chr.1: 92842938-92842938 on Build GRCh38
rs2244588 Chr.1: 92843241-92843241 on Build GRCh38
rs142261326 Chr.1: 92843309-92843309 on Build GRCh38
rs955418135 Chr.1: 92843156-92843156 on Build GRCh38
rs573650418 Chr.1: 92843021-92843021 on Build GRCh38
rs1035054815 Chr.1: 92842950-92842950 on Build GRCh38
rs780258784 Chr.1: 92842991-92842991 on Build GRCh38
rs914266729 Chr.1: 92843265-92843265 on Build GRCh38
rs945126691 Chr.1: 92842868-92842868 on Build GRCh38
rs1043821737 Chr.1: 92842918-92842918 on Build GRCh38
rs1050684648 Chr.1: 92842990-92842990 on Build GRCh38
rs556177828 Chr.1: 92843155-92843155 on Build GRCh38
rs565252580 Chr.1: 92843314-92843314 on Build GRCh38
rs183736361 Chr.1: 92843079-92843079 on Build GRCh38
rs903510498 Chr.1: 92842966-92842966 on Build GRCh38
rs945153237 Chr.1: 92843150-92843150 on Build GRCh38
rs1010010721 Chr.1: 92842862-92842863 on Build GRCh38
rs781190147 Chr.1: 92843077-92843077 on Build GRCh38
rs938392716 Chr.1: 92843299-92843299 on Build GRCh38
rs553081344 Chr.1: 92843098-92843098 on Build GRCh38
rs911013740 Chr.1: 92843162-92843162 on Build GRCh38
rs768469555 Chr.1: 92843031-92843031 on Build GRCh38
rs769650278 Chr.1: 92843139-92843139 on Build GRCh38

View Full Product Details