Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38479504-38480008 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDELR3
Entrez Gene ID
11015
Gene Name
KDEL endoplasmic reticulum protein retention receptor 3
Gene Aliases
ERD2L3
UniGene
Hs.745072
Gene Chromosome Location
Chr.22: 38468062-38483447 on Build GRCh38
Transcripts
NM_006855.3 NM_016657.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3992160 c.328A>G p.N110D Chr.22: 38479728-38479728 on Build GRCh38
4836181 c.202C>G p.L68V Chr.22: 38479602-38479602 on Build GRCh38
7322756 c.339C>A p.F113L Chr.22: 38479739-38479739 on Build GRCh38
3800200 c.195G>A p.V65V Chr.22: 38479595-38479595 on Build GRCh38
3842682 c.349G>C p.E117Q Chr.22: 38479749-38479749 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779947023 Chr.22: 38479797-38479797 on Build GRCh38
rs750373209 Chr.22: 38479922-38479922 on Build GRCh38
rs994597575 Chr.22: 38479876-38479876 on Build GRCh38
rs754478616 Chr.22: 38479577-38479577 on Build GRCh38
rs771470669 Chr.22: 38479573-38479574 on Build GRCh38
rs774317224 Chr.22: 38479623-38479623 on Build GRCh38
rs753518121 Chr.22: 38479767-38479767 on Build GRCh38
rs1046330270 Chr.22: 38479649-38479650 on Build GRCh38
rs762005807 Chr.22: 38479732-38479732 on Build GRCh38
rs567165767 Chr.22: 38479732-38479733 on Build GRCh38
rs775563815 Chr.22: 38479635-38479635 on Build GRCh38
rs778706793 Chr.22: 38479772-38479772 on Build GRCh38
rs997239713 Chr.22: 38479851-38479852 on Build GRCh38
rs775800725 Chr.22: 38479696-38479696 on Build GRCh38
rs765621536 Chr.22: 38479741-38479741 on Build GRCh38
rs138428820 Chr.22: 38479627-38479627 on Build GRCh38
rs898913858 Chr.22: 38479872-38479872 on Build GRCh38
rs767020631 Chr.22: 38479755-38479755 on Build GRCh38
rs766759831 Chr.22: 38479565-38479565 on Build GRCh38
rs189590805 Chr.22: 38479724-38479724 on Build GRCh38
rs752815424 Chr.22: 38479743-38479743 on Build GRCh38
rs778883688 Chr.22: 38479907-38479907 on Build GRCh38
rs763581658 Chr.22: 38479766-38479766 on Build GRCh38
rs768815947 Chr.22: 38479772-38479772 on Build GRCh38
rs747788768 Chr.22: 38479580-38479580 on Build GRCh38
rs181843947 Chr.22: 38479970-38479970 on Build GRCh38
rs981527360 Chr.22: 38479937-38479937 on Build GRCh38
rs1053959583 Chr.22: 38479685-38479685 on Build GRCh38
rs200575951 Chr.22: 38479801-38479801 on Build GRCh38
rs767615263 Chr.22: 38479692-38479692 on Build GRCh38
rs965824187 Chr.22: 38479606-38479606 on Build GRCh38
rs758283215 Chr.22: 38479795-38479795 on Build GRCh38
rs770721832 Chr.22: 38479615-38479615 on Build GRCh38
rs879201942 Chr.22: 38479759-38479759 on Build GRCh38
rs957415042 Chr.22: 38479879-38479879 on Build GRCh38
rs1007619701 Chr.22: 38479746-38479746 on Build GRCh38
rs1001984096 Chr.22: 38479534-38479534 on Build GRCh38
rs401933 Chr.22: 38479544-38479544 on Build GRCh38
rs146375082 Chr.22: 38479596-38479596 on Build GRCh38
rs912601645 Chr.22: 38479605-38479605 on Build GRCh38
rs778634923 Chr.22: 38479690-38479690 on Build GRCh38
rs577967921 Chr.22: 38479962-38479962 on Build GRCh38
rs372505402 Chr.22: 38479609-38479609 on Build GRCh38
rs752256020 Chr.22: 38479665-38479665 on Build GRCh38
rs1020802245 Chr.22: 38479854-38479854 on Build GRCh38
rs372780986 Chr.22: 38479754-38479754 on Build GRCh38
rs781490519 Chr.22: 38479614-38479615 on Build GRCh38
rs368527543 Chr.22: 38479592-38479592 on Build GRCh38
rs1002292417 Chr.22: 38479790-38479790 on Build GRCh38
rs747115395 Chr.22: 38479717-38479717 on Build GRCh38
rs553218030 Chr.22: 38479956-38479956 on Build GRCh38
rs745695546 Chr.22: 38479776-38479776 on Build GRCh38
rs569201267 Chr.22: 38479680-38479680 on Build GRCh38
rs950635869 Chr.22: 38479914-38479914 on Build GRCh38
rs966624251 Chr.22: 38479587-38479588 on Build GRCh38
rs141148010 Chr.22: 38479659-38479659 on Build GRCh38
rs201618436 Chr.22: 38479730-38479730 on Build GRCh38
rs919440180 Chr.22: 38479950-38479950 on Build GRCh38
rs369236434 Chr.22: 38479637-38479637 on Build GRCh38
rs755785070 Chr.22: 38479673-38479673 on Build GRCh38
rs757666985 Chr.22: 38479564-38479564 on Build GRCh38
rs201816064 Chr.22: 38479681-38479681 on Build GRCh38
rs991893689 Chr.22: 38479888-38479888 on Build GRCh38
rs573857697 Chr.22: 38479843-38479843 on Build GRCh38
rs184487322 Chr.22: 38479531-38479531 on Build GRCh38
rs755660288 Chr.22: 38479761-38479761 on Build GRCh38
rs754105116 Chr.22: 38479647-38479647 on Build GRCh38
rs369081501 Chr.22: 38479575-38479575 on Build GRCh38
rs752202908 Chr.22: 38479758-38479758 on Build GRCh38
rs1032245347 Chr.22: 38479878-38479878 on Build GRCh38
rs375590658 Chr.22: 38479624-38479624 on Build GRCh38
rs772002860 Chr.22: 38479703-38479703 on Build GRCh38
rs950647197 Chr.22: 38479584-38479584 on Build GRCh38
rs764331396 Chr.22: 38479645-38479645 on Build GRCh38
rs528692609 Chr.22: 38479579-38479579 on Build GRCh38
rs867335980 Chr.22: 38479734-38479734 on Build GRCh38
rs746369189 Chr.22: 38479654-38479655 on Build GRCh38
rs754235418 Chr.22: 38479552-38479552 on Build GRCh38
rs773461810 Chr.22: 38479750-38479750 on Build GRCh38
rs745764222 Chr.22: 38479702-38479702 on Build GRCh38
rs1020018391 Chr.22: 38479934-38479934 on Build GRCh38
rs371405125 Chr.22: 38479955-38479955 on Build GRCh38
rs756878248 Chr.22: 38479770-38479770 on Build GRCh38
rs748937233 Chr.22: 38479683-38479683 on Build GRCh38
rs147788461 Chr.22: 38479648-38479648 on Build GRCh38
rs775508451 Chr.22: 38479711-38479711 on Build GRCh38
rs750935487 Chr.22: 38479655-38479655 on Build GRCh38
rs915882033 Chr.22: 38479904-38479904 on Build GRCh38
rs776879564 Chr.22: 38479725-38479725 on Build GRCh38
rs966508724 Chr.22: 38479862-38479862 on Build GRCh38
rs763938623 Chr.22: 38479530-38479530 on Build GRCh38
rs914122008 Chr.22: 38479718-38479718 on Build GRCh38

View Full Product Details