Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 14490595-14491137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIP2
Entrez Gene ID
80852
Gene Name
glutamate receptor interacting protein 2
Gene Aliases
-
UniGene
Hs.517819
Gene Chromosome Location
Chr.3: 14489111-14602996 on Build GRCh38
Transcripts
NM_001080423.3

SNP Detail

SNP ID Chromosome Location
rs372613541 Chr.3: 14490652-14490652 on Build GRCh38
rs780584588 Chr.3: 14490927-14490927 on Build GRCh38
rs563989670 Chr.3: 14490734-14490734 on Build GRCh38
rs369572184 Chr.3: 14490729-14490729 on Build GRCh38
rs1042461055 Chr.3: 14490912-14490912 on Build GRCh38
rs997917970 Chr.3: 14490735-14490735 on Build GRCh38
rs1050893551 Chr.3: 14490972-14490972 on Build GRCh38
rs564475870 Chr.3: 14490680-14490680 on Build GRCh38
rs932734223 Chr.3: 14490722-14490722 on Build GRCh38
rs923608243 Chr.3: 14490957-14490957 on Build GRCh38
rs919747257 Chr.3: 14490968-14490968 on Build GRCh38
rs565875143 Chr.3: 14490869-14490869 on Build GRCh38
rs75866505 Chr.3: 14490991-14490991 on Build GRCh38
rs1049819374 Chr.3: 14490744-14490744 on Build GRCh38
rs974421896 Chr.3: 14490967-14490967 on Build GRCh38
rs145900754 Chr.3: 14490835-14490835 on Build GRCh38
rs891098520 Chr.3: 14490746-14490746 on Build GRCh38
rs892045308 Chr.3: 14490775-14490775 on Build GRCh38
rs1006767744 Chr.3: 14490996-14490996 on Build GRCh38
rs898421466 Chr.3: 14491056-14491056 on Build GRCh38
rs373024808 Chr.3: 14490788-14490788 on Build GRCh38
rs75851328 Chr.3: 14490638-14490638 on Build GRCh38
rs1015304958 Chr.3: 14490901-14490901 on Build GRCh38
rs553787325 Chr.3: 14491078-14491078 on Build GRCh38
rs967816181 Chr.3: 14490631-14490631 on Build GRCh38
rs551037409 Chr.3: 14490628-14490628 on Build GRCh38
rs750756363 Chr.3: 14490836-14490836 on Build GRCh38
rs1041998005 Chr.3: 14490697-14490697 on Build GRCh38
rs191786239 Chr.3: 14490888-14490888 on Build GRCh38
rs114399383 Chr.3: 14490644-14490644 on Build GRCh38
rs928017919 Chr.3: 14490844-14490844 on Build GRCh38
rs376488059 Chr.3: 14490683-14490683 on Build GRCh38
rs956173352 Chr.3: 14490625-14490625 on Build GRCh38
rs570521935 Chr.3: 14490892-14490892 on Build GRCh38
rs994054969 Chr.3: 14491062-14491062 on Build GRCh38
rs1021780103 Chr.3: 14490819-14490819 on Build GRCh38
rs368530001 Chr.3: 14490881-14490881 on Build GRCh38
rs980956058 Chr.3: 14490635-14490635 on Build GRCh38
rs962616816 Chr.3: 14490816-14490816 on Build GRCh38
rs992045638 Chr.3: 14490645-14490645 on Build GRCh38
rs756453818 Chr.3: 14490896-14490896 on Build GRCh38
rs76221136 Chr.3: 14491001-14491001 on Build GRCh38
rs535485098 Chr.3: 14491055-14491055 on Build GRCh38
rs939974099 Chr.3: 14490675-14490675 on Build GRCh38
rs912648010 Chr.3: 14490994-14490994 on Build GRCh38
rs754525005 Chr.3: 14491074-14491074 on Build GRCh38
rs569964023 Chr.3: 14491018-14491018 on Build GRCh38
rs1011747583 Chr.3: 14490627-14490627 on Build GRCh38
rs1053260063 Chr.3: 14490764-14490764 on Build GRCh38
rs764224931 Chr.3: 14490660-14490660 on Build GRCh38
rs889651478 Chr.3: 14490761-14490761 on Build GRCh38
rs1033439772 Chr.3: 14490843-14490843 on Build GRCh38
rs529275623 Chr.3: 14490759-14490759 on Build GRCh38
rs762796837 Chr.3: 14490976-14490976 on Build GRCh38
rs35854097 Chr.3: 14490720-14490720 on Build GRCh38
rs541652668 Chr.3: 14490655-14490655 on Build GRCh38
rs938282787 Chr.3: 14490649-14490649 on Build GRCh38
rs994841359 Chr.3: 14490822-14490822 on Build GRCh38

View Full Product Details