Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 14493179-14493681 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIP2
Entrez Gene ID
80852
Gene Name
glutamate receptor interacting protein 2
Gene Aliases
-
UniGene
Hs.517819
Gene Chromosome Location
Chr.3: 14489111-14602996 on Build GRCh38
Transcripts
NM_001080423.3 XM_011534141.2 XM_011534142.2 XM_011534143.2 XM_011534145.2 XM_011534146.2

SNP Detail

SNP ID Chromosome Location
rs779700342 Chr.3: 14493652-14493652 on Build GRCh38
rs923299985 Chr.3: 14493437-14493437 on Build GRCh38
rs758050575 Chr.3: 14493648-14493648 on Build GRCh38
rs147230097 Chr.3: 14493267-14493267 on Build GRCh38
rs997753266 Chr.3: 14493214-14493214 on Build GRCh38
rs935653857 Chr.3: 14493308-14493308 on Build GRCh38
rs933563067 Chr.3: 14493446-14493446 on Build GRCh38
rs61547342 Chr.3: 14493514-14493514 on Build GRCh38
rs781148651 Chr.3: 14493658-14493658 on Build GRCh38
rs531705591 Chr.3: 14493631-14493631 on Build GRCh38
rs144434618 Chr.3: 14493519-14493519 on Build GRCh38
rs778444899 Chr.3: 14493630-14493630 on Build GRCh38
rs917499227 Chr.3: 14493266-14493266 on Build GRCh38
rs892883711 Chr.3: 14493476-14493476 on Build GRCh38
rs535822341 Chr.3: 14493505-14493505 on Build GRCh38
rs984854145 Chr.3: 14493380-14493380 on Build GRCh38
rs767972191 Chr.3: 14493620-14493620 on Build GRCh38
rs769624099 Chr.3: 14493604-14493604 on Build GRCh38
rs73031847 Chr.3: 14493539-14493539 on Build GRCh38
rs931789421 Chr.3: 14493578-14493578 on Build GRCh38
rs530471361 Chr.3: 14493409-14493409 on Build GRCh38
rs1043043400 Chr.3: 14493449-14493449 on Build GRCh38
rs887687951 Chr.3: 14493585-14493585 on Build GRCh38
rs909298663 Chr.3: 14493401-14493401 on Build GRCh38
rs116409960 Chr.3: 14493208-14493208 on Build GRCh38
rs1040197523 Chr.3: 14493351-14493351 on Build GRCh38
rs891539283 Chr.3: 14493314-14493314 on Build GRCh38
rs144232410 Chr.3: 14493453-14493453 on Build GRCh38
rs1052909660 Chr.3: 14493309-14493309 on Build GRCh38
rs991149968 Chr.3: 14493304-14493304 on Build GRCh38
rs554961383 Chr.3: 14493550-14493550 on Build GRCh38
rs768451044 Chr.3: 14493657-14493657 on Build GRCh38
rs1021105854 Chr.3: 14493221-14493221 on Build GRCh38
rs765819360 Chr.3: 14493272-14493272 on Build GRCh38
rs952925362 Chr.3: 14493238-14493238 on Build GRCh38
rs950703655 Chr.3: 14493586-14493586 on Build GRCh38
rs1022670736 Chr.3: 14493325-14493325 on Build GRCh38
rs961719012 Chr.3: 14493447-14493447 on Build GRCh38
rs548713934 Chr.3: 14493248-14493248 on Build GRCh38
rs756790766 Chr.3: 14493532-14493532 on Build GRCh38
rs750031591 Chr.3: 14493638-14493638 on Build GRCh38
rs745610042 Chr.3: 14493565-14493565 on Build GRCh38
rs902098469 Chr.3: 14493213-14493213 on Build GRCh38
rs565011324 Chr.3: 14493358-14493358 on Build GRCh38
rs559450764 Chr.3: 14493332-14493332 on Build GRCh38
rs908606836 Chr.3: 14493246-14493246 on Build GRCh38
rs1001441708 Chr.3: 14493457-14493457 on Build GRCh38
rs964748574 Chr.3: 14493649-14493649 on Build GRCh38
rs140610749 Chr.3: 14493285-14493285 on Build GRCh38
rs929140755 Chr.3: 14493379-14493379 on Build GRCh38
rs1024398430 Chr.3: 14493459-14493459 on Build GRCh38
rs189495110 Chr.3: 14493414-14493414 on Build GRCh38
rs753296572 Chr.3: 14493291-14493291 on Build GRCh38
rs567019189 Chr.3: 14493500-14493500 on Build GRCh38
rs561422681 Chr.3: 14493352-14493352 on Build GRCh38
rs528393288 Chr.3: 14493342-14493342 on Build GRCh38
rs369914186 Chr.3: 14493621-14493621 on Build GRCh38
rs546814739 Chr.3: 14493467-14493467 on Build GRCh38
rs754115407 Chr.3: 14493240-14493240 on Build GRCh38
rs373836618 Chr.3: 14493653-14493653 on Build GRCh38
rs756453422 Chr.3: 14493628-14493628 on Build GRCh38
rs1015974348 Chr.3: 14493645-14493645 on Build GRCh38
rs1049040623 Chr.3: 14493579-14493579 on Build GRCh38
rs944410725 Chr.3: 14493560-14493560 on Build GRCh38
rs1027553727 Chr.3: 14493391-14493391 on Build GRCh38
rs567034577 Chr.3: 14493436-14493436 on Build GRCh38

View Full Product Details