Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15223344-15223872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN7
Entrez Gene ID
23473
Gene Name
calpain 7
Gene Aliases
CALPAIN7, PALBH
UniGene
Hs.595234
Gene Chromosome Location
Chr.3: 15206226-15252916 on Build GRCh38
Transcripts
NM_014296.2 XM_011533555.2 XM_011533556.2 XM_011533557.2 XM_017006043.1 XM_017006044.1 XM_017006045.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1247326 c.647C>G p.S216* Chr.3: 15223483-15223483 on Build GRCh38
3588918 c.673C>T p.P225S Chr.3: 15223509-15223509 on Build GRCh38
1040092 c.697G>T p.E233* Chr.3: 15223533-15223533 on Build GRCh38
5025901 c.715A>G p.M239V Chr.3: 15223551-15223551 on Build GRCh38
1040093 c.700C>T p.R234C Chr.3: 15223536-15223536 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1038016816 Chr.3: 15223611-15223611 on Build GRCh38
rs773192523 Chr.3: 15223490-15223490 on Build GRCh38
rs201692968 Chr.3: 15223481-15223481 on Build GRCh38
rs987160827 Chr.3: 15223815-15223815 on Build GRCh38
rs538264446 Chr.3: 15223658-15223658 on Build GRCh38
rs756110533 Chr.3: 15223457-15223457 on Build GRCh38
rs746176017 Chr.3: 15223471-15223471 on Build GRCh38
rs1031625209 Chr.3: 15223789-15223789 on Build GRCh38
rs199866982 Chr.3: 15223460-15223460 on Build GRCh38
rs770806737 Chr.3: 15223385-15223385 on Build GRCh38
rs1057041445 Chr.3: 15223657-15223657 on Build GRCh38
rs1021590181 Chr.3: 15223830-15223830 on Build GRCh38
rs760863604 Chr.3: 15223550-15223550 on Build GRCh38
rs1008519777 Chr.3: 15223716-15223716 on Build GRCh38
rs762875536 Chr.3: 15223499-15223499 on Build GRCh38
rs926418492 Chr.3: 15223523-15223523 on Build GRCh38
rs1000120044 Chr.3: 15223779-15223779 on Build GRCh38
rs753403699 Chr.3: 15223805-15223805 on Build GRCh38
rs955617487 Chr.3: 15223417-15223417 on Build GRCh38
rs114148981 Chr.3: 15223464-15223464 on Build GRCh38
rs183150729 Chr.3: 15223818-15223818 on Build GRCh38
rs17040876 Chr.3: 15223581-15223581 on Build GRCh38
rs774149066 Chr.3: 15223519-15223519 on Build GRCh38
rs764072733 Chr.3: 15223551-15223551 on Build GRCh38
rs766730519 Chr.3: 15223695-15223695 on Build GRCh38
rs538852855 Chr.3: 15223564-15223564 on Build GRCh38
rs780948356 Chr.3: 15223587-15223587 on Build GRCh38
rs752834980 Chr.3: 15223538-15223538 on Build GRCh38
rs779421617 Chr.3: 15223465-15223465 on Build GRCh38
rs767816389 Chr.3: 15223537-15223537 on Build GRCh38
rs368395737 Chr.3: 15223565-15223565 on Build GRCh38
rs113192849 Chr.3: 15223633-15223633 on Build GRCh38
rs767579784 Chr.3: 15223433-15223433 on Build GRCh38
rs892026576 Chr.3: 15223793-15223793 on Build GRCh38
rs759762285 Chr.3: 15223428-15223428 on Build GRCh38
rs766150686 Chr.3: 15223424-15223424 on Build GRCh38
rs932385953 Chr.3: 15223676-15223676 on Build GRCh38
rs754379549 Chr.3: 15223463-15223463 on Build GRCh38
rs887118439 Chr.3: 15223645-15223645 on Build GRCh38
rs1031917646 Chr.3: 15223777-15223777 on Build GRCh38
rs769166335 Chr.3: 15223484-15223484 on Build GRCh38
rs752847829 Chr.3: 15223449-15223449 on Build GRCh38
rs375269916 Chr.3: 15223586-15223586 on Build GRCh38
rs747743157 Chr.3: 15223589-15223589 on Build GRCh38
rs757850285 Chr.3: 15223575-15223575 on Build GRCh38
rs780976936 Chr.3: 15223453-15223454 on Build GRCh38
rs891299313 Chr.3: 15223708-15223708 on Build GRCh38
rs746839406 Chr.3: 15223375-15223375 on Build GRCh38
rs759205888 Chr.3: 15223534-15223534 on Build GRCh38
rs554973126 Chr.3: 15223679-15223679 on Build GRCh38
rs940059594 Chr.3: 15223656-15223659 on Build GRCh38
rs770789118 Chr.3: 15223504-15223504 on Build GRCh38
rs964832964 Chr.3: 15223819-15223819 on Build GRCh38
rs140164718 Chr.3: 15223478-15223478 on Build GRCh38
rs773516235 Chr.3: 15223606-15223606 on Build GRCh38
rs765788642 Chr.3: 15223579-15223579 on Build GRCh38
rs566674844 Chr.3: 15223479-15223479 on Build GRCh38
rs745789424 Chr.3: 15223750-15223750 on Build GRCh38
rs370780259 Chr.3: 15223426-15223426 on Build GRCh38

View Full Product Details