Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 101025583-101026081 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPH5
Entrez Gene ID
51611
Gene Name
diphthamide biosynthesis 5
Gene Aliases
AD-018, CGI-30, HSPC143, NPD015
UniGene
Hs.440776
Gene Chromosome Location
Chr.1: 100989623-101025806 on Build GRCh38
Transcripts
NM_001077394.1 NM_001077395.1 NM_015958.2 XM_005270938.1 XM_005270939.4 XM_005270941.3 XM_006710683.3 XM_011541569.1 XM_011541570.2 XM_017001460.1 XM_017001461.1 XM_017001462.1 NR_109849.1

Gene Symbol
LOC102606465
Entrez Gene ID
102606465
Gene Name
uncharacterized LOC102606465
Gene Aliases
-
UniGene
Hs.593669
Gene Chromosome Location
Chr.1: 101025846-101087379 on Build GRCh38
Transcripts
NM_001077394.1 NM_001077395.1 NM_015958.2 XM_005270938.1 XM_005270939.4 XM_005270941.3 XM_006710683.3 XM_011541569.1 XM_011541570.2 XM_017001460.1 XM_017001461.1 XM_017001462.1 NR_109849.1

SNP Detail

SNP ID Chromosome Location
rs949422978 Chr.1: 101025951-101025951 on Build GRCh38
rs893680176 Chr.1: 101025886-101025886 on Build GRCh38
rs776576297 Chr.1: 101025934-101025934 on Build GRCh38
rs1055233665 Chr.1: 101025945-101025945 on Build GRCh38
rs1015940196 Chr.1: 101025841-101025841 on Build GRCh38
rs184857255 Chr.1: 101025784-101025784 on Build GRCh38
rs889634765 Chr.1: 101025987-101025987 on Build GRCh38
rs556107158 Chr.1: 101025814-101025814 on Build GRCh38
rs943315957 Chr.1: 101025883-101025883 on Build GRCh38
rs961903365 Chr.1: 101025922-101025922 on Build GRCh38
rs934041882 Chr.1: 101025862-101025862 on Build GRCh38
rs965116129 Chr.1: 101026061-101026061 on Build GRCh38
rs981760310 Chr.1: 101025762-101025762 on Build GRCh38
rs998192985 Chr.1: 101025638-101025638 on Build GRCh38
rs201869101 Chr.1: 101025665-101025665 on Build GRCh38
rs922696422 Chr.1: 101025847-101025847 on Build GRCh38
rs1006647318 Chr.1: 101026005-101026005 on Build GRCh38
rs545389832 Chr.1: 101026003-101026003 on Build GRCh38
rs906315240 Chr.1: 101025891-101025891 on Build GRCh38
rs896511435 Chr.1: 101025808-101025808 on Build GRCh38
rs12090113 Chr.1: 101025658-101025658 on Build GRCh38
rs534656062 Chr.1: 101025936-101025936 on Build GRCh38
rs985199956 Chr.1: 101025890-101025890 on Build GRCh38
rs1026808239 Chr.1: 101025885-101025885 on Build GRCh38
rs1053706848 Chr.1: 101025884-101025884 on Build GRCh38
rs144025500 Chr.1: 101025872-101025872 on Build GRCh38
rs189277330 Chr.1: 101025918-101025918 on Build GRCh38
rs1055205530 Chr.1: 101025802-101025802 on Build GRCh38
rs984380645 Chr.1: 101025776-101025776 on Build GRCh38
rs938145340 Chr.1: 101025938-101025938 on Build GRCh38
rs959026642 Chr.1: 101025915-101025915 on Build GRCh38
rs568196177 Chr.1: 101025682-101025682 on Build GRCh38
rs189941207 Chr.1: 101026039-101026039 on Build GRCh38
rs867781655 Chr.1: 101025772-101025772 on Build GRCh38
rs911264900 Chr.1: 101025978-101025978 on Build GRCh38
rs112985750 Chr.1: 101025650-101025650 on Build GRCh38
rs1003233106 Chr.1: 101025897-101025897 on Build GRCh38
rs184235661 Chr.1: 101026025-101026025 on Build GRCh38
rs61742627 Chr.1: 101025958-101025958 on Build GRCh38
rs547648241 Chr.1: 101025736-101025736 on Build GRCh38
rs2281828 Chr.1: 101025709-101025709 on Build GRCh38
rs538075064 Chr.1: 101025763-101025763 on Build GRCh38
rs574642835 Chr.1: 101025919-101025919 on Build GRCh38
rs6693101 Chr.1: 101025901-101025901 on Build GRCh38
rs896544021 Chr.1: 101025946-101025946 on Build GRCh38
rs973280221 Chr.1: 101025829-101025829 on Build GRCh38
rs910077904 Chr.1: 101025790-101025790 on Build GRCh38
rs1009415719 Chr.1: 101025887-101025887 on Build GRCh38
rs879002991 Chr.1: 101025607-101025607 on Build GRCh38
rs1007463157 Chr.1: 101025692-101025692 on Build GRCh38
rs867740756 Chr.1: 101025948-101025948 on Build GRCh38
rs893741183 Chr.1: 101026021-101026021 on Build GRCh38
rs537504657 Chr.1: 101025909-101025909 on Build GRCh38
rs181024251 Chr.1: 101025944-101025944 on Build GRCh38
rs1021182395 Chr.1: 101025888-101025888 on Build GRCh38
rs769272309 Chr.1: 101025724-101025724 on Build GRCh38
rs541766979 Chr.1: 101025980-101025980 on Build GRCh38
rs914127985 Chr.1: 101025753-101025753 on Build GRCh38
rs34799907 Chr.1: 101025969-101025970 on Build GRCh38
rs553176353 Chr.1: 101025952-101025952 on Build GRCh38
rs985535688 Chr.1: 101025961-101025961 on Build GRCh38
rs773901355 Chr.1: 101025755-101025755 on Build GRCh38
rs565958255 Chr.1: 101025771-101025771 on Build GRCh38
rs957888900 Chr.1: 101025719-101025719 on Build GRCh38
rs772957874 Chr.1: 101025900-101025900 on Build GRCh38
rs1049782317 Chr.1: 101025869-101025869 on Build GRCh38
rs1054835085 Chr.1: 101026006-101026006 on Build GRCh38

View Full Product Details