Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 42692986-42693483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HHATL
Entrez Gene ID
57467
Gene Name
hedgehog acyltransferase-like
Gene Aliases
C3orf3, GUP1, MBOAT3, MSTP002, OACT3
UniGene
Hs.476041
Gene Chromosome Location
Chr.3: 42692663-42702849 on Build GRCh38
Transcripts
NM_020707.3 NR_027753.1 XM_006713274.2 XM_006713275.2 XM_011533969.2 XM_011533970.2 XM_017006935.1 XM_017006936.1 XM_017006937.1 XM_017006938.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3729236 c.1376G>A p.R459H Chr.3: 42693091-42693091 on Build GRCh38
4979251 c.1283G>A p.R428Q Chr.3: 42693184-42693184 on Build GRCh38
5497706 c.1333G>A p.V445M Chr.3: 42693134-42693134 on Build GRCh38
3594077 c.1286C>T p.A429V Chr.3: 42693181-42693181 on Build GRCh38
5399362 c.1302G>A p.M434I Chr.3: 42693165-42693165 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749756603 Chr.3: 42693108-42693109 on Build GRCh38
rs946780925 Chr.3: 42693315-42693315 on Build GRCh38
rs926596759 Chr.3: 42693331-42693331 on Build GRCh38
rs756401064 Chr.3: 42693066-42693067 on Build GRCh38
rs557543678 Chr.3: 42693211-42693211 on Build GRCh38
rs373396126 Chr.3: 42693059-42693059 on Build GRCh38
rs776300731 Chr.3: 42693065-42693065 on Build GRCh38
rs767924380 Chr.3: 42693048-42693048 on Build GRCh38
rs150455507 Chr.3: 42693095-42693095 on Build GRCh38
rs542446946 Chr.3: 42693336-42693336 on Build GRCh38
rs572206736 Chr.3: 42693045-42693045 on Build GRCh38
rs761502352 Chr.3: 42693089-42693089 on Build GRCh38
rs988531624 Chr.3: 42693296-42693296 on Build GRCh38
rs758799639 Chr.3: 42693243-42693243 on Build GRCh38
rs559136325 Chr.3: 42693347-42693347 on Build GRCh38
rs753887214 Chr.3: 42693225-42693225 on Build GRCh38
rs746392510 Chr.3: 42693044-42693044 on Build GRCh38
rs764293071 Chr.3: 42693224-42693224 on Build GRCh38
rs761776371 Chr.3: 42693316-42693316 on Build GRCh38
rs992460162 Chr.3: 42693209-42693209 on Build GRCh38
rs747532998 Chr.3: 42693247-42693247 on Build GRCh38
rs756770176 Chr.3: 42693233-42693233 on Build GRCh38
rs980801134 Chr.3: 42693244-42693244 on Build GRCh38
rs975284995 Chr.3: 42693232-42693232 on Build GRCh38
rs568800153 Chr.3: 42693015-42693015 on Build GRCh38
rs776066700 Chr.3: 42693194-42693194 on Build GRCh38
rs778459338 Chr.3: 42693263-42693278 on Build GRCh38
rs200263986 Chr.3: 42693184-42693184 on Build GRCh38
rs772900005 Chr.3: 42693201-42693201 on Build GRCh38
rs763778428 Chr.3: 42693127-42693127 on Build GRCh38
rs750604620 Chr.3: 42693240-42693240 on Build GRCh38
rs1056662803 Chr.3: 42693092-42693092 on Build GRCh38
rs760234905 Chr.3: 42693056-42693056 on Build GRCh38
rs766926218 Chr.3: 42693310-42693310 on Build GRCh38
rs779253437 Chr.3: 42693229-42693229 on Build GRCh38
rs147953207 Chr.3: 42693094-42693094 on Build GRCh38
rs756329889 Chr.3: 42693105-42693105 on Build GRCh38
rs567601087 Chr.3: 42693335-42693335 on Build GRCh38
rs113952517 Chr.3: 42693374-42693374 on Build GRCh38
rs929470785 Chr.3: 42693354-42693354 on Build GRCh38
rs933225334 Chr.3: 42693321-42693321 on Build GRCh38
rs768201980 Chr.3: 42693193-42693193 on Build GRCh38
rs879083361 Chr.3: 42693319-42693319 on Build GRCh38
rs754281929 Chr.3: 42693318-42693318 on Build GRCh38
rs772411332 Chr.3: 42693180-42693180 on Build GRCh38
rs768452390 Chr.3: 42693252-42693252 on Build GRCh38
rs189346366 Chr.3: 42693255-42693255 on Build GRCh38
rs1014219274 Chr.3: 42693460-42693460 on Build GRCh38
rs747479497 Chr.3: 42693185-42693185 on Build GRCh38
rs895514735 Chr.3: 42693451-42693451 on Build GRCh38
rs779148223 Chr.3: 42693140-42693140 on Build GRCh38
rs892366147 Chr.3: 42693021-42693021 on Build GRCh38
rs759525991 Chr.3: 42693223-42693223 on Build GRCh38
rs774994838 Chr.3: 42693051-42693051 on Build GRCh38
rs746325431 Chr.3: 42693145-42693145 on Build GRCh38
rs778178815 Chr.3: 42693067-42693067 on Build GRCh38
rs35468649 Chr.3: 42693346-42693346 on Build GRCh38
rs886895278 Chr.3: 42693413-42693413 on Build GRCh38
rs1037425755 Chr.3: 42693346-42693346 on Build GRCh38
rs762867804 Chr.3: 42693207-42693207 on Build GRCh38
rs74853274 Chr.3: 42693258-42693258 on Build GRCh38
rs987958403 Chr.3: 42693276-42693276 on Build GRCh38
rs376754935 Chr.3: 42693246-42693246 on Build GRCh38
rs751587988 Chr.3: 42693217-42693217 on Build GRCh38
rs774758083 Chr.3: 42693037-42693037 on Build GRCh38
rs373774262 Chr.3: 42693141-42693141 on Build GRCh38
rs1043708073 Chr.3: 42693022-42693022 on Build GRCh38
rs1026395079 Chr.3: 42693377-42693377 on Build GRCh38
rs757458637 Chr.3: 42693129-42693129 on Build GRCh38
rs368835249 Chr.3: 42693195-42693195 on Build GRCh38
rs563642995 Chr.3: 42693317-42693317 on Build GRCh38
rs148117407 Chr.3: 42693371-42693371 on Build GRCh38
rs372425768 Chr.3: 42693196-42693196 on Build GRCh38
rs771242961 Chr.3: 42693031-42693031 on Build GRCh38
rs753069063 Chr.3: 42693056-42693057 on Build GRCh38
rs764961715 Chr.3: 42693091-42693091 on Build GRCh38

View Full Product Details