Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 47511009-47511540 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELP6
Entrez Gene ID
54859
Gene Name
elongator acetyltransferase complex subunit 6
Gene Aliases
C3orf75, TMEM103
UniGene
Hs.311100
Gene Chromosome Location
Chr.3: 47495640-47513711 on Build GRCh38
Transcripts
NM_001031703.2 XM_005265241.3 XM_011533851.2 XM_011533852.2 XM_011533853.2 XM_011533854.2 XM_011533856.2 XM_011533857.2 XM_017006669.1 XM_017006670.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4407228 c.133+3A>G p.? Chr.3: 47511145-47511145 on Build GRCh38
1045630 c.133+1G>A p.? Chr.3: 47511147-47511147 on Build GRCh38
7131827 c.91G>T p.G31W Chr.3: 47511190-47511190 on Build GRCh38
4913094 c.81C>A p.A27A Chr.3: 47511200-47511200 on Build GRCh38
1246843 c.92G>T p.G31V Chr.3: 47511189-47511189 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776110688 Chr.3: 47511394-47511394 on Build GRCh38
rs773523949 Chr.3: 47511104-47511104 on Build GRCh38
rs749619300 Chr.3: 47511161-47511161 on Build GRCh38
rs200181192 Chr.3: 47511193-47511193 on Build GRCh38
rs368475325 Chr.3: 47511152-47511152 on Build GRCh38
rs371037952 Chr.3: 47511185-47511185 on Build GRCh38
rs938750452 Chr.3: 47511329-47511329 on Build GRCh38
rs999132186 Chr.3: 47511501-47511501 on Build GRCh38
rs935684728 Chr.3: 47511060-47511060 on Build GRCh38
rs575192064 Chr.3: 47511286-47511286 on Build GRCh38
rs771038790 Chr.3: 47511113-47511113 on Build GRCh38
rs182677594 Chr.3: 47511493-47511493 on Build GRCh38
rs772419197 Chr.3: 47511162-47511163 on Build GRCh38
rs903435117 Chr.3: 47511454-47511454 on Build GRCh38
rs954468131 Chr.3: 47511039-47511039 on Build GRCh38
rs768587716 Chr.3: 47511203-47511203 on Build GRCh38
rs932815635 Chr.3: 47511096-47511096 on Build GRCh38
rs753195680 Chr.3: 47511145-47511145 on Build GRCh38
rs927814707 Chr.3: 47511314-47511314 on Build GRCh38
rs775636498 Chr.3: 47511195-47511195 on Build GRCh38
rs995507935 Chr.3: 47511262-47511262 on Build GRCh38
rs201970071 Chr.3: 47511208-47511208 on Build GRCh38
rs761602538 Chr.3: 47511225-47511225 on Build GRCh38
rs745387307 Chr.3: 47511181-47511181 on Build GRCh38
rs924526518 Chr.3: 47511475-47511475 on Build GRCh38
rs751641610 Chr.3: 47511124-47511124 on Build GRCh38
rs894435386 Chr.3: 47511341-47511341 on Build GRCh38
rs1043115096 Chr.3: 47511407-47511407 on Build GRCh38
rs760865819 Chr.3: 47511496-47511496 on Build GRCh38
rs746218722 Chr.3: 47511157-47511159 on Build GRCh38
rs766522243 Chr.3: 47511231-47511231 on Build GRCh38
rs991316778 Chr.3: 47511366-47511366 on Build GRCh38
rs5022852 Chr.3: 47511374-47511374 on Build GRCh38
rs748837352 Chr.3: 47511092-47511092 on Build GRCh38
rs768067320 Chr.3: 47511135-47511135 on Build GRCh38
rs778563813 Chr.3: 47511177-47511177 on Build GRCh38
rs143211714 Chr.3: 47511267-47511267 on Build GRCh38
rs1056301656 Chr.3: 47511336-47511336 on Build GRCh38
rs756894975 Chr.3: 47511164-47511164 on Build GRCh38
rs563948749 Chr.3: 47511305-47511305 on Build GRCh38
rs539973873 Chr.3: 47511360-47511360 on Build GRCh38
rs545545852 Chr.3: 47511303-47511303 on Build GRCh38
rs1024165342 Chr.3: 47511375-47511375 on Build GRCh38
rs376279998 Chr.3: 47511090-47511090 on Build GRCh38
rs187474789 Chr.3: 47511106-47511106 on Build GRCh38
rs534046075 Chr.3: 47511093-47511093 on Build GRCh38
rs774818568 Chr.3: 47511156-47511156 on Build GRCh38
rs766583754 Chr.3: 47511111-47511111 on Build GRCh38
rs754992653 Chr.3: 47511127-47511127 on Build GRCh38
rs973256092 Chr.3: 47511211-47511211 on Build GRCh38
rs779396596 Chr.3: 47511153-47511153 on Build GRCh38
rs572859673 Chr.3: 47511310-47511310 on Build GRCh38
rs775994937 Chr.3: 47511200-47511200 on Build GRCh38
rs771333651 Chr.3: 47511146-47511146 on Build GRCh38
rs774539239 Chr.3: 47511241-47511241 on Build GRCh38
rs192324660 Chr.3: 47511258-47511258 on Build GRCh38
rs753248907 Chr.3: 47511277-47511277 on Build GRCh38
rs746914304 Chr.3: 47511197-47511197 on Build GRCh38

View Full Product Details