Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48449989-48450523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATRIP
Entrez Gene ID
84126
Gene Name
ATR interacting protein
Gene Aliases
-
UniGene
Hs.694840
Gene Chromosome Location
Chr.3: 48446710-48465655 on Build GRCh38
Transcripts
NM_001271022.1 NM_001271023.1 NM_032166.3 NM_130384.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4187932 c.274G>A p.D92N Chr.3: 48450063-48450063 on Build GRCh38
6097868 c.274G>T p.D92Y Chr.3: 48450063-48450063 on Build GRCh38
6608494 c.248-10delT p.? Chr.3: 48450027-48450027 on Build GRCh38
292369 c.288A>G p.K96K Chr.3: 48450077-48450077 on Build GRCh38
1045816 c.336C>T p.F112F Chr.3: 48450125-48450125 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367760362 Chr.3: 48450016-48450016 on Build GRCh38
rs760847721 Chr.3: 48450034-48450034 on Build GRCh38
rs779266318 Chr.3: 48450135-48450135 on Build GRCh38
rs578158086 Chr.3: 48450481-48450481 on Build GRCh38
rs149518027 Chr.3: 48450126-48450126 on Build GRCh38
rs142063821 Chr.3: 48450109-48450109 on Build GRCh38
rs766393286 Chr.3: 48450467-48450467 on Build GRCh38
rs370870241 Chr.3: 48450216-48450216 on Build GRCh38
rs182238067 Chr.3: 48450284-48450284 on Build GRCh38
rs11922041 Chr.3: 48450188-48450188 on Build GRCh38
rs757630635 Chr.3: 48450129-48450129 on Build GRCh38
rs893775783 Chr.3: 48450380-48450380 on Build GRCh38
rs754491690 Chr.3: 48450121-48450121 on Build GRCh38
rs998056753 Chr.3: 48450136-48450136 on Build GRCh38
rs925195192 Chr.3: 48450279-48450279 on Build GRCh38
rs764399960 Chr.3: 48450042-48450042 on Build GRCh38
rs531253407 Chr.3: 48450190-48450190 on Build GRCh38
rs761053270 Chr.3: 48450366-48450368 on Build GRCh38
rs775553560 Chr.3: 48450026-48450026 on Build GRCh38
rs1042621923 Chr.3: 48450249-48450249 on Build GRCh38
rs547592863 Chr.3: 48450289-48450289 on Build GRCh38
rs746435130 Chr.3: 48450020-48450020 on Build GRCh38
rs973593597 Chr.3: 48450229-48450229 on Build GRCh38
rs751131967 Chr.3: 48450120-48450120 on Build GRCh38
rs549456831 Chr.3: 48450217-48450217 on Build GRCh38
rs775500131 Chr.3: 48450164-48450164 on Build GRCh38
rs761592490 Chr.3: 48450264-48450265 on Build GRCh38
rs902464923 Chr.3: 48450393-48450393 on Build GRCh38
rs749995083 Chr.3: 48450360-48450361 on Build GRCh38
rs995745348 Chr.3: 48450491-48450491 on Build GRCh38
rs182766845 Chr.3: 48450019-48450019 on Build GRCh38
rs929475119 Chr.3: 48450281-48450281 on Build GRCh38
rs1042325219 Chr.3: 48450390-48450390 on Build GRCh38
rs538250510 Chr.3: 48450436-48450436 on Build GRCh38
rs761663934 Chr.3: 48450096-48450096 on Build GRCh38
rs140650148 Chr.3: 48450130-48450130 on Build GRCh38
rs763116772 Chr.3: 48450433-48450433 on Build GRCh38
rs191497815 Chr.3: 48450269-48450269 on Build GRCh38
rs747070222 Chr.3: 48450167-48450167 on Build GRCh38
rs368911609 Chr.3: 48450105-48450105 on Build GRCh38
rs962916164 Chr.3: 48450485-48450485 on Build GRCh38
rs534776780 Chr.3: 48450405-48450405 on Build GRCh38
rs374799413 Chr.3: 48450100-48450100 on Build GRCh38
rs1029935080 Chr.3: 48450470-48450470 on Build GRCh38
rs917740843 Chr.3: 48450237-48450237 on Build GRCh38
rs919515391 Chr.3: 48450245-48450245 on Build GRCh38
rs371133192 Chr.3: 48450187-48450187 on Build GRCh38
rs761952854 Chr.3: 48450193-48450193 on Build GRCh38
rs765945400 Chr.3: 48450111-48450111 on Build GRCh38
rs150341628 Chr.3: 48450125-48450125 on Build GRCh38
rs946702092 Chr.3: 48450388-48450388 on Build GRCh38
rs928050859 Chr.3: 48450330-48450330 on Build GRCh38
rs866210291 Chr.3: 48450492-48450492 on Build GRCh38
rs1016988588 Chr.3: 48450206-48450206 on Build GRCh38
rs996963848 Chr.3: 48450021-48450021 on Build GRCh38
rs1004358677 Chr.3: 48450454-48450454 on Build GRCh38
rs11925638 Chr.3: 48450162-48450162 on Build GRCh38
rs891205529 Chr.3: 48450432-48450432 on Build GRCh38
rs751297998 Chr.3: 48450178-48450182 on Build GRCh38
rs953958703 Chr.3: 48450182-48450182 on Build GRCh38
rs1030227037 Chr.3: 48450023-48450023 on Build GRCh38
rs556646454 Chr.3: 48450440-48450440 on Build GRCh38
rs567652194 Chr.3: 48450412-48450412 on Build GRCh38
rs1054439756 Chr.3: 48450358-48450358 on Build GRCh38
rs754773325 Chr.3: 48450222-48450223 on Build GRCh38
rs11918058 Chr.3: 48450351-48450351 on Build GRCh38
rs772082890 Chr.3: 48450022-48450022 on Build GRCh38
rs982723251 Chr.3: 48450299-48450299 on Build GRCh38
rs766409015 Chr.3: 48450479-48450479 on Build GRCh38
rs188036456 Chr.3: 48450341-48450341 on Build GRCh38
rs746308489 Chr.3: 48450144-48450144 on Build GRCh38
rs996924474 Chr.3: 48450399-48450399 on Build GRCh38
rs565804267 Chr.3: 48450311-48450311 on Build GRCh38
rs745473839 Chr.3: 48450081-48450081 on Build GRCh38

View Full Product Details