Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109612675-109613176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAT2
Entrez Gene ID
2780
Gene Name
G protein subunit alpha transducin 2
Gene Aliases
ACHM4, GNATC
UniGene
Hs.36973
Gene Chromosome Location
Chr.1: 109603091-109619733 on Build GRCh38
Transcripts
NM_005272.3 XM_011541264.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1208400 c.63G>T p.K21N Chr.1: 109612808-109612808 on Build GRCh38
893549 c.74A>T p.E25V Chr.1: 109612797-109612797 on Build GRCh38
4020394 c.47C>A p.S16Y Chr.1: 109612824-109612824 on Build GRCh38
893548 c.109C>A p.L37I Chr.1: 109612762-109612762 on Build GRCh38
4020393 c.75G>T p.E25D Chr.1: 109612796-109612796 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs116819755 Chr.1: 109612999-109612999 on Build GRCh38
rs772922679 Chr.1: 109612819-109612819 on Build GRCh38
rs946827795 Chr.1: 109613046-109613046 on Build GRCh38
rs1008134895 Chr.1: 109613014-109613014 on Build GRCh38
rs776479555 Chr.1: 109612833-109612833 on Build GRCh38
rs1027082056 Chr.1: 109613082-109613082 on Build GRCh38
rs917910292 Chr.1: 109612715-109612715 on Build GRCh38
rs780494235 Chr.1: 109612742-109612742 on Build GRCh38
rs770886199 Chr.1: 109612823-109612823 on Build GRCh38
rs758154861 Chr.1: 109612720-109612720 on Build GRCh38
rs765367881 Chr.1: 109612866-109612866 on Build GRCh38
rs963851109 Chr.1: 109613030-109613030 on Build GRCh38
rs755513575 Chr.1: 109612891-109612891 on Build GRCh38
rs199503029 Chr.1: 109612857-109612857 on Build GRCh38
rs762637247 Chr.1: 109612867-109612867 on Build GRCh38
rs759055694 Chr.1: 109612840-109612840 on Build GRCh38
rs577203330 Chr.1: 109612700-109612700 on Build GRCh38
rs372507125 Chr.1: 109612856-109612856 on Build GRCh38
rs754140721 Chr.1: 109612714-109612714 on Build GRCh38
rs748227612 Chr.1: 109612911-109612911 on Build GRCh38
rs191124987 Chr.1: 109613084-109613084 on Build GRCh38
rs1018115608 Chr.1: 109613015-109613015 on Build GRCh38
rs748317500 Chr.1: 109612794-109612794 on Build GRCh38
rs1025434520 Chr.1: 109613122-109613122 on Build GRCh38
rs951430058 Chr.1: 109612695-109612695 on Build GRCh38
rs772291136 Chr.1: 109612809-109612809 on Build GRCh38
rs2304355 Chr.1: 109612902-109612902 on Build GRCh38
rs180838035 Chr.1: 109612934-109612934 on Build GRCh38
rs561211882 Chr.1: 109612744-109612747 on Build GRCh38
rs879119030 Chr.1: 109613107-109613107 on Build GRCh38
rs982867768 Chr.1: 109613086-109613086 on Build GRCh38
rs750766880 Chr.1: 109612739-109612739 on Build GRCh38
rs752278435 Chr.1: 109612863-109612863 on Build GRCh38
rs763845288 Chr.1: 109612868-109612868 on Build GRCh38
rs113070910 Chr.1: 109612737-109612737 on Build GRCh38
rs547633476 Chr.1: 109612973-109612973 on Build GRCh38
rs914349631 Chr.1: 109613032-109613032 on Build GRCh38
rs1054229968 Chr.1: 109612909-109612909 on Build GRCh38
rs779079945 Chr.1: 109612760-109612760 on Build GRCh38
rs756496666 Chr.1: 109612878-109612878 on Build GRCh38
rs542546686 Chr.1: 109612701-109612702 on Build GRCh38
rs373698555 Chr.1: 109612913-109612913 on Build GRCh38
rs886045029 Chr.1: 109612958-109612958 on Build GRCh38
rs535445250 Chr.1: 109613110-109613110 on Build GRCh38
rs749377354 Chr.1: 109612753-109612753 on Build GRCh38
rs1045338055 Chr.1: 109612842-109612842 on Build GRCh38
rs750796967 Chr.1: 109612877-109612877 on Build GRCh38
rs979958186 Chr.1: 109613048-109613048 on Build GRCh38
rs34046712 Chr.1: 109612922-109612923 on Build GRCh38
rs564936072 Chr.1: 109612907-109612908 on Build GRCh38
rs756579867 Chr.1: 109612741-109612741 on Build GRCh38
rs186255282 Chr.1: 109613009-109613009 on Build GRCh38
rs1040644318 Chr.1: 109612871-109612871 on Build GRCh38
rs951574359 Chr.1: 109613085-109613085 on Build GRCh38
rs532113128 Chr.1: 109613001-109613001 on Build GRCh38
rs746796374 Chr.1: 109612914-109612914 on Build GRCh38
rs946765939 Chr.1: 109612757-109612757 on Build GRCh38
rs780590839 Chr.1: 109612885-109612885 on Build GRCh38
rs562449373 Chr.1: 109612886-109612886 on Build GRCh38
rs1043733773 Chr.1: 109612789-109612789 on Build GRCh38
rs527385567 Chr.1: 109612943-109612943 on Build GRCh38
rs114457486 Chr.1: 109612696-109612696 on Build GRCh38
rs995274892 Chr.1: 109613052-109613052 on Build GRCh38
rs760450896 Chr.1: 109612820-109612820 on Build GRCh38
rs546168176 Chr.1: 109612733-109612733 on Build GRCh38

View Full Product Details