Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 120167302-120167800 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR156
Entrez Gene ID
165829
Gene Name
G protein-coupled receptor 156
Gene Aliases
GABABL, PGR28
UniGene
Hs.333358
Gene Chromosome Location
Chr.3: 120165481-120285122 on Build GRCh38
Transcripts
NM_001168271.1 NM_153002.2 XM_005247165.2 XM_011512484.2 XM_011512487.1 XM_011512488.2 XM_017005795.1 XM_017005796.1 XM_017005797.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6480222 c.2157C>G p.D719E Chr.3: 120167320-120167320 on Build GRCh38
5863283 c.1787C>T p.P596L Chr.3: 120167690-120167690 on Build GRCh38
3845937 c.1896C>T p.H632H Chr.3: 120167581-120167581 on Build GRCh38
1418075 c.1820G>A p.R607Q Chr.3: 120167657-120167657 on Build GRCh38
6439519 c.1762C>T p.L588F Chr.3: 120167715-120167715 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375975333 Chr.3: 120167681-120167681 on Build GRCh38
rs867700471 Chr.3: 120167415-120167415 on Build GRCh38
rs886482415 Chr.3: 120167617-120167617 on Build GRCh38
rs1006667011 Chr.3: 120167575-120167575 on Build GRCh38
rs757923817 Chr.3: 120167611-120167611 on Build GRCh38
rs112549051 Chr.3: 120167475-120167475 on Build GRCh38
rs200929093 Chr.3: 120167387-120167387 on Build GRCh38
rs769422019 Chr.3: 120167743-120167743 on Build GRCh38
rs778495665 Chr.3: 120167609-120167609 on Build GRCh38
rs202063599 Chr.3: 120167760-120167760 on Build GRCh38
rs760809125 Chr.3: 120167761-120167761 on Build GRCh38
rs114542754 Chr.3: 120167418-120167418 on Build GRCh38
rs756049689 Chr.3: 120167723-120167723 on Build GRCh38
rs553212940 Chr.3: 120167514-120167514 on Build GRCh38
rs372902834 Chr.3: 120167657-120167657 on Build GRCh38
rs750786372 Chr.3: 120167422-120167422 on Build GRCh38
rs550290864 Chr.3: 120167748-120167748 on Build GRCh38
rs769408777 Chr.3: 120167331-120167331 on Build GRCh38
rs768922573 Chr.3: 120167647-120167647 on Build GRCh38
rs760092352 Chr.3: 120167773-120167773 on Build GRCh38
rs140788830 Chr.3: 120167779-120167779 on Build GRCh38
rs867162378 Chr.3: 120167342-120167342 on Build GRCh38
rs768056878 Chr.3: 120167421-120167421 on Build GRCh38
rs560588984 Chr.3: 120167652-120167652 on Build GRCh38
rs776945664 Chr.3: 120167768-120167768 on Build GRCh38
rs773109242 Chr.3: 120167367-120167367 on Build GRCh38
rs766727029 Chr.3: 120167595-120167595 on Build GRCh38
rs947753177 Chr.3: 120167554-120167554 on Build GRCh38
rs150922548 Chr.3: 120167765-120167765 on Build GRCh38
rs756619608 Chr.3: 120167588-120167588 on Build GRCh38
rs923796467 Chr.3: 120167676-120167676 on Build GRCh38
rs748905811 Chr.3: 120167441-120167441 on Build GRCh38
rs376129523 Chr.3: 120167651-120167651 on Build GRCh38
rs9858566 Chr.3: 120167701-120167701 on Build GRCh38
rs143524747 Chr.3: 120167670-120167670 on Build GRCh38
rs751162137 Chr.3: 120167711-120167711 on Build GRCh38
rs536737767 Chr.3: 120167730-120167730 on Build GRCh38
rs761123808 Chr.3: 120167669-120167669 on Build GRCh38
rs754620284 Chr.3: 120167455-120167455 on Build GRCh38
rs148653681 Chr.3: 120167610-120167610 on Build GRCh38
rs146755184 Chr.3: 120167677-120167677 on Build GRCh38
rs761951530 Chr.3: 120167569-120167569 on Build GRCh38
rs758950332 Chr.3: 120167632-120167632 on Build GRCh38
rs773739887 Chr.3: 120167361-120167361 on Build GRCh38
rs779752844 Chr.3: 120167346-120167346 on Build GRCh38
rs765648012 Chr.3: 120167685-120167685 on Build GRCh38
rs939480246 Chr.3: 120167407-120167407 on Build GRCh38
rs779904863 Chr.3: 120167746-120167746 on Build GRCh38
rs188225112 Chr.3: 120167774-120167774 on Build GRCh38
rs768002076 Chr.3: 120167570-120167570 on Build GRCh38
rs911897667 Chr.3: 120167379-120167379 on Build GRCh38
rs748735036 Chr.3: 120167350-120167350 on Build GRCh38
rs200587090 Chr.3: 120167427-120167427 on Build GRCh38
rs772131075 Chr.3: 120167499-120167499 on Build GRCh38
rs560879228 Chr.3: 120167776-120167778 on Build GRCh38
rs781060681 Chr.3: 120167733-120167733 on Build GRCh38
rs746900032 Chr.3: 120167627-120167627 on Build GRCh38
rs763184111 Chr.3: 120167527-120167527 on Build GRCh38
rs752144877 Chr.3: 120167501-120167503 on Build GRCh38
rs115973935 Chr.3: 120167419-120167419 on Build GRCh38
rs906074777 Chr.3: 120167780-120167780 on Build GRCh38
rs754544859 Chr.3: 120167396-120167398 on Build GRCh38
rs763087402 Chr.3: 120167405-120167405 on Build GRCh38
rs761772174 Chr.3: 120167366-120167366 on Build GRCh38
rs974761592 Chr.3: 120167654-120167654 on Build GRCh38
rs745666133 Chr.3: 120167736-120167736 on Build GRCh38
rs565334408 Chr.3: 120167560-120167560 on Build GRCh38
rs759874526 Chr.3: 120167682-120167682 on Build GRCh38
rs772305456 Chr.3: 120167751-120167751 on Build GRCh38
rs771433011 Chr.3: 120167658-120167658 on Build GRCh38
rs764273875 Chr.3: 120167538-120167538 on Build GRCh38
rs776319315 Chr.3: 120167520-120167520 on Build GRCh38
rs763876310 Chr.3: 120167778-120167778 on Build GRCh38
rs367758171 Chr.3: 120167357-120167357 on Build GRCh38
rs757734122 Chr.3: 120167325-120167325 on Build GRCh38
rs762670033 Chr.3: 120167775-120167775 on Build GRCh38
rs771011344 Chr.3: 120167631-120167631 on Build GRCh38
rs201531419 Chr.3: 120167655-120167655 on Build GRCh38
rs751263400 Chr.3: 120167324-120167324 on Build GRCh38
rs752327252 Chr.3: 120167691-120167691 on Build GRCh38
rs778461783 Chr.3: 120167459-120167459 on Build GRCh38
rs200604393 Chr.3: 120167469-120167469 on Build GRCh38
rs145001260 Chr.3: 120167596-120167596 on Build GRCh38
rs150001619 Chr.3: 120167513-120167513 on Build GRCh38
rs139313892 Chr.3: 120167511-120167511 on Build GRCh38
rs760612038 Chr.3: 120167386-120167386 on Build GRCh38
rs386665142 Chr.3: 120167418-120167419 on Build GRCh38
rs777746158 Chr.3: 120167765-120167765 on Build GRCh38
rs780411492 Chr.3: 120167431-120167431 on Build GRCh38
rs923653303 Chr.3: 120167449-120167449 on Build GRCh38
rs754625557 Chr.3: 120167601-120167601 on Build GRCh38
rs939513149 Chr.3: 120167633-120167633 on Build GRCh38
rs777422969 Chr.3: 120167624-120167624 on Build GRCh38
rs962580007 Chr.3: 120167599-120167599 on Build GRCh38
rs767034751 Chr.3: 120167460-120167464 on Build GRCh38
rs140248719 Chr.3: 120167694-120167694 on Build GRCh38
rs973736400 Chr.3: 120167360-120167360 on Build GRCh38
rs775719509 Chr.3: 120167388-120167388 on Build GRCh38

View Full Product Details