Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 161103285-161103693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GALNT1
Entrez Gene ID
8706
Gene Name
beta-1,3-N-acetylgalactosaminyltransferase 1 (globoside blood group)
Gene Aliases
B3GALT3, GLCT3, GLOB, Gb4Cer, P, P1, beta3Gal-T3, galT3
UniGene
Hs.418062
Gene Chromosome Location
Chr.3: 161083883-161105528 on Build GRCh38
Transcripts
NM_001038628.1 NM_003781.3 NM_033167.2 NM_033168.2 NM_033169.2 XM_005247859.4 XM_005247861.3 XM_005247862.3 XM_005247864.3 XM_006713803.3 XM_006713804.3 XM_006713805.3 XM_017007402.1 XM_017007403.1 XM_017007404.1 XM_017007405.1 XM_017007406.1 XM_017007407.1 XM_017007408.1 XM_017007409.1 XM_017007410.1 XM_017007411.1 XM_017007412.1 XM_017007413.1

SNP Detail

SNP ID Chromosome Location
rs531532021 Chr.3: 161103467-161103467 on Build GRCh38
rs974139396 Chr.3: 161103347-161103347 on Build GRCh38
rs996550357 Chr.3: 161103532-161103532 on Build GRCh38
rs767362312 Chr.3: 161103630-161103630 on Build GRCh38
rs185667635 Chr.3: 161103423-161103423 on Build GRCh38
rs762979783 Chr.3: 161103376-161103376 on Build GRCh38
rs548787698 Chr.3: 161103390-161103390 on Build GRCh38
rs571526794 Chr.3: 161103534-161103534 on Build GRCh38
rs1032294674 Chr.3: 161103450-161103450 on Build GRCh38
rs987138908 Chr.3: 161103588-161103588 on Build GRCh38
rs528946682 Chr.3: 161103371-161103371 on Build GRCh38
rs1022058691 Chr.3: 161103508-161103508 on Build GRCh38
rs868692857 Chr.3: 161103342-161103342 on Build GRCh38
rs907484855 Chr.3: 161103402-161103402 on Build GRCh38
rs1021215957 Chr.3: 161103540-161103543 on Build GRCh38
rs997685109 Chr.3: 161103414-161103414 on Build GRCh38
rs148395014 Chr.3: 161103483-161103483 on Build GRCh38
rs188930363 Chr.3: 161103550-161103550 on Build GRCh38
rs764168541 Chr.3: 161103325-161103325 on Build GRCh38
rs761722441 Chr.3: 161103523-161103523 on Build GRCh38
rs1053352011 Chr.3: 161103447-161103447 on Build GRCh38
rs938914533 Chr.3: 161103422-161103422 on Build GRCh38
rs986678864 Chr.3: 161103328-161103328 on Build GRCh38
rs1007621848 Chr.3: 161103374-161103374 on Build GRCh38
rs369335123 Chr.3: 161103403-161103407 on Build GRCh38
rs373892479 Chr.3: 161103655-161103655 on Build GRCh38
rs36051722 Chr.3: 161103327-161103327 on Build GRCh38
rs919976854 Chr.3: 161103364-161103364 on Build GRCh38
rs1044371598 Chr.3: 161103385-161103385 on Build GRCh38
rs908352193 Chr.3: 161103329-161103329 on Build GRCh38
rs753387954 Chr.3: 161103495-161103496 on Build GRCh38
rs961509999 Chr.3: 161103608-161103608 on Build GRCh38
rs1015480728 Chr.3: 161103594-161103594 on Build GRCh38
rs948725468 Chr.3: 161103373-161103373 on Build GRCh38
rs34791869 Chr.3: 161103648-161103648 on Build GRCh38
rs939844343 Chr.3: 161103341-161103341 on Build GRCh38
rs774378292 Chr.3: 161103503-161103503 on Build GRCh38
rs979761340 Chr.3: 161103574-161103574 on Build GRCh38
rs869154804 Chr.3: 161103402-161103406 on Build GRCh38
rs540176884 Chr.3: 161103315-161103315 on Build GRCh38

View Full Product Details