Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 172040450-172040953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNDC3B
Entrez Gene ID
64778
Gene Name
fibronectin type III domain containing 3B
Gene Aliases
FAD104, PRO4979, YVTM2421
UniGene
Hs.744888
Gene Chromosome Location
Chr.3: 172039628-172400703 on Build GRCh38
Transcripts
NM_001135095.1 NM_022763.3 XM_017007062.1 XM_017007064.1

SNP Detail

SNP ID Chromosome Location
rs899192542 Chr.3: 172040471-172040471 on Build GRCh38
rs1006789481 Chr.3: 172040500-172040500 on Build GRCh38
rs978481409 Chr.3: 172040686-172040686 on Build GRCh38
rs886746142 Chr.3: 172040531-172040551 on Build GRCh38
rs1010687237 Chr.3: 172040796-172040796 on Build GRCh38
rs929623037 Chr.3: 172040483-172040483 on Build GRCh38
rs58908502 Chr.3: 172040899-172040899 on Build GRCh38
rs974302172 Chr.3: 172040817-172040817 on Build GRCh38
rs912240302 Chr.3: 172040864-172040864 on Build GRCh38
rs960919877 Chr.3: 172040748-172040748 on Build GRCh38
rs933537499 Chr.3: 172040588-172040588 on Build GRCh38
rs960699707 Chr.3: 172040930-172040930 on Build GRCh38
rs891022820 Chr.3: 172040494-172040494 on Build GRCh38
rs575780247 Chr.3: 172040893-172040893 on Build GRCh38
rs978159143 Chr.3: 172040543-172040543 on Build GRCh38
rs1048088801 Chr.3: 172040483-172040493 on Build GRCh38
rs999630288 Chr.3: 172040925-172040925 on Build GRCh38
rs993488009 Chr.3: 172040476-172040476 on Build GRCh38
rs910701735 Chr.3: 172040632-172040632 on Build GRCh38
rs966394265 Chr.3: 172040808-172040808 on Build GRCh38
rs183288040 Chr.3: 172040467-172040467 on Build GRCh38
rs918328519 Chr.3: 172040830-172040830 on Build GRCh38
rs1048157519 Chr.3: 172040707-172040707 on Build GRCh38
rs982532203 Chr.3: 172040564-172040564 on Build GRCh38
rs559633164 Chr.3: 172040724-172040724 on Build GRCh38
rs906461990 Chr.3: 172040740-172040740 on Build GRCh38
rs959132526 Chr.3: 172040498-172040498 on Build GRCh38
rs993513758 Chr.3: 172040689-172040689 on Build GRCh38
rs1039597306 Chr.3: 172040871-172040871 on Build GRCh38
rs1018032142 Chr.3: 172040544-172040544 on Build GRCh38
rs916711065 Chr.3: 172040804-172040804 on Build GRCh38
rs1026268025 Chr.3: 172040483-172040483 on Build GRCh38
rs564694405 Chr.3: 172040892-172040892 on Build GRCh38
rs966860854 Chr.3: 172040677-172040677 on Build GRCh38
rs902583841 Chr.3: 172040908-172040908 on Build GRCh38
rs899244463 Chr.3: 172040685-172040685 on Build GRCh38
rs1051188368 Chr.3: 172040860-172040860 on Build GRCh38
rs977795447 Chr.3: 172040811-172040811 on Build GRCh38
rs1035049280 Chr.3: 172040717-172040717 on Build GRCh38
rs943502777 Chr.3: 172040648-172040648 on Build GRCh38
rs966382863 Chr.3: 172040534-172040534 on Build GRCh38
rs111903995 Chr.3: 172040518-172040518 on Build GRCh38
rs892988263 Chr.3: 172040563-172040563 on Build GRCh38
rs942595703 Chr.3: 172040868-172040868 on Build GRCh38
rs1022796807 Chr.3: 172040599-172040599 on Build GRCh38
rs1050821132 Chr.3: 172040491-172040491 on Build GRCh38
rs929764851 Chr.3: 172040849-172040849 on Build GRCh38
rs1033538823 Chr.3: 172040756-172040756 on Build GRCh38
rs941532900 Chr.3: 172040806-172040806 on Build GRCh38
rs543069100 Chr.3: 172040694-172040694 on Build GRCh38
rs116145421 Chr.3: 172040882-172040882 on Build GRCh38
rs991089215 Chr.3: 172040774-172040774 on Build GRCh38
rs1033900186 Chr.3: 172040493-172040494 on Build GRCh38
rs1034996067 Chr.3: 172040927-172040927 on Build GRCh38

View Full Product Details