Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 180983906-180984447 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC19
Entrez Gene ID
131118
Gene Name
DnaJ heat shock protein family (Hsp40) member C19
Gene Aliases
PAM18, TIM14, TIMM14
UniGene
Hs.230601
Gene Chromosome Location
Chr.3: 180983709-180989774 on Build GRCh38
Transcripts
NM_001190233.1 NM_145261.3 NR_033721.1 NR_033722.1 NR_033723.1 NR_046073.1

SNP Detail

SNP ID Chromosome Location
rs747455339 Chr.3: 180984032-180984032 on Build GRCh38
rs947602721 Chr.3: 180984027-180984027 on Build GRCh38
rs186247662 Chr.3: 180984150-180984150 on Build GRCh38
rs1041026360 Chr.3: 180984054-180984054 on Build GRCh38
rs969387870 Chr.3: 180984294-180984294 on Build GRCh38
rs774343049 Chr.3: 180984087-180984087 on Build GRCh38
rs886058202 Chr.3: 180984315-180984315 on Build GRCh38
rs763157750 Chr.3: 180984225-180984225 on Build GRCh38
rs899940959 Chr.3: 180983969-180983969 on Build GRCh38
rs576015416 Chr.3: 180983953-180983953 on Build GRCh38
rs143458718 Chr.3: 180984275-180984275 on Build GRCh38
rs922592862 Chr.3: 180984419-180984419 on Build GRCh38
rs779379432 Chr.3: 180984405-180984405 on Build GRCh38
rs746098932 Chr.3: 180984350-180984350 on Build GRCh38
rs564709350 Chr.3: 180984111-180984111 on Build GRCh38
rs780680646 Chr.3: 180984010-180984010 on Build GRCh38
rs745435747 Chr.3: 180983968-180983968 on Build GRCh38
rs903237371 Chr.3: 180984049-180984049 on Build GRCh38
rs997423816 Chr.3: 180984104-180984104 on Build GRCh38
rs776304531 Chr.3: 180983942-180983942 on Build GRCh38
rs548246397 Chr.3: 180984403-180984403 on Build GRCh38
rs576646391 Chr.3: 180984152-180984152 on Build GRCh38
rs555888288 Chr.3: 180983948-180983948 on Build GRCh38
rs764012878 Chr.3: 180984283-180984283 on Build GRCh38
rs762130203 Chr.3: 180984124-180984124 on Build GRCh38
rs769028541 Chr.3: 180984091-180984091 on Build GRCh38
rs1047323892 Chr.3: 180984156-180984156 on Build GRCh38
rs11547440 Chr.3: 180984143-180984143 on Build GRCh38
rs778688606 Chr.3: 180984327-180984327 on Build GRCh38
rs768968975 Chr.3: 180983934-180983934 on Build GRCh38
rs912833362 Chr.3: 180984176-180984177 on Build GRCh38
rs888930915 Chr.3: 180984113-180984113 on Build GRCh38
rs991682423 Chr.3: 180984251-180984251 on Build GRCh38
rs901181077 Chr.3: 180984090-180984090 on Build GRCh38
rs750041383 Chr.3: 180984369-180984369 on Build GRCh38
rs760642586 Chr.3: 180984281-180984281 on Build GRCh38
rs114637069 Chr.3: 180984261-180984261 on Build GRCh38
rs753726273 Chr.3: 180984316-180984316 on Build GRCh38
rs961799533 Chr.3: 180984176-180984176 on Build GRCh38
rs148074891 Chr.3: 180984097-180984098 on Build GRCh38
rs1018290358 Chr.3: 180983956-180983956 on Build GRCh38
rs34661228 Chr.3: 180984066-180984067 on Build GRCh38
rs920139228 Chr.3: 180984115-180984115 on Build GRCh38
rs1011696769 Chr.3: 180984243-180984243 on Build GRCh38
rs934758233 Chr.3: 180984053-180984053 on Build GRCh38
rs36016478 Chr.3: 180983966-180983966 on Build GRCh38
rs749423439 Chr.3: 180984173-180984173 on Build GRCh38
rs538879345 Chr.3: 180984096-180984097 on Build GRCh38
rs757933163 Chr.3: 180984400-180984400 on Build GRCh38
rs773292491 Chr.3: 180984212-180984212 on Build GRCh38
rs1039686087 Chr.3: 180984209-180984209 on Build GRCh38
rs146964003 Chr.3: 180983995-180983995 on Build GRCh38
rs899874966 Chr.3: 180984233-180984233 on Build GRCh38
rs1021498576 Chr.3: 180984013-180984013 on Build GRCh38
rs757100034 Chr.3: 180984320-180984320 on Build GRCh38
rs1805635 Chr.3: 180984199-180984199 on Build GRCh38
rs934697111 Chr.3: 180984401-180984401 on Build GRCh38
rs561308095 Chr.3: 180984208-180984208 on Build GRCh38
rs375439278 Chr.3: 180984392-180984392 on Build GRCh38
rs1016525460 Chr.3: 180984164-180984164 on Build GRCh38
rs938898464 Chr.3: 180983938-180983938 on Build GRCh38
rs752694507 Chr.3: 180984267-180984267 on Build GRCh38
rs761431774 Chr.3: 180983952-180983952 on Build GRCh38
rs978928094 Chr.3: 180984380-180984380 on Build GRCh38
rs993211828 Chr.3: 180984198-180984198 on Build GRCh38
rs777982585 Chr.3: 180984116-180984116 on Build GRCh38
rs1043521824 Chr.3: 180984041-180984041 on Build GRCh38
rs1035336239 Chr.3: 180984057-180984057 on Build GRCh38
rs372954525 Chr.3: 180984105-180984105 on Build GRCh38

View Full Product Details