Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 186581878-186582422 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB11
Entrez Gene ID
51726
Gene Name
DnaJ heat shock protein family (Hsp40) member B11
Gene Aliases
ABBP-2, ABBP2, DJ9, Dj-9, EDJ, ERdj3, ERj3, ERj3p, PRO1080, UNQ537
UniGene
Hs.317192
Gene Chromosome Location
Chr.3: 186570676-186585800 on Build GRCh38
Transcripts
NM_016306.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1204173 c.616C>T p.R206* Chr.3: 186582011-186582011 on Build GRCh38
5891344 c.667C>T p.P223S Chr.3: 186582062-186582062 on Build GRCh38
6266276 c.643G>T p.G215W Chr.3: 186582038-186582038 on Build GRCh38
23322 c.613G>T p.E205* Chr.3: 186582008-186582008 on Build GRCh38
5459043 c.620C>T p.T207M Chr.3: 186582015-186582015 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs6784026 Chr.3: 186582116-186582116 on Build GRCh38
rs754070952 Chr.3: 186582045-186582045 on Build GRCh38
rs374509746 Chr.3: 186582012-186582012 on Build GRCh38
rs1018326576 Chr.3: 186582187-186582187 on Build GRCh38
rs937248387 Chr.3: 186582386-186582386 on Build GRCh38
rs941660944 Chr.3: 186582325-186582325 on Build GRCh38
rs200565661 Chr.3: 186582069-186582069 on Build GRCh38
rs762271812 Chr.3: 186582048-186582048 on Build GRCh38
rs201578569 Chr.3: 186582126-186582126 on Build GRCh38
rs1020379385 Chr.3: 186582167-186582167 on Build GRCh38
rs907625402 Chr.3: 186582319-186582319 on Build GRCh38
rs768333865 Chr.3: 186581970-186581970 on Build GRCh38
rs969553860 Chr.3: 186582145-186582145 on Build GRCh38
rs941713150 Chr.3: 186582011-186582011 on Build GRCh38
rs773149769 Chr.3: 186581996-186581996 on Build GRCh38
rs1030107179 Chr.3: 186581911-186581912 on Build GRCh38
rs372977627 Chr.3: 186582186-186582186 on Build GRCh38
rs201458470 Chr.3: 186581981-186581981 on Build GRCh38
rs1030225159 Chr.3: 186582265-186582265 on Build GRCh38
rs767082355 Chr.3: 186582271-186582272 on Build GRCh38
rs969087434 Chr.3: 186582180-186582180 on Build GRCh38
rs760852405 Chr.3: 186582024-186582024 on Build GRCh38
rs149200786 Chr.3: 186582193-186582193 on Build GRCh38
rs773710743 Chr.3: 186581982-186581989 on Build GRCh38
rs755739463 Chr.3: 186581905-186581905 on Build GRCh38
rs1057074131 Chr.3: 186582397-186582397 on Build GRCh38
rs769638452 Chr.3: 186582110-186582110 on Build GRCh38
rs56084565 Chr.3: 186582217-186582217 on Build GRCh38
rs1001448127 Chr.3: 186582093-186582093 on Build GRCh38
rs764395900 Chr.3: 186582040-186582040 on Build GRCh38
rs780775457 Chr.3: 186581979-186581979 on Build GRCh38
rs41268611 Chr.3: 186582003-186582003 on Build GRCh38
rs751024041 Chr.3: 186582050-186582050 on Build GRCh38
rs756926511 Chr.3: 186582212-186582212 on Build GRCh38
rs368496191 Chr.3: 186582016-186582016 on Build GRCh38
rs140614246 Chr.3: 186582341-186582341 on Build GRCh38
rs762867519 Chr.3: 186582009-186582009 on Build GRCh38
rs747742959 Chr.3: 186582103-186582103 on Build GRCh38
rs926998247 Chr.3: 186582385-186582385 on Build GRCh38
rs375052103 Chr.3: 186582095-186582095 on Build GRCh38
rs371013780 Chr.3: 186582211-186582211 on Build GRCh38
rs951597443 Chr.3: 186582272-186582272 on Build GRCh38
rs770875769 Chr.3: 186582129-186582129 on Build GRCh38
rs777543921 Chr.3: 186582114-186582114 on Build GRCh38
rs779479780 Chr.3: 186582061-186582061 on Build GRCh38
rs772113983 Chr.3: 186582015-186582015 on Build GRCh38
rs114859750 Chr.3: 186582305-186582305 on Build GRCh38
rs533556965 Chr.3: 186582288-186582288 on Build GRCh38
rs34707834 Chr.3: 186582285-186582285 on Build GRCh38
rs370808915 Chr.3: 186582051-186582051 on Build GRCh38
rs982242780 Chr.3: 186582149-186582149 on Build GRCh38
rs941341786 Chr.3: 186582256-186582256 on Build GRCh38
rs151292368 Chr.3: 186582280-186582280 on Build GRCh38
rs371152451 Chr.3: 186581988-186581988 on Build GRCh38
rs199675514 Chr.3: 186582092-186582092 on Build GRCh38
rs764238700 Chr.3: 186582049-186582049 on Build GRCh38

View Full Product Details