Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 68191176-68191701 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FTLP10
Entrez Gene ID
100130017
Gene Name
ferritin light chain pseudogene 10
Gene Aliases
-
UniGene
Hs.582037
Gene Chromosome Location
Chr.4: 68182292-68212470 on Build GRCh38
Transcripts
NR_015446.1 NR_104048.1

Gene Symbol
TMPRSS11BNL
Entrez Gene ID
401136
Gene Name
TMPRSS11B N-terminal like, pseudogene
Gene Aliases
-
UniGene
Hs.652046
Gene Chromosome Location
Chr.4: 68184129-68218080 on Build GRCh38
Transcripts
NR_015446.1 NR_104048.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5540563 c.131G>A p.R44K Chr.4: 68191518-68191518 on Build GRCh38
5463362 c.242G>T p.R81I Chr.4: 68191407-68191407 on Build GRCh38
5401430 c.302C>T p.S101L Chr.4: 68191257-68191257 on Build GRCh38
4937732 c.222T>C p.L74L Chr.4: 68191427-68191427 on Build GRCh38
4797167 c.275C>T p.T92I Chr.4: 68191284-68191284 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1020994092 Chr.4: 68191401-68191401 on Build GRCh38
rs567807204 Chr.4: 68191362-68191362 on Build GRCh38
rs921993111 Chr.4: 68191516-68191516 on Build GRCh38
rs1013878093 Chr.4: 68191268-68191268 on Build GRCh38
rs144301451 Chr.4: 68191273-68191273 on Build GRCh38
rs751965735 Chr.4: 68191325-68191325 on Build GRCh38
rs998115304 Chr.4: 68191497-68191497 on Build GRCh38
rs907986870 Chr.4: 68191549-68191549 on Build GRCh38
rs754587424 Chr.4: 68191407-68191407 on Build GRCh38
rs976686258 Chr.4: 68191473-68191473 on Build GRCh38
rs1029550113 Chr.4: 68191499-68191499 on Build GRCh38
rs951164049 Chr.4: 68191225-68191225 on Build GRCh38
rs763300637 Chr.4: 68191562-68191562 on Build GRCh38
rs536695949 Chr.4: 68191386-68191386 on Build GRCh38
rs762466312 Chr.4: 68191346-68191346 on Build GRCh38
rs572958728 Chr.4: 68191644-68191644 on Build GRCh38
rs183591149 Chr.4: 68191253-68191253 on Build GRCh38
rs777649985 Chr.4: 68191505-68191505 on Build GRCh38
rs535191519 Chr.4: 68191318-68191318 on Build GRCh38
rs1046610918 Chr.4: 68191274-68191274 on Build GRCh38
rs771084152 Chr.4: 68191355-68191356 on Build GRCh38
rs1037700296 Chr.4: 68191260-68191260 on Build GRCh38
rs530124479 Chr.4: 68191208-68191208 on Build GRCh38
rs75202564 Chr.4: 68191617-68191617 on Build GRCh38
rs749602481 Chr.4: 68191388-68191388 on Build GRCh38
rs745970514 Chr.4: 68191528-68191528 on Build GRCh38
rs770508560 Chr.4: 68191470-68191470 on Build GRCh38
rs114552880 Chr.4: 68191256-68191256 on Build GRCh38
rs1046641092 Chr.4: 68191623-68191623 on Build GRCh38
rs569664095 Chr.4: 68191543-68191543 on Build GRCh38
rs556518039 Chr.4: 68191500-68191500 on Build GRCh38
rs775045343 Chr.4: 68191545-68191545 on Build GRCh38
rs778829294 Chr.4: 68191442-68191442 on Build GRCh38
rs747918720 Chr.4: 68191467-68191467 on Build GRCh38
rs764452807 Chr.4: 68191315-68191315 on Build GRCh38
rs753467165 Chr.4: 68191402-68191402 on Build GRCh38
rs576763384 Chr.4: 68191525-68191525 on Build GRCh38
rs906724979 Chr.4: 68191280-68191280 on Build GRCh38
rs917853792 Chr.4: 68191664-68191664 on Build GRCh38
rs1000507742 Chr.4: 68191308-68191308 on Build GRCh38
rs533027711 Chr.4: 68191283-68191283 on Build GRCh38
rs775579055 Chr.4: 68191548-68191548 on Build GRCh38
rs4860284 Chr.4: 68191355-68191355 on Build GRCh38
rs370555471 Chr.4: 68191267-68191267 on Build GRCh38
rs539366456 Chr.4: 68191583-68191583 on Build GRCh38
rs963482470 Chr.4: 68191563-68191563 on Build GRCh38
rs1012416163 Chr.4: 68191641-68191641 on Build GRCh38
rs1052173455 Chr.4: 68191337-68191337 on Build GRCh38
rs749990772 Chr.4: 68191358-68191358 on Build GRCh38
rs551554040 Chr.4: 68191230-68191230 on Build GRCh38
rs964026801 Chr.4: 68191487-68191487 on Build GRCh38
rs1017918547 Chr.4: 68191449-68191449 on Build GRCh38
rs935533316 Chr.4: 68191672-68191672 on Build GRCh38
rs748631191 Chr.4: 68191582-68191582 on Build GRCh38
rs922447869 Chr.4: 68191201-68191201 on Build GRCh38
rs916606194 Chr.4: 68191615-68191615 on Build GRCh38
rs1044822151 Chr.4: 68191272-68191272 on Build GRCh38

View Full Product Details