Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70634219-70634724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENAM
Entrez Gene ID
10117
Gene Name
enamelin
Gene Aliases
ADAI, AI1C, AIH2
UniGene
Hs.667018
Gene Chromosome Location
Chr.4: 70627471-70646819 on Build GRCh38
Transcripts
NM_031889.2 XM_006714056.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6653971 c.313C>T p.R105W Chr.4: 70634410-70634410 on Build GRCh38
4135870 c.446C>A p.P149H Chr.4: 70634543-70634543 on Build GRCh38
3775956 c.381G>A p.Q127Q Chr.4: 70634478-70634478 on Build GRCh38
3826139 c.399A>C p.P133P Chr.4: 70634496-70634496 on Build GRCh38
1057097 c.363A>G p.E121E Chr.4: 70634460-70634460 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370035808 Chr.4: 70634260-70634260 on Build GRCh38
rs959670000 Chr.4: 70634388-70634388 on Build GRCh38
rs917934946 Chr.4: 70634659-70634659 on Build GRCh38
rs766042938 Chr.4: 70634522-70634522 on Build GRCh38
rs1007205246 Chr.4: 70634493-70634493 on Build GRCh38
rs768924783 Chr.4: 70634474-70634474 on Build GRCh38
rs761239828 Chr.4: 70634521-70634521 on Build GRCh38
rs750910565 Chr.4: 70634410-70634410 on Build GRCh38
rs748423230 Chr.4: 70634394-70634395 on Build GRCh38
rs769829982 Chr.4: 70634373-70634373 on Build GRCh38
rs772524304 Chr.4: 70634495-70634495 on Build GRCh38
rs776515910 Chr.4: 70634325-70634327 on Build GRCh38
rs773076296 Chr.4: 70634369-70634369 on Build GRCh38
rs770975485 Chr.4: 70634272-70634272 on Build GRCh38
rs766814379 Chr.4: 70634311-70634311 on Build GRCh38
rs764490016 Chr.4: 70634313-70634313 on Build GRCh38
rs532584416 Chr.4: 70634429-70634429 on Build GRCh38
rs1018691670 Chr.4: 70634327-70634327 on Build GRCh38
rs774770647 Chr.4: 70634395-70634395 on Build GRCh38
rs778745635 Chr.4: 70634350-70634350 on Build GRCh38
rs565258194 Chr.4: 70634360-70634360 on Build GRCh38
rs544910871 Chr.4: 70634442-70634442 on Build GRCh38
rs374823061 Chr.4: 70634561-70634561 on Build GRCh38
rs563240539 Chr.4: 70634470-70634470 on Build GRCh38
rs777525143 Chr.4: 70634548-70634548 on Build GRCh38
rs903768178 Chr.4: 70634605-70634605 on Build GRCh38
rs373525837 Chr.4: 70634375-70634375 on Build GRCh38
rs765954002 Chr.4: 70634427-70634427 on Build GRCh38
rs924456253 Chr.4: 70634412-70634412 on Build GRCh38
rs200233913 Chr.4: 70634348-70634348 on Build GRCh38
rs765458823 Chr.4: 70634293-70634293 on Build GRCh38
rs888786519 Chr.4: 70634459-70634459 on Build GRCh38
rs752256214 Chr.4: 70634546-70634546 on Build GRCh38
rs755623510 Chr.4: 70634336-70634336 on Build GRCh38
rs749953744 Chr.4: 70634334-70634334 on Build GRCh38
rs966758447 Chr.4: 70634665-70634665 on Build GRCh38
rs377238799 Chr.4: 70634611-70634611 on Build GRCh38
rs747989947 Chr.4: 70634435-70634435 on Build GRCh38
rs149086531 Chr.4: 70634545-70634545 on Build GRCh38
rs34251790 Chr.4: 70634331-70634331 on Build GRCh38
rs771484720 Chr.4: 70634610-70634610 on Build GRCh38
rs774375347 Chr.4: 70634476-70634476 on Build GRCh38
rs778365039 Chr.4: 70634432-70634432 on Build GRCh38
rs762288141 Chr.4: 70634494-70634494 on Build GRCh38
rs764420953 Chr.4: 70634376-70634376 on Build GRCh38
rs949849858 Chr.4: 70634404-70634404 on Build GRCh38
rs755717210 Chr.4: 70634417-70634417 on Build GRCh38
rs201940627 Chr.4: 70634537-70634537 on Build GRCh38
rs145049132 Chr.4: 70634431-70634431 on Build GRCh38
rs758140330 Chr.4: 70634441-70634441 on Build GRCh38
rs769913792 Chr.4: 70634447-70634447 on Build GRCh38
rs773696608 Chr.4: 70634608-70634608 on Build GRCh38
rs188794413 Chr.4: 70634609-70634609 on Build GRCh38
rs753395224 Chr.4: 70634523-70634523 on Build GRCh38
rs1054090199 Chr.4: 70634250-70634250 on Build GRCh38
rs752058237 Chr.4: 70634319-70634319 on Build GRCh38
rs773896528 Chr.4: 70634507-70634507 on Build GRCh38
rs567213429 Chr.4: 70634589-70634589 on Build GRCh38
rs769865727 Chr.4: 70634398-70634398 on Build GRCh38
rs35554152 Chr.4: 70634603-70634603 on Build GRCh38
rs762013731 Chr.4: 70634400-70634400 on Build GRCh38
rs768470071 Chr.4: 70634338-70634338 on Build GRCh38
rs775645248 Chr.4: 70634297-70634297 on Build GRCh38
rs775629492 Chr.4: 70634454-70634454 on Build GRCh38
rs573297420 Chr.4: 70634329-70634329 on Build GRCh38
rs1052695462 Chr.4: 70634639-70634639 on Build GRCh38
rs757179405 Chr.4: 70634559-70634559 on Build GRCh38
rs759545856 Chr.4: 70634290-70634290 on Build GRCh38
rs891913969 Chr.4: 70634642-70634642 on Build GRCh38
rs758234051 Chr.4: 70634547-70634547 on Build GRCh38
rs554718725 Chr.4: 70634301-70634301 on Build GRCh38
rs779873942 Chr.4: 70634337-70634337 on Build GRCh38
rs992188058 Chr.4: 70634649-70634649 on Build GRCh38
rs199855542 Chr.4: 70634265-70634265 on Build GRCh38
rs748349248 Chr.4: 70634591-70634591 on Build GRCh38
rs748101666 Chr.4: 70634352-70634352 on Build GRCh38
rs978182243 Chr.4: 70634697-70634697 on Build GRCh38
rs992240660 Chr.4: 70634392-70634392 on Build GRCh38
rs907639982 Chr.4: 70634536-70634536 on Build GRCh38
rs370811268 Chr.4: 70634446-70634446 on Build GRCh38
rs1010705565 Chr.4: 70634701-70634701 on Build GRCh38
rs748930307 Chr.4: 70634342-70634342 on Build GRCh38
rs745972859 Chr.4: 70634371-70634371 on Build GRCh38
rs749395719 Chr.4: 70634567-70634567 on Build GRCh38
rs985348709 Chr.4: 70634526-70634526 on Build GRCh38
rs1026401742 Chr.4: 70634576-70634576 on Build GRCh38
rs959381485 Chr.4: 70634641-70634641 on Build GRCh38
rs202231676 Chr.4: 70634330-70634330 on Build GRCh38
rs776689511 Chr.4: 70634274-70634274 on Build GRCh38
rs758914802 Chr.4: 70634544-70634544 on Build GRCh38

View Full Product Details