Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 71745809-71746311 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GC
Entrez Gene ID
2638
Gene Name
GC, vitamin D binding protein
Gene Aliases
DBP, DBP/GC, GRD3, Gc-MAF, GcMAF, HEL-S-51, VDBG, VDBP
UniGene
Hs.418497
Gene Chromosome Location
Chr.4: 71741693-71805520 on Build GRCh38
Transcripts
NM_000583.3 NM_001204306.1 NM_001204307.1 XM_006714177.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6056745 c.1467G>A p.L489L Chr.4: 71746191-71746191 on Build GRCh38
1594584 c.1477C>A p.L493M Chr.4: 71746181-71746181 on Build GRCh38
4653830 c.1418T>C p.I473T Chr.4: 71746183-71746183 on Build GRCh38
6056746 c.1410G>A p.L470L Chr.4: 71746191-71746191 on Build GRCh38
1057200 c.1420C>A p.L474M Chr.4: 71746181-71746181 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187720029 Chr.4: 71745922-71745922 on Build GRCh38
rs1027003749 Chr.4: 71746174-71746174 on Build GRCh38
rs763545498 Chr.4: 71746253-71746253 on Build GRCh38
rs777052549 Chr.4: 71746224-71746224 on Build GRCh38
rs955432050 Chr.4: 71746103-71746103 on Build GRCh38
rs370532508 Chr.4: 71746160-71746160 on Build GRCh38
rs567906149 Chr.4: 71745928-71745928 on Build GRCh38
rs1009248049 Chr.4: 71746029-71746029 on Build GRCh38
rs917621673 Chr.4: 71745926-71745926 on Build GRCh38
rs1026471434 Chr.4: 71745840-71745840 on Build GRCh38
rs779820003 Chr.4: 71746188-71746188 on Build GRCh38
rs761301070 Chr.4: 71746228-71746228 on Build GRCh38
rs756905671 Chr.4: 71746172-71746172 on Build GRCh38
rs887913209 Chr.4: 71746180-71746180 on Build GRCh38
rs958997509 Chr.4: 71745903-71745903 on Build GRCh38
rs551043420 Chr.4: 71745843-71745843 on Build GRCh38
rs781391886 Chr.4: 71746098-71746098 on Build GRCh38
rs771293200 Chr.4: 71746215-71746215 on Build GRCh38
rs222045 Chr.4: 71745878-71745878 on Build GRCh38
rs755712569 Chr.4: 71746183-71746183 on Build GRCh38
rs191456559 Chr.4: 71746205-71746205 on Build GRCh38
rs1040381293 Chr.4: 71746120-71746120 on Build GRCh38
rs775811747 Chr.4: 71746249-71746249 on Build GRCh38
rs1054502265 Chr.4: 71745993-71745993 on Build GRCh38
rs537218225 Chr.4: 71746259-71746259 on Build GRCh38
rs111611682 Chr.4: 71746149-71746149 on Build GRCh38
rs761804967 Chr.4: 71746284-71746284 on Build GRCh38
rs945073565 Chr.4: 71746261-71746261 on Build GRCh38
rs77319027 Chr.4: 71745941-71745941 on Build GRCh38
rs376989511 Chr.4: 71746130-71746130 on Build GRCh38
rs903683190 Chr.4: 71746075-71746075 on Build GRCh38
rs764540894 Chr.4: 71746168-71746168 on Build GRCh38
rs113336164 Chr.4: 71746150-71746150 on Build GRCh38
rs760064008 Chr.4: 71746129-71746129 on Build GRCh38
rs367874733 Chr.4: 71746238-71746238 on Build GRCh38
rs375391497 Chr.4: 71745882-71745882 on Build GRCh38
rs776469725 Chr.4: 71746208-71746208 on Build GRCh38
rs222043 Chr.4: 71745982-71745982 on Build GRCh38
rs1042726317 Chr.4: 71745960-71745960 on Build GRCh38
rs842999 Chr.4: 71745973-71745973 on Build GRCh38
rs374292547 Chr.4: 71746229-71746229 on Build GRCh38
rs753313845 Chr.4: 71746148-71746148 on Build GRCh38
rs749300653 Chr.4: 71746199-71746199 on Build GRCh38
rs183398214 Chr.4: 71745873-71745873 on Build GRCh38
rs111451482 Chr.4: 71746008-71746008 on Build GRCh38
rs980715495 Chr.4: 71746204-71746204 on Build GRCh38
rs926377729 Chr.4: 71745880-71745880 on Build GRCh38
rs952327708 Chr.4: 71745846-71745846 on Build GRCh38
rs920066255 Chr.4: 71745891-71745891 on Build GRCh38
rs1008155306 Chr.4: 71746111-71746111 on Build GRCh38
rs1043480337 Chr.4: 71746062-71746062 on Build GRCh38
rs768471185 Chr.4: 71746155-71746155 on Build GRCh38
rs222044 Chr.4: 71745943-71745943 on Build GRCh38
rs1044637334 Chr.4: 71745934-71745934 on Build GRCh38
rs778802715 Chr.4: 71746213-71746213 on Build GRCh38
rs76732050 Chr.4: 71746011-71746011 on Build GRCh38
rs749918294 Chr.4: 71746178-71746178 on Build GRCh38
rs369345140 Chr.4: 71746121-71746121 on Build GRCh38
rs996670057 Chr.4: 71746079-71746079 on Build GRCh38
rs747163730 Chr.4: 71746214-71746214 on Build GRCh38

View Full Product Details