Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 88651892-88652397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HERC3
Entrez Gene ID
8916
Gene Name
HECT and RLD domain containing E3 ubiquitin protein ligase 3
Gene Aliases
-
UniGene
Hs.35804
Gene Chromosome Location
Chr.4: 88592423-88709303 on Build GRCh38
Transcripts
NM_001271602.1 NM_001318505.1 NM_014606.2 XM_005263327.3 XM_005263329.3 XM_005263330.3 XM_005263331.1 XM_005263332.3 XM_017008806.1 XM_017008807.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4690125 c.452C>T p.A151V Chr.4: 88652077-88652077 on Build GRCh38
6999885 c.458C>T p.A153V Chr.4: 88652083-88652083 on Build GRCh38
6861194 c.463+2T>G p.? Chr.4: 88652090-88652090 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761863880 Chr.4: 88651977-88651977 on Build GRCh38
rs757824365 Chr.4: 88652005-88652005 on Build GRCh38
rs753708459 Chr.4: 88652113-88652113 on Build GRCh38
rs377069890 Chr.4: 88652141-88652141 on Build GRCh38
rs771554401 Chr.4: 88651976-88651977 on Build GRCh38
rs772001461 Chr.4: 88652091-88652091 on Build GRCh38
rs776774314 Chr.4: 88652105-88652105 on Build GRCh38
rs565940188 Chr.4: 88652246-88652246 on Build GRCh38
rs769936214 Chr.4: 88652293-88652293 on Build GRCh38
rs751724169 Chr.4: 88651955-88651955 on Build GRCh38
rs986114663 Chr.4: 88652172-88652172 on Build GRCh38
rs746335012 Chr.4: 88652022-88652022 on Build GRCh38
rs993409140 Chr.4: 88651927-88651927 on Build GRCh38
rs983813907 Chr.4: 88652173-88652173 on Build GRCh38
rs764792053 Chr.4: 88651997-88651997 on Build GRCh38
rs1015036906 Chr.4: 88651938-88651938 on Build GRCh38
rs760628360 Chr.4: 88652098-88652098 on Build GRCh38
rs975099689 Chr.4: 88652130-88652130 on Build GRCh38
rs952478079 Chr.4: 88652161-88652161 on Build GRCh38
rs756619836 Chr.4: 88652062-88652062 on Build GRCh38
rs773426654 Chr.4: 88652094-88652094 on Build GRCh38
rs1019488483 Chr.4: 88652116-88652116 on Build GRCh38
rs752242358 Chr.4: 88652118-88652118 on Build GRCh38
rs777365529 Chr.4: 88652021-88652021 on Build GRCh38
rs143699412 Chr.4: 88652084-88652084 on Build GRCh38
rs894442170 Chr.4: 88652326-88652326 on Build GRCh38
rs375161830 Chr.4: 88652078-88652078 on Build GRCh38
rs10015739 Chr.4: 88652119-88652119 on Build GRCh38
rs763447717 Chr.4: 88652132-88652132 on Build GRCh38
rs773231730 Chr.4: 88651964-88651964 on Build GRCh38
rs911965414 Chr.4: 88652194-88652194 on Build GRCh38
rs745355480 Chr.4: 88651972-88651972 on Build GRCh38
rs537769608 Chr.4: 88652261-88652261 on Build GRCh38
rs759220213 Chr.4: 88652109-88652109 on Build GRCh38
rs752434591 Chr.4: 88651998-88651998 on Build GRCh38
rs548383989 Chr.4: 88652342-88652342 on Build GRCh38
rs557348723 Chr.4: 88652316-88652316 on Build GRCh38
rs1055426602 Chr.4: 88652348-88652348 on Build GRCh38
rs192303165 Chr.4: 88652099-88652099 on Build GRCh38
rs780340195 Chr.4: 88652069-88652069 on Build GRCh38
rs928892819 Chr.4: 88652270-88652270 on Build GRCh38
rs528617177 Chr.4: 88651949-88651949 on Build GRCh38
rs1011438108 Chr.4: 88652097-88652097 on Build GRCh38
rs532461561 Chr.4: 88652101-88652101 on Build GRCh38
rs201105775 Chr.4: 88652070-88652070 on Build GRCh38
rs909400212 Chr.4: 88652189-88652189 on Build GRCh38
rs1043014174 Chr.4: 88652324-88652324 on Build GRCh38
rs940830309 Chr.4: 88652225-88652225 on Build GRCh38
rs1040934792 Chr.4: 88652269-88652269 on Build GRCh38
rs373556353 Chr.4: 88652304-88652304 on Build GRCh38
rs9995890 Chr.4: 88652329-88652329 on Build GRCh38
rs754876351 Chr.4: 88651982-88651982 on Build GRCh38
rs766207290 Chr.4: 88651976-88651976 on Build GRCh38
rs751085164 Chr.4: 88652140-88652140 on Build GRCh38
rs75814248 Chr.4: 88652090-88652090 on Build GRCh38

View Full Product Details