Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 88655746-88656213 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HERC3
Entrez Gene ID
8916
Gene Name
HECT and RLD domain containing E3 ubiquitin protein ligase 3
Gene Aliases
-
UniGene
Hs.35804
Gene Chromosome Location
Chr.4: 88592423-88709303 on Build GRCh38
Transcripts
NM_001271602.1 NM_001318505.1 NM_014606.2 XM_005263327.3 XM_005263329.3 XM_005263330.3 XM_005263331.1 XM_005263332.3 XM_017008806.1 XM_017008807.1 XM_017008808.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7371032 c.1008G>T p.K336N Chr.4: 88655974-88655974 on Build GRCh38
6679442 c.932C>T p.P311L Chr.4: 88655898-88655898 on Build GRCh38
481672 c.950A>G p.Y317C Chr.4: 88655916-88655916 on Build GRCh38
1486161 c.940G>A p.G314R Chr.4: 88655906-88655906 on Build GRCh38
1209597 c.978A>C p.Q326H Chr.4: 88655944-88655944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs538167482 Chr.4: 88655996-88655996 on Build GRCh38
rs763143167 Chr.4: 88656082-88656082 on Build GRCh38
rs1000700656 Chr.4: 88655822-88655822 on Build GRCh38
rs780986349 Chr.4: 88655969-88655969 on Build GRCh38
rs759601114 Chr.4: 88655916-88655916 on Build GRCh38
rs138394673 Chr.4: 88655834-88655834 on Build GRCh38
rs765262466 Chr.4: 88656060-88656060 on Build GRCh38
rs201937749 Chr.4: 88655897-88655897 on Build GRCh38
rs200532115 Chr.4: 88655971-88655971 on Build GRCh38
rs552306781 Chr.4: 88655792-88655792 on Build GRCh38
rs763900260 Chr.4: 88655944-88655944 on Build GRCh38
rs553928306 Chr.4: 88656032-88656032 on Build GRCh38
rs755625380 Chr.4: 88656012-88656012 on Build GRCh38
rs567830574 Chr.4: 88655793-88655793 on Build GRCh38
rs151018103 Chr.4: 88655881-88655881 on Build GRCh38
rs767431411 Chr.4: 88656112-88656112 on Build GRCh38
rs751290394 Chr.4: 88655956-88655956 on Build GRCh38
rs1053632512 Chr.4: 88656171-88656171 on Build GRCh38
rs765315774 Chr.4: 88655920-88655920 on Build GRCh38
rs926677006 Chr.4: 88656135-88656135 on Build GRCh38
rs756988628 Chr.4: 88656104-88656104 on Build GRCh38
rs757114976 Chr.4: 88655966-88655966 on Build GRCh38
rs771388611 Chr.4: 88656030-88656030 on Build GRCh38
rs73841610 Chr.4: 88656164-88656164 on Build GRCh38
rs773813906 Chr.4: 88656027-88656027 on Build GRCh38
rs192424672 Chr.4: 88655850-88655850 on Build GRCh38
rs866881807 Chr.4: 88656150-88656150 on Build GRCh38
rs147433297 Chr.4: 88656093-88656093 on Build GRCh38
rs766483118 Chr.4: 88655894-88655894 on Build GRCh38
rs772474731 Chr.4: 88656020-88656020 on Build GRCh38
rs949501671 Chr.4: 88655789-88655789 on Build GRCh38
rs779712451 Chr.4: 88656016-88656016 on Build GRCh38
rs905062711 Chr.4: 88655819-88655819 on Build GRCh38
rs1045213999 Chr.4: 88656108-88656108 on Build GRCh38
rs774983935 Chr.4: 88655826-88655826 on Build GRCh38
rs73841608 Chr.4: 88655980-88655980 on Build GRCh38
rs761154826 Chr.4: 88655893-88655893 on Build GRCh38
rs964217572 Chr.4: 88656005-88656005 on Build GRCh38
rs759105522 Chr.4: 88655807-88655810 on Build GRCh38
rs1007270484 Chr.4: 88655964-88655964 on Build GRCh38
rs758300366 Chr.4: 88655943-88655943 on Build GRCh38
rs1003640966 Chr.4: 88655801-88655801 on Build GRCh38
rs747334811 Chr.4: 88656029-88656029 on Build GRCh38
rs549564675 Chr.4: 88655771-88655771 on Build GRCh38
rs763905360 Chr.4: 88656085-88656085 on Build GRCh38
rs139661725 Chr.4: 88656156-88656156 on Build GRCh38
rs1047042923 Chr.4: 88655769-88655769 on Build GRCh38
rs776779224 Chr.4: 88656054-88656054 on Build GRCh38
rs748645971 Chr.4: 88656019-88656019 on Build GRCh38
rs2972040 Chr.4: 88656063-88656063 on Build GRCh38
rs896447701 Chr.4: 88655779-88655779 on Build GRCh38

View Full Product Details