Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 152829445-152829974 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARFIP1
Entrez Gene ID
27236
Gene Name
ADP ribosylation factor interacting protein 1
Gene Aliases
HSU52521
UniGene
Hs.416089
Gene Chromosome Location
Chr.4: 152779937-152911912 on Build GRCh38
Transcripts
NM_001025593.2 NM_001025595.2 NM_001287431.1 NM_001287432.1 NM_001287433.1 NM_014447.3 XM_011531869.2 XM_011531870.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3600955 c.40C>T p.P14S Chr.4: 152829673-152829673 on Build GRCh38
5865884 c.16_17CC>TT p.P6F Chr.4: 152829649-152829650 on Build GRCh38
6898115 c.93G>A p.R31R Chr.4: 152829726-152829726 on Build GRCh38
5865886 c.17C>T p.P6L Chr.4: 152829650-152829650 on Build GRCh38
4909744 c.58G>A p.E20K Chr.4: 152829691-152829691 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750927874 Chr.4: 152829595-152829596 on Build GRCh38
rs746436354 Chr.4: 152829684-152829684 on Build GRCh38
rs547462540 Chr.4: 152829613-152829613 on Build GRCh38
rs759984298 Chr.4: 152829595-152829595 on Build GRCh38
rs550011612 Chr.4: 152829713-152829713 on Build GRCh38
rs895932238 Chr.4: 152829786-152829786 on Build GRCh38
rs753491075 Chr.4: 152829760-152829760 on Build GRCh38
rs752258619 Chr.4: 152829632-152829632 on Build GRCh38
rs755204314 Chr.4: 152829602-152829603 on Build GRCh38
rs897743177 Chr.4: 152829564-152829564 on Build GRCh38
rs930604777 Chr.4: 152829879-152829879 on Build GRCh38
rs751473678 Chr.4: 152829729-152829729 on Build GRCh38
rs772724793 Chr.4: 152829772-152829772 on Build GRCh38
rs954354610 Chr.4: 152829688-152829688 on Build GRCh38
rs772544773 Chr.4: 152829686-152829686 on Build GRCh38
rs199986353 Chr.4: 152829722-152829722 on Build GRCh38
rs369510451 Chr.4: 152829682-152829682 on Build GRCh38
rs767259177 Chr.4: 152829735-152829735 on Build GRCh38
rs759507226 Chr.4: 152829733-152829733 on Build GRCh38
rs748423090 Chr.4: 152829610-152829613 on Build GRCh38
rs985763212 Chr.4: 152829920-152829920 on Build GRCh38
rs140758872 Chr.4: 152829762-152829762 on Build GRCh38
rs10010297 Chr.4: 152829766-152829766 on Build GRCh38
rs370236805 Chr.4: 152829598-152829598 on Build GRCh38
rs947630102 Chr.4: 152829750-152829750 on Build GRCh38
rs771491000 Chr.4: 152829585-152829585 on Build GRCh38
rs1012309715 Chr.4: 152829628-152829628 on Build GRCh38
rs201260045 Chr.4: 152829526-152829526 on Build GRCh38
rs1027019708 Chr.4: 152829596-152829596 on Build GRCh38
rs1016060316 Chr.4: 152829856-152829856 on Build GRCh38
rs776503493 Chr.4: 152829711-152829711 on Build GRCh38
rs1006559309 Chr.4: 152829842-152829842 on Build GRCh38
rs1023693032 Chr.4: 152829629-152829629 on Build GRCh38
rs529283440 Chr.4: 152829610-152829610 on Build GRCh38
rs931755392 Chr.4: 152829491-152829491 on Build GRCh38
rs577802620 Chr.4: 152829763-152829763 on Build GRCh38
rs748844373 Chr.4: 152829674-152829674 on Build GRCh38
rs1023493567 Chr.4: 152829521-152829521 on Build GRCh38
rs79395166 Chr.4: 152829773-152829773 on Build GRCh38
rs183711375 Chr.4: 152829601-152829601 on Build GRCh38
rs759038565 Chr.4: 152829555-152829555 on Build GRCh38
rs771499421 Chr.4: 152829681-152829681 on Build GRCh38
rs374989924 Chr.4: 152829651-152829651 on Build GRCh38
rs768061896 Chr.4: 152829597-152829597 on Build GRCh38
rs750952818 Chr.4: 152829508-152829508 on Build GRCh38
rs761009754 Chr.4: 152829600-152829600 on Build GRCh38
rs1049431543 Chr.4: 152829897-152829897 on Build GRCh38
rs115502962 Chr.4: 152829540-152829540 on Build GRCh38
rs897653429 Chr.4: 152829857-152829857 on Build GRCh38
rs781012339 Chr.4: 152829623-152829623 on Build GRCh38
rs202111498 Chr.4: 152829687-152829687 on Build GRCh38
rs897643738 Chr.4: 152829835-152829835 on Build GRCh38
rs780398939 Chr.4: 152829601-152829602 on Build GRCh38
rs993380723 Chr.4: 152829860-152829860 on Build GRCh38
rs954460567 Chr.4: 152829914-152829914 on Build GRCh38
rs890319308 Chr.4: 152829509-152829509 on Build GRCh38
rs753985869 Chr.4: 152829603-152829603 on Build GRCh38
rs12505637 Chr.4: 152829928-152829928 on Build GRCh38
rs1030128267 Chr.4: 152829640-152829640 on Build GRCh38
rs948704130 Chr.4: 152829796-152829796 on Build GRCh38
rs777097416 Chr.4: 152829660-152829660 on Build GRCh38
rs752569609 Chr.4: 152829736-152829736 on Build GRCh38
rs768949193 Chr.4: 152829707-152829707 on Build GRCh38
rs774798384 Chr.4: 152829593-152829593 on Build GRCh38
rs751010670 Chr.4: 152829704-152829704 on Build GRCh38
rs751561439 Chr.4: 152829618-152829618 on Build GRCh38
rs755820775 Chr.4: 152829659-152829659 on Build GRCh38
rs370284819 Chr.4: 152829741-152829741 on Build GRCh38
rs995158032 Chr.4: 152829578-152829578 on Build GRCh38
rs754750805 Chr.4: 152829619-152829619 on Build GRCh38
rs775966703 Chr.4: 152829703-152829703 on Build GRCh38
rs905091528 Chr.4: 152829819-152829819 on Build GRCh38
rs1006030186 Chr.4: 152829570-152829570 on Build GRCh38
rs1036235179 Chr.4: 152829830-152829830 on Build GRCh38
rs747392701 Chr.4: 152829603-152829603 on Build GRCh38
rs1017286890 Chr.4: 152829947-152829947 on Build GRCh38
rs377362694 Chr.4: 152829631-152829631 on Build GRCh38
rs895817774 Chr.4: 152829497-152829497 on Build GRCh38
rs376458448 Chr.4: 152829723-152829723 on Build GRCh38
rs145739616 Chr.4: 152829913-152829913 on Build GRCh38
rs540170800 Chr.4: 152829802-152829802 on Build GRCh38
rs1057460787 Chr.4: 152829781-152829781 on Build GRCh38
rs145188297 Chr.4: 152829706-152829706 on Build GRCh38

View Full Product Details