Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 863561-864054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD9
Entrez Gene ID
65980
Gene Name
bromodomain containing 9
Gene Aliases
LAVS3040, PRO9856
UniGene
Hs.449278
Gene Chromosome Location
Chr.5: 863735-892824 on Build GRCh38
Transcripts
NM_001009877.2 NM_001317951.1 NM_023924.4 NR_134296.1 XM_017009870.1 XM_017009871.1

Gene Symbol
ZDHHC11
Entrez Gene ID
79844
Gene Name
zinc finger DHHC-type containing 11
Gene Aliases
ZNF399
UniGene
Hs.654874
Gene Chromosome Location
Chr.5: 795605-863634 on Build GRCh38
Transcripts
NM_001009877.2 NM_001317951.1 NM_023924.4 NR_134296.1 XM_017009870.1 XM_017009871.1

SNP Detail

SNP ID Chromosome Location
rs536443015 Chr.5: 863667-863668 on Build GRCh38
rs13436859 Chr.5: 863682-863682 on Build GRCh38
rs1027330535 Chr.5: 864006-864006 on Build GRCh38
rs192869078 Chr.5: 863684-863684 on Build GRCh38
rs536148184 Chr.5: 863940-863940 on Build GRCh38
rs531031437 Chr.5: 863857-863857 on Build GRCh38
rs536466636 Chr.5: 863767-863767 on Build GRCh38
rs13436651 Chr.5: 864017-864017 on Build GRCh38
rs1042246955 Chr.5: 863908-863908 on Build GRCh38
rs976112453 Chr.5: 863973-863973 on Build GRCh38
rs541690319 Chr.5: 863785-863785 on Build GRCh38
rs189909985 Chr.5: 863745-863745 on Build GRCh38
rs138372628 Chr.5: 863768-863768 on Build GRCh38
rs1054867050 Chr.5: 863708-863708 on Build GRCh38
rs547782361 Chr.5: 863942-863942 on Build GRCh38
rs936482107 Chr.5: 863813-863813 on Build GRCh38
rs1063602 Chr.5: 863830-863830 on Build GRCh38
rs893760122 Chr.5: 863878-863878 on Build GRCh38
rs560061985 Chr.5: 863796-863796 on Build GRCh38
rs1024561082 Chr.5: 863586-863586 on Build GRCh38
rs1010786989 Chr.5: 863887-863887 on Build GRCh38
rs964423622 Chr.5: 863800-863800 on Build GRCh38
rs577966027 Chr.5: 863993-863993 on Build GRCh38
rs575227478 Chr.5: 863810-863810 on Build GRCh38
rs1040306267 Chr.5: 863666-863666 on Build GRCh38
rs1013251965 Chr.5: 863734-863734 on Build GRCh38
rs952266059 Chr.5: 863818-863818 on Build GRCh38
rs569355495 Chr.5: 863971-863971 on Build GRCh38
rs145316738 Chr.5: 863855-863855 on Build GRCh38
rs964812310 Chr.5: 863980-863980 on Build GRCh38
rs750427155 Chr.5: 864022-864022 on Build GRCh38
rs565313103 Chr.5: 863700-863700 on Build GRCh38
rs774929855 Chr.5: 863772-863772 on Build GRCh38
rs984813618 Chr.5: 863637-863637 on Build GRCh38
rs1024613428 Chr.5: 863736-863736 on Build GRCh38
rs1030345128 Chr.5: 864015-864015 on Build GRCh38
rs967100428 Chr.5: 864016-864017 on Build GRCh38
rs563737940 Chr.5: 863591-863591 on Build GRCh38
rs887993270 Chr.5: 863868-863868 on Build GRCh38
rs532232709 Chr.5: 863888-863888 on Build GRCh38
rs1045826310 Chr.5: 863838-863838 on Build GRCh38
rs905880641 Chr.5: 863761-863761 on Build GRCh38
rs13436858 Chr.5: 863585-863585 on Build GRCh38
rs571397812 Chr.5: 863662-863662 on Build GRCh38
rs150493681 Chr.5: 863634-863634 on Build GRCh38
rs1047923303 Chr.5: 863668-863668 on Build GRCh38
rs908035709 Chr.5: 863832-863832 on Build GRCh38
rs987577503 Chr.5: 863615-863615 on Build GRCh38
rs972007757 Chr.5: 863608-863608 on Build GRCh38
rs781504730 Chr.5: 863849-863849 on Build GRCh38
rs1046115311 Chr.5: 863841-863841 on Build GRCh38
rs184913588 Chr.5: 863724-863724 on Build GRCh38
rs180764534 Chr.5: 863774-863774 on Build GRCh38
rs141557050 Chr.5: 863769-863769 on Build GRCh38
rs886556394 Chr.5: 863679-863679 on Build GRCh38
rs971676685 Chr.5: 863743-863743 on Build GRCh38
rs930821135 Chr.5: 863665-863665 on Build GRCh38
rs923223842 Chr.5: 863799-863799 on Build GRCh38
rs73730991 Chr.5: 863881-863881 on Build GRCh38
rs923182076 Chr.5: 863671-863671 on Build GRCh38
rs559100331 Chr.5: 863773-863773 on Build GRCh38
rs950238073 Chr.5: 863837-863837 on Build GRCh38
rs903661271 Chr.5: 863698-863698 on Build GRCh38
rs16877782 Chr.5: 863938-863938 on Build GRCh38
rs112501833 Chr.5: 863815-863815 on Build GRCh38
rs978056988 Chr.5: 864016-864016 on Build GRCh38
rs547208609 Chr.5: 863911-863911 on Build GRCh38
rs73018873 Chr.5: 863635-863635 on Build GRCh38
rs975011707 Chr.5: 863660-863660 on Build GRCh38
rs925169315 Chr.5: 863811-863811 on Build GRCh38
rs552430710 Chr.5: 863863-863863 on Build GRCh38
rs945250816 Chr.5: 863677-863677 on Build GRCh38
rs966912511 Chr.5: 863780-863780 on Build GRCh38

View Full Product Details