Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 37122097-37122617 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf42
Entrez Gene ID
65250
Gene Name
chromosome 5 open reading frame 42
Gene Aliases
Hug, JBTS17, OFD6
UniGene
Hs.586199
Gene Chromosome Location
Chr.5: 37063928-37249428 on Build GRCh38
Transcripts
NM_023073.3 XM_005248345.3 XM_005248346.3 XM_005248347.3 XM_005248349.3 XM_005248350.3 XM_006714491.3 XM_011514085.2 XM_011514086.2 XM_011514087.2 XM_011514088.2 XM_011514089.2 XM_011514090.2 XM_011514092.2 XM_011514094.2 XM_017009760.1 XM_017009761.1 XM_017009762.1 XM_017009763.1 XM_017009764.1 XM_017009765.1 XM_017009766.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4561363 c.5507G>A p.R1836K Chr.5: 37122472-37122472 on Build GRCh38
6038800 c.5539G>T p.E1847* Chr.5: 37122440-37122440 on Build GRCh38
2688154 c.5499G>A p.M1833I Chr.5: 37122480-37122480 on Build GRCh38
2688153 c.8805G>A p.M2935I Chr.5: 37122480-37122480 on Build GRCh38
127087 c.5549+1G>T p.? Chr.5: 37122429-37122429 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs967055027 Chr.5: 37122170-37122170 on Build GRCh38
rs532477255 Chr.5: 37122188-37122188 on Build GRCh38
rs771246667 Chr.5: 37122313-37122313 on Build GRCh38
rs767604260 Chr.5: 37122526-37122526 on Build GRCh38
rs1019583712 Chr.5: 37122254-37122254 on Build GRCh38
rs764211169 Chr.5: 37122494-37122494 on Build GRCh38
rs989847255 Chr.5: 37122133-37122133 on Build GRCh38
rs863225154 Chr.5: 37122429-37122429 on Build GRCh38
rs759378976 Chr.5: 37122466-37122466 on Build GRCh38
rs926372895 Chr.5: 37122571-37122571 on Build GRCh38
rs145867174 Chr.5: 37122471-37122471 on Build GRCh38
rs369404796 Chr.5: 37122374-37122374 on Build GRCh38
rs775563318 Chr.5: 37122482-37122482 on Build GRCh38
rs544926381 Chr.5: 37122587-37122587 on Build GRCh38
rs113214197 Chr.5: 37122433-37122433 on Build GRCh38
rs374386158 Chr.5: 37122440-37122440 on Build GRCh38
rs750480289 Chr.5: 37122204-37122204 on Build GRCh38
rs375394384 Chr.5: 37122528-37122528 on Build GRCh38
rs924779439 Chr.5: 37122462-37122462 on Build GRCh38
rs958181845 Chr.5: 37122537-37122537 on Build GRCh38
rs975465326 Chr.5: 37122357-37122357 on Build GRCh38
rs371727527 Chr.5: 37122503-37122503 on Build GRCh38
rs571350425 Chr.5: 37122135-37122135 on Build GRCh38
rs148987541 Chr.5: 37122448-37122448 on Build GRCh38
rs1037772115 Chr.5: 37122343-37122343 on Build GRCh38
rs977781565 Chr.5: 37122182-37122182 on Build GRCh38
rs566931989 Chr.5: 37122444-37122444 on Build GRCh38
rs949140790 Chr.5: 37122134-37122134 on Build GRCh38
rs897860506 Chr.5: 37122368-37122368 on Build GRCh38
rs1044742341 Chr.5: 37122551-37122551 on Build GRCh38
rs949058727 Chr.5: 37122514-37122514 on Build GRCh38
rs757963426 Chr.5: 37122419-37122419 on Build GRCh38
rs116198390 Chr.5: 37122431-37122431 on Build GRCh38
rs749313183 Chr.5: 37122536-37122536 on Build GRCh38
rs777968527 Chr.5: 37122533-37122536 on Build GRCh38
rs757462684 Chr.5: 37122519-37122519 on Build GRCh38
rs556601819 Chr.5: 37122321-37122321 on Build GRCh38
rs965779676 Chr.5: 37122329-37122329 on Build GRCh38
rs577359016 Chr.5: 37122556-37122556 on Build GRCh38
rs569302817 Chr.5: 37122208-37122208 on Build GRCh38
rs1004545394 Chr.5: 37122213-37122214 on Build GRCh38
rs367827156 Chr.5: 37122501-37122501 on Build GRCh38
rs376997400 Chr.5: 37122588-37122588 on Build GRCh38
rs536621339 Chr.5: 37122214-37122214 on Build GRCh38
rs550585886 Chr.5: 37122192-37122192 on Build GRCh38
rs942147266 Chr.5: 37122280-37122280 on Build GRCh38
rs916007621 Chr.5: 37122237-37122237 on Build GRCh38
rs771569470 Chr.5: 37122383-37122383 on Build GRCh38
rs74950744 Chr.5: 37122489-37122489 on Build GRCh38

View Full Product Details