Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138468128-138468630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGR1
Entrez Gene ID
1958
Gene Name
early growth response 1
Gene Aliases
AT225, G0S30, KROX-24, NGFI-A, TIS8, ZIF-268, ZNF225
UniGene
Hs.326035
Gene Chromosome Location
Chr.5: 138465492-138469315 on Build GRCh38
Transcripts
NM_001964.2

SNP Detail

SNP ID Chromosome Location
rs188002849 Chr.5: 138468278-138468278 on Build GRCh38
rs201135481 Chr.5: 138468262-138468262 on Build GRCh38
rs898940454 Chr.5: 138468209-138468209 on Build GRCh38
rs1022790337 Chr.5: 138468423-138468423 on Build GRCh38
rs202201976 Chr.5: 138468268-138468268 on Build GRCh38
rs921766656 Chr.5: 138468549-138468549 on Build GRCh38
rs955386560 Chr.5: 138468430-138468430 on Build GRCh38
rs200874249 Chr.5: 138468221-138468221 on Build GRCh38
rs200025697 Chr.5: 138468213-138468213 on Build GRCh38
rs199797866 Chr.5: 138468387-138468387 on Build GRCh38
rs776725459 Chr.5: 138468393-138468393 on Build GRCh38
rs761470879 Chr.5: 138468586-138468586 on Build GRCh38
rs200583571 Chr.5: 138468485-138468485 on Build GRCh38
rs995383525 Chr.5: 138468212-138468212 on Build GRCh38
rs200394040 Chr.5: 138468334-138468334 on Build GRCh38
rs199627249 Chr.5: 138468433-138468433 on Build GRCh38
rs908313859 Chr.5: 138468514-138468514 on Build GRCh38
rs759581124 Chr.5: 138468153-138468153 on Build GRCh38
rs2294175 Chr.5: 138468304-138468304 on Build GRCh38
rs200295874 Chr.5: 138468303-138468303 on Build GRCh38
rs546954332 Chr.5: 138468167-138468167 on Build GRCh38
rs962414905 Chr.5: 138468550-138468550 on Build GRCh38
rs201306291 Chr.5: 138468157-138468157 on Build GRCh38
rs1042197 Chr.5: 138468598-138468598 on Build GRCh38
rs534036144 Chr.5: 138468599-138468600 on Build GRCh38
rs200950069 Chr.5: 138468590-138468590 on Build GRCh38
rs1009594134 Chr.5: 138468394-138468394 on Build GRCh38
rs893194804 Chr.5: 138468159-138468159 on Build GRCh38
rs758759231 Chr.5: 138468275-138468275 on Build GRCh38
rs978980129 Chr.5: 138468506-138468506 on Build GRCh38
rs964693958 Chr.5: 138468265-138468265 on Build GRCh38
rs200648293 Chr.5: 138468443-138468443 on Build GRCh38
rs199889910 Chr.5: 138468190-138468190 on Build GRCh38
rs201202983 Chr.5: 138468418-138468418 on Build GRCh38
rs780082755 Chr.5: 138468175-138468175 on Build GRCh38
rs1003758725 Chr.5: 138468273-138468273 on Build GRCh38
rs201027575 Chr.5: 138468372-138468372 on Build GRCh38
rs555058438 Chr.5: 138468544-138468544 on Build GRCh38
rs202053052 Chr.5: 138468446-138468446 on Build GRCh38
rs200283860 Chr.5: 138468602-138468602 on Build GRCh38
rs201807055 Chr.5: 138468586-138468586 on Build GRCh38
rs200398803 Chr.5: 138468408-138468408 on Build GRCh38
rs201213451 Chr.5: 138468585-138468585 on Build GRCh38
rs138171053 Chr.5: 138468258-138468258 on Build GRCh38
rs199660024 Chr.5: 138468453-138468453 on Build GRCh38
rs968173802 Chr.5: 138468431-138468431 on Build GRCh38
rs200993915 Chr.5: 138468193-138468193 on Build GRCh38
rs201616201 Chr.5: 138468307-138468307 on Build GRCh38
rs879193807 Chr.5: 138468317-138468317 on Build GRCh38
rs201870338 Chr.5: 138468236-138468236 on Build GRCh38
rs747195555 Chr.5: 138468346-138468346 on Build GRCh38
rs1803556 Chr.5: 138468552-138468552 on Build GRCh38
rs201567147 Chr.5: 138468581-138468581 on Build GRCh38
rs200019675 Chr.5: 138468587-138468587 on Build GRCh38
rs199900872 Chr.5: 138468584-138468584 on Build GRCh38
rs201813217 Chr.5: 138468195-138468195 on Build GRCh38

View Full Product Details