Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140127345-140127846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGIP
Entrez Gene ID
492311
Gene Name
IgA inducing protein
Gene Aliases
C5orf53
UniGene
Hs.696360
Gene Chromosome Location
Chr.5: 140125936-140128806 on Build GRCh38
Transcripts
NM_001007189.1

SNP Detail

SNP ID Chromosome Location
rs531261457 Chr.5: 140127760-140127760 on Build GRCh38
rs911373033 Chr.5: 140127676-140127676 on Build GRCh38
rs1040173268 Chr.5: 140127373-140127373 on Build GRCh38
rs898146538 Chr.5: 140127691-140127691 on Build GRCh38
rs976765398 Chr.5: 140127708-140127708 on Build GRCh38
rs1004126275 Chr.5: 140127633-140127633 on Build GRCh38
rs1050952866 Chr.5: 140127615-140127615 on Build GRCh38
rs985686834 Chr.5: 140127652-140127652 on Build GRCh38
rs971179475 Chr.5: 140127747-140127747 on Build GRCh38
rs543108555 Chr.5: 140127794-140127794 on Build GRCh38
rs1038325874 Chr.5: 140127644-140127644 on Build GRCh38
rs567377901 Chr.5: 140127823-140127823 on Build GRCh38
rs879128085 Chr.5: 140127433-140127433 on Build GRCh38
rs937246440 Chr.5: 140127817-140127818 on Build GRCh38
rs200535818 Chr.5: 140127581-140127581 on Build GRCh38
rs778678613 Chr.5: 140127495-140127496 on Build GRCh38
rs865929424 Chr.5: 140127809-140127809 on Build GRCh38
rs1023109321 Chr.5: 140127704-140127704 on Build GRCh38
rs868471689 Chr.5: 140127660-140127660 on Build GRCh38
rs574207746 Chr.5: 140127448-140127448 on Build GRCh38
rs759175373 Chr.5: 140127465-140127465 on Build GRCh38
rs1012607915 Chr.5: 140127703-140127703 on Build GRCh38
rs772704091 Chr.5: 140127626-140127626 on Build GRCh38
rs918600391 Chr.5: 140127746-140127746 on Build GRCh38
rs946747042 Chr.5: 140127369-140127369 on Build GRCh38
rs1048856688 Chr.5: 140127777-140127777 on Build GRCh38
rs909838886 Chr.5: 140127815-140127815 on Build GRCh38
rs775795659 Chr.5: 140127817-140127817 on Build GRCh38
rs921374581 Chr.5: 140127403-140127403 on Build GRCh38
rs560974771 Chr.5: 140127367-140127367 on Build GRCh38

View Full Product Details