Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 149354058-149354598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRPEL2
Entrez Gene ID
134266
Gene Name
GrpE like 2, mitochondrial
Gene Aliases
Mt-GrpE#2
UniGene
Hs.511816
Gene Chromosome Location
Chr.5: 149345414-149354583 on Build GRCh38
Transcripts
NM_152407.3 NR_132366.1

Gene Symbol
GRPEL2-AS1
Entrez Gene ID
106144529
Gene Name
GRPEL2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.607162
Gene Chromosome Location
Chr.5: 149348116-149357642 on Build GRCh38
Transcripts
NM_152407.3 NR_132366.1

SNP Detail

SNP ID Chromosome Location
rs908825070 Chr.5: 149354106-149354106 on Build GRCh38
rs951567587 Chr.5: 149354334-149354334 on Build GRCh38
rs925705418 Chr.5: 149354357-149354357 on Build GRCh38
rs141681853 Chr.5: 149354429-149354429 on Build GRCh38
rs920894751 Chr.5: 149354206-149354206 on Build GRCh38
rs1012663114 Chr.5: 149354187-149354187 on Build GRCh38
rs553776184 Chr.5: 149354275-149354275 on Build GRCh38
rs538064719 Chr.5: 149354278-149354278 on Build GRCh38
rs1014086198 Chr.5: 149354544-149354544 on Build GRCh38
rs550756143 Chr.5: 149354369-149354369 on Build GRCh38
rs982954611 Chr.5: 149354098-149354098 on Build GRCh38
rs550404639 Chr.5: 149354349-149354349 on Build GRCh38
rs752401823 Chr.5: 149354397-149354397 on Build GRCh38
rs975503530 Chr.5: 149354411-149354411 on Build GRCh38
rs35528284 Chr.5: 149354085-149354085 on Build GRCh38
rs778375098 Chr.5: 149354244-149354244 on Build GRCh38
rs963147344 Chr.5: 149354392-149354392 on Build GRCh38
rs750822018 Chr.5: 149354140-149354140 on Build GRCh38
rs965614168 Chr.5: 149354122-149354122 on Build GRCh38
rs1024254123 Chr.5: 149354230-149354230 on Build GRCh38
rs895776733 Chr.5: 149354459-149354459 on Build GRCh38
rs565615870 Chr.5: 149354522-149354522 on Build GRCh38
rs892918883 Chr.5: 149354147-149354147 on Build GRCh38
rs933839959 Chr.5: 149354518-149354518 on Build GRCh38
rs983228112 Chr.5: 149354358-149354358 on Build GRCh38
rs1035277155 Chr.5: 149354326-149354326 on Build GRCh38
rs539572025 Chr.5: 149354566-149354566 on Build GRCh38
rs571015978 Chr.5: 149354231-149354231 on Build GRCh38
rs552036179 Chr.5: 149354203-149354203 on Build GRCh38
rs986563939 Chr.5: 149354564-149354564 on Build GRCh38
rs1016928116 Chr.5: 149354364-149354368 on Build GRCh38
rs147927292 Chr.5: 149354376-149354376 on Build GRCh38

View Full Product Details