Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157237973-157238479 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITK
Entrez Gene ID
3702
Gene Name
IL2 inducible T-cell kinase
Gene Aliases
EMT, LPFS1, LYK, PSCTK2
UniGene
Hs.558348
Gene Chromosome Location
Chr.5: 157180896-157255185 on Build GRCh38
Transcripts
NM_005546.3 XM_017009443.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
377678 c.786C>T p.F262F Chr.5: 157238126-157238126 on Build GRCh38
6688552 c.774A>C p.K258N Chr.5: 157238114-157238114 on Build GRCh38
71223 c.781G>C p.A261P Chr.5: 157238121-157238121 on Build GRCh38
6244219 c.804G>T p.R268S Chr.5: 157238144-157238144 on Build GRCh38
4389238 c.823G>A p.V275M Chr.5: 157238163-157238163 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772820944 Chr.5: 157238209-157238209 on Build GRCh38
rs761415992 Chr.5: 157238062-157238062 on Build GRCh38
rs539907506 Chr.5: 157238441-157238442 on Build GRCh38
rs200351890 Chr.5: 157238162-157238162 on Build GRCh38
rs538711147 Chr.5: 157238058-157238058 on Build GRCh38
rs762473738 Chr.5: 157238221-157238221 on Build GRCh38
rs528697627 Chr.5: 157238392-157238392 on Build GRCh38
rs776226101 Chr.5: 157238061-157238061 on Build GRCh38
rs753847568 Chr.5: 157238130-157238130 on Build GRCh38
rs138548010 Chr.5: 157238060-157238060 on Build GRCh38
rs752615063 Chr.5: 157238243-157238243 on Build GRCh38
rs917745624 Chr.5: 157238234-157238234 on Build GRCh38
rs1006929318 Chr.5: 157238194-157238194 on Build GRCh38
rs67533376 Chr.5: 157238256-157238256 on Build GRCh38
rs34184802 Chr.5: 157238258-157238258 on Build GRCh38
rs774073557 Chr.5: 157238228-157238228 on Build GRCh38
rs969360463 Chr.5: 157238327-157238327 on Build GRCh38
rs1030045561 Chr.5: 157238330-157238330 on Build GRCh38
rs768805648 Chr.5: 157238404-157238404 on Build GRCh38
rs34064633 Chr.5: 157238256-157238256 on Build GRCh38
rs3214572 Chr.5: 157238061-157238068 on Build GRCh38
rs528889449 Chr.5: 157238314-157238314 on Build GRCh38
rs925298445 Chr.5: 157238390-157238390 on Build GRCh38
rs997262726 Chr.5: 157238022-157238022 on Build GRCh38
rs956685259 Chr.5: 157238014-157238014 on Build GRCh38
rs775449466 Chr.5: 157238222-157238222 on Build GRCh38
rs780145031 Chr.5: 157238344-157238344 on Build GRCh38
rs56100638 Chr.5: 157238096-157238096 on Build GRCh38
rs770640503 Chr.5: 157238225-157238225 on Build GRCh38
rs916462268 Chr.5: 157238310-157238310 on Build GRCh38
rs979879612 Chr.5: 157238328-157238328 on Build GRCh38
rs780522355 Chr.5: 157238163-157238163 on Build GRCh38
rs1053064787 Chr.5: 157238100-157238100 on Build GRCh38
rs960504990 Chr.5: 157238286-157238286 on Build GRCh38
rs986349805 Chr.5: 157238090-157238090 on Build GRCh38
rs779105268 Chr.5: 157238147-157238147 on Build GRCh38
rs566225025 Chr.5: 157238355-157238355 on Build GRCh38
rs1007655017 Chr.5: 157238385-157238385 on Build GRCh38
rs202191474 Chr.5: 157238184-157238184 on Build GRCh38
rs749513868 Chr.5: 157238397-157238397 on Build GRCh38
rs750585884 Chr.5: 157238150-157238150 on Build GRCh38
rs59272074 Chr.5: 157238271-157238271 on Build GRCh38
rs115402136 Chr.5: 157238230-157238230 on Build GRCh38
rs972060964 Chr.5: 157238109-157238109 on Build GRCh38
rs757692774 Chr.5: 157238168-157238168 on Build GRCh38
rs947834237 Chr.5: 157238253-157238253 on Build GRCh38
rs767365690 Chr.5: 157238241-157238241 on Build GRCh38
rs370800001 Chr.5: 157238056-157238056 on Build GRCh38
rs572233794 Chr.5: 157238108-157238108 on Build GRCh38
rs199870150 Chr.5: 157238085-157238085 on Build GRCh38
rs551183755 Chr.5: 157238353-157238353 on Build GRCh38
rs769166363 Chr.5: 157238196-157238196 on Build GRCh38
rs889292606 Chr.5: 157238382-157238382 on Build GRCh38
rs767455537 Chr.5: 157238122-157238122 on Build GRCh38
rs55646940 Chr.5: 157238099-157238099 on Build GRCh38
rs572655455 Chr.5: 157238050-157238050 on Build GRCh38
rs1043823890 Chr.5: 157238084-157238084 on Build GRCh38
rs769649354 Chr.5: 157238105-157238105 on Build GRCh38
rs540853883 Chr.5: 157238192-157238192 on Build GRCh38
rs759419301 Chr.5: 157238119-157238119 on Build GRCh38
rs992049777 Chr.5: 157238297-157238297 on Build GRCh38
rs372206530 Chr.5: 157238078-157238078 on Build GRCh38
rs757278824 Chr.5: 157238132-157238132 on Build GRCh38

View Full Product Details