Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 7726858-7727388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP6
Entrez Gene ID
654
Gene Name
bone morphogenetic protein 6
Gene Aliases
VGR, VGR1
UniGene
Hs.285671
Gene Chromosome Location
Chr.6: 7726778-7881728 on Build GRCh38
Transcripts
NM_001718.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1238310 c.395C>T p.A132V Chr.6: 7727350-7727350 on Build GRCh38
1312397 c.415C>T p.L139L Chr.6: 7727370-7727370 on Build GRCh38
5345211 c.387G>A p.L129L Chr.6: 7727342-7727342 on Build GRCh38
7272799 c.279C>A p.P93P Chr.6: 7727234-7727234 on Build GRCh38
6373145 c.4C>T p.P2S Chr.6: 7726959-7726959 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759464815 Chr.6: 7727127-7727128 on Build GRCh38
rs746449569 Chr.6: 7727171-7727171 on Build GRCh38
rs758374667 Chr.6: 7727224-7727225 on Build GRCh38
rs767201995 Chr.6: 7727348-7727348 on Build GRCh38
rs775095706 Chr.6: 7727169-7727169 on Build GRCh38
rs771495326 Chr.6: 7727261-7727261 on Build GRCh38
rs146710919 Chr.6: 7726884-7726894 on Build GRCh38
rs769021981 Chr.6: 7727329-7727329 on Build GRCh38
rs767598408 Chr.6: 7727204-7727204 on Build GRCh38
rs187155245 Chr.6: 7727340-7727340 on Build GRCh38
rs201935310 Chr.6: 7727327-7727327 on Build GRCh38
rs771574320 Chr.6: 7727166-7727166 on Build GRCh38
rs758074194 Chr.6: 7727220-7727220 on Build GRCh38
rs755551885 Chr.6: 7727321-7727321 on Build GRCh38
rs759757384 Chr.6: 7727233-7727233 on Build GRCh38
rs7745236 Chr.6: 7727292-7727292 on Build GRCh38
rs111588693 Chr.6: 7727038-7727038 on Build GRCh38
rs752779336 Chr.6: 7727105-7727105 on Build GRCh38
rs199518216 Chr.6: 7727238-7727238 on Build GRCh38
rs1036932521 Chr.6: 7726990-7726990 on Build GRCh38
rs753918736 Chr.6: 7727236-7727236 on Build GRCh38
rs765090274 Chr.6: 7727176-7727178 on Build GRCh38
rs747427445 Chr.6: 7727364-7727364 on Build GRCh38
rs1001122292 Chr.6: 7727070-7727070 on Build GRCh38
rs537332654 Chr.6: 7727289-7727290 on Build GRCh38
rs780705858 Chr.6: 7727254-7727254 on Build GRCh38
rs779625403 Chr.6: 7727228-7727228 on Build GRCh38
rs751383633 Chr.6: 7727343-7727343 on Build GRCh38
rs953331948 Chr.6: 7726922-7726922 on Build GRCh38
rs746635386 Chr.6: 7727244-7727244 on Build GRCh38
rs761992924 Chr.6: 7727335-7727335 on Build GRCh38
rs756246256 Chr.6: 7727205-7727205 on Build GRCh38
rs763377219 Chr.6: 7727181-7727181 on Build GRCh38
rs760992636 Chr.6: 7727281-7727281 on Build GRCh38
rs377443730 Chr.6: 7727296-7727296 on Build GRCh38
rs549761145 Chr.6: 7727134-7727134 on Build GRCh38
rs201788022 Chr.6: 7727180-7727180 on Build GRCh38
rs919392084 Chr.6: 7727003-7727003 on Build GRCh38
rs201486498 Chr.6: 7727289-7727289 on Build GRCh38
rs904968871 Chr.6: 7727031-7727031 on Build GRCh38
rs759273978 Chr.6: 7727347-7727347 on Build GRCh38
rs367791101 Chr.6: 7727154-7727154 on Build GRCh38
rs770753032 Chr.6: 7727147-7727147 on Build GRCh38
rs376104548 Chr.6: 7727350-7727350 on Build GRCh38
rs753793938 Chr.6: 7727209-7727209 on Build GRCh38
rs533887534 Chr.6: 7727278-7727278 on Build GRCh38
rs754534061 Chr.6: 7727246-7727246 on Build GRCh38
rs768933997 Chr.6: 7727282-7727282 on Build GRCh38
rs1001806413 Chr.6: 7727036-7727036 on Build GRCh38
rs777792391 Chr.6: 7727133-7727133 on Build GRCh38
rs778790527 Chr.6: 7727210-7727210 on Build GRCh38
rs986151349 Chr.6: 7726925-7726925 on Build GRCh38
rs537870091 Chr.6: 7727326-7727326 on Build GRCh38
rs752496918 Chr.6: 7727215-7727216 on Build GRCh38
rs1012098266 Chr.6: 7727247-7727247 on Build GRCh38
rs746349898 Chr.6: 7727325-7727325 on Build GRCh38
rs764190979 Chr.6: 7727207-7727207 on Build GRCh38
rs376612665 Chr.6: 7727174-7727174 on Build GRCh38
rs766234867 Chr.6: 7727342-7727342 on Build GRCh38
rs896399711 Chr.6: 7727042-7727042 on Build GRCh38
rs1009526839 Chr.6: 7727051-7727051 on Build GRCh38
rs945009915 Chr.6: 7726975-7726975 on Build GRCh38
rs750246590 Chr.6: 7727239-7727239 on Build GRCh38
rs943346289 Chr.6: 7727253-7727253 on Build GRCh38
rs868027339 Chr.6: 7727108-7727108 on Build GRCh38
rs772563237 Chr.6: 7727270-7727270 on Build GRCh38
rs376922072 Chr.6: 7727235-7727235 on Build GRCh38
rs766146566 Chr.6: 7727302-7727302 on Build GRCh38
rs967873795 Chr.6: 7727060-7727060 on Build GRCh38
rs377639547 Chr.6: 7727265-7727265 on Build GRCh38
rs755710426 Chr.6: 7727351-7727351 on Build GRCh38
rs747541472 Chr.6: 7727255-7727255 on Build GRCh38
rs746287546 Chr.6: 7727268-7727268 on Build GRCh38
rs751803774 Chr.6: 7727081-7727081 on Build GRCh38
rs752305807 Chr.6: 7727311-7727311 on Build GRCh38
rs773751065 Chr.6: 7727230-7727230 on Build GRCh38
rs780427949 Chr.6: 7727290-7727298 on Build GRCh38
rs774658607 Chr.6: 7727303-7727311 on Build GRCh38
rs777164496 Chr.6: 7727353-7727353 on Build GRCh38
rs758116388 Chr.6: 7727241-7727241 on Build GRCh38
rs774813618 Chr.6: 7727232-7727232 on Build GRCh38
rs773257606 Chr.6: 7727339-7727339 on Build GRCh38
rs749567930 Chr.6: 7727290-7727295 on Build GRCh38
rs754626098 Chr.6: 7727308-7727308 on Build GRCh38
rs755363611 Chr.6: 7727367-7727367 on Build GRCh38
rs759005904 Chr.6: 7727355-7727355 on Build GRCh38
rs778468984 Chr.6: 7727160-7727160 on Build GRCh38
rs774488996 Chr.6: 7727189-7727189 on Build GRCh38
rs753269481 Chr.6: 7727309-7727309 on Build GRCh38
rs757277220 Chr.6: 7727278-7727279 on Build GRCh38
rs751662817 Chr.6: 7727236-7727237 on Build GRCh38
rs781313847 Chr.6: 7727094-7727094 on Build GRCh38
rs1021295129 Chr.6: 7727054-7727055 on Build GRCh38
rs755054887 Chr.6: 7727091-7727091 on Build GRCh38
rs5025862 Chr.6: 7726889-7726889 on Build GRCh38
rs542708147 Chr.6: 7726935-7726935 on Build GRCh38
rs745510672 Chr.6: 7727164-7727164 on Build GRCh38
rs753324025 Chr.6: 7727354-7727354 on Build GRCh38
rs781449388 Chr.6: 7727283-7727284 on Build GRCh38
rs200554660 Chr.6: 7727294-7727294 on Build GRCh38
rs1028191991 Chr.6: 7727314-7727314 on Build GRCh38
rs775914813 Chr.6: 7727279-7727279 on Build GRCh38
rs986460047 Chr.6: 7727312-7727312 on Build GRCh38
rs759829490 Chr.6: 7727193-7727193 on Build GRCh38
rs371499962 Chr.6: 7727186-7727186 on Build GRCh38
rs771616962 Chr.6: 7727292-7727293 on Build GRCh38
rs200573175 Chr.6: 7727242-7727242 on Build GRCh38
rs745610164 Chr.6: 7727214-7727214 on Build GRCh38
rs370312610 Chr.6: 7727307-7727307 on Build GRCh38
rs1033072339 Chr.6: 7727072-7727072 on Build GRCh38
rs561342836 Chr.6: 7726961-7726961 on Build GRCh38
rs763878792 Chr.6: 7727225-7727230 on Build GRCh38
rs746580433 Chr.6: 7727229-7727229 on Build GRCh38
rs762871776 Chr.6: 7727299-7727299 on Build GRCh38
rs1028244492 Chr.6: 7727295-7727295 on Build GRCh38
rs779544660 Chr.6: 7727324-7727324 on Build GRCh38
rs745504783 Chr.6: 7727290-7727292 on Build GRCh38
rs930813516 Chr.6: 7727018-7727018 on Build GRCh38
rs1048774251 Chr.6: 7727022-7727022 on Build GRCh38
rs756121523 Chr.6: 7727124-7727124 on Build GRCh38
rs769758049 Chr.6: 7727336-7727336 on Build GRCh38

View Full Product Details