Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 70225658-70226156 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL9A1
Entrez Gene ID
1297
Gene Name
collagen type IX alpha 1
Gene Aliases
DJ149L1.1.2, EDM6, MED, STL4
UniGene
Hs.590892
Gene Chromosome Location
Chr.6: 70216040-70303083 on Build GRCh38
Transcripts
NM_001851.4 NM_078485.3 XM_011535429.2 XM_011535430.2 XM_017010246.1 XM_017010247.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5746604 c.1820C>A p.P607H Chr.6: 70225964-70225964 on Build GRCh38
4932927 c.2561C>A p.P854H Chr.6: 70225952-70225952 on Build GRCh38
4932928 c.1832C>A p.P611H Chr.6: 70225952-70225952 on Build GRCh38
6012365 c.1826G>A p.G609D Chr.6: 70225958-70225958 on Build GRCh38
5764656 c.1817G>T p.G606V Chr.6: 70225967-70225967 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779808424 Chr.6: 70226041-70226041 on Build GRCh38
rs992484011 Chr.6: 70226064-70226064 on Build GRCh38
rs374185119 Chr.6: 70226044-70226044 on Build GRCh38
rs201978369 Chr.6: 70225976-70225976 on Build GRCh38
rs578034223 Chr.6: 70225961-70225961 on Build GRCh38
rs528372012 Chr.6: 70226047-70226047 on Build GRCh38
rs748046538 Chr.6: 70226049-70226049 on Build GRCh38
rs1135057 Chr.6: 70225951-70225951 on Build GRCh38
rs755509700 Chr.6: 70225882-70225882 on Build GRCh38
rs79282554 Chr.6: 70226056-70226056 on Build GRCh38
rs546606054 Chr.6: 70226108-70226108 on Build GRCh38
rs754167994 Chr.6: 70226025-70226025 on Build GRCh38
rs200103796 Chr.6: 70225900-70225900 on Build GRCh38
rs899351727 Chr.6: 70225752-70225752 on Build GRCh38
rs746481353 Chr.6: 70225943-70225943 on Build GRCh38
rs955977873 Chr.6: 70226062-70226062 on Build GRCh38
rs1054864480 Chr.6: 70225903-70225903 on Build GRCh38
rs547618159 Chr.6: 70225985-70225985 on Build GRCh38
rs745496117 Chr.6: 70225908-70225908 on Build GRCh38
rs756400521 Chr.6: 70225899-70225899 on Build GRCh38
rs895021995 Chr.6: 70225937-70225937 on Build GRCh38
rs761742449 Chr.6: 70225952-70225952 on Build GRCh38
rs985584771 Chr.6: 70226069-70226069 on Build GRCh38
rs1044556397 Chr.6: 70225746-70225746 on Build GRCh38
rs534752381 Chr.6: 70225960-70225960 on Build GRCh38
rs916929188 Chr.6: 70226065-70226065 on Build GRCh38
rs753195491 Chr.6: 70226012-70226012 on Build GRCh38
rs368114172 Chr.6: 70225954-70225954 on Build GRCh38
rs532443730 Chr.6: 70225888-70225888 on Build GRCh38
rs143217938 Chr.6: 70225986-70225986 on Build GRCh38
rs569795420 Chr.6: 70225816-70225816 on Build GRCh38
rs778015289 Chr.6: 70226016-70226016 on Build GRCh38
rs886702234 Chr.6: 70225955-70225955 on Build GRCh38
rs370699377 Chr.6: 70226042-70226042 on Build GRCh38
rs1042464097 Chr.6: 70225896-70225896 on Build GRCh38
rs977328651 Chr.6: 70226112-70226112 on Build GRCh38
rs768642091 Chr.6: 70225926-70225926 on Build GRCh38
rs778692300 Chr.6: 70225906-70225906 on Build GRCh38
rs944271652 Chr.6: 70225783-70225783 on Build GRCh38
rs562791036 Chr.6: 70225831-70225831 on Build GRCh38
rs568295931 Chr.6: 70226014-70226014 on Build GRCh38
rs748558592 Chr.6: 70225897-70225897 on Build GRCh38
rs577125555 Chr.6: 70226080-70226080 on Build GRCh38
rs945024661 Chr.6: 70225868-70225868 on Build GRCh38
rs977038028 Chr.6: 70225790-70225790 on Build GRCh38
rs751660212 Chr.6: 70225978-70225978 on Build GRCh38
rs768737537 Chr.6: 70225944-70225944 on Build GRCh38
rs537226988 Chr.6: 70226028-70226028 on Build GRCh38
rs965504284 Chr.6: 70226104-70226104 on Build GRCh38
rs911416987 Chr.6: 70225757-70225758 on Build GRCh38
rs774871148 Chr.6: 70225932-70225932 on Build GRCh38

View Full Product Details