Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 73761926-73762424 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD109
Entrez Gene ID
135228
Gene Name
CD109 molecule
Gene Aliases
CPAMD7, p180, r150
UniGene
Hs.399891
Gene Chromosome Location
Chr.6: 73679192-73828317 on Build GRCh38
Transcripts
NM_001159587.2 NM_001159588.2 NM_133493.4 XM_005248659.3 XM_011535473.1 XM_017010283.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6623386 c.790G>A p.V264I Chr.6: 73762415-73762415 on Build GRCh38
5698911 c.777A>T p.P259P Chr.6: 73762402-73762402 on Build GRCh38
1445865 c.794C>T p.T265M Chr.6: 73762419-73762419 on Build GRCh38
6471381 c.793A>T p.T265S Chr.6: 73762418-73762418 on Build GRCh38
3875446 c.795G>A p.T265T Chr.6: 73762420-73762420 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1019985984 Chr.6: 73761982-73761982 on Build GRCh38
rs549856484 Chr.6: 73762025-73762025 on Build GRCh38
rs963360194 Chr.6: 73762088-73762088 on Build GRCh38
rs1039383814 Chr.6: 73761954-73761954 on Build GRCh38
rs9293939 Chr.6: 73762128-73762128 on Build GRCh38
rs560900997 Chr.6: 73762146-73762146 on Build GRCh38
rs190304780 Chr.6: 73762180-73762180 on Build GRCh38
rs907114006 Chr.6: 73761956-73761956 on Build GRCh38
rs531505898 Chr.6: 73761960-73761960 on Build GRCh38
rs762608750 Chr.6: 73762337-73762337 on Build GRCh38
rs375716383 Chr.6: 73762092-73762092 on Build GRCh38
rs563439014 Chr.6: 73762386-73762386 on Build GRCh38
rs1003047036 Chr.6: 73762001-73762001 on Build GRCh38
rs181642822 Chr.6: 73761958-73761958 on Build GRCh38
rs12661860 Chr.6: 73762257-73762257 on Build GRCh38
rs992129315 Chr.6: 73762065-73762065 on Build GRCh38
rs367957225 Chr.6: 73762347-73762347 on Build GRCh38
rs986657621 Chr.6: 73762184-73762184 on Build GRCh38
rs1041328675 Chr.6: 73762216-73762216 on Build GRCh38
rs963412216 Chr.6: 73762275-73762275 on Build GRCh38
rs942576744 Chr.6: 73761950-73761950 on Build GRCh38
rs938567144 Chr.6: 73762295-73762295 on Build GRCh38
rs113691268 Chr.6: 73761949-73761949 on Build GRCh38
rs1026675036 Chr.6: 73762204-73762204 on Build GRCh38
rs563599085 Chr.6: 73762253-73762253 on Build GRCh38
rs200534962 Chr.6: 73761950-73761950 on Build GRCh38
rs912354646 Chr.6: 73762197-73762197 on Build GRCh38
rs761242650 Chr.6: 73762374-73762374 on Build GRCh38
rs184241237 Chr.6: 73762389-73762389 on Build GRCh38
rs568337343 Chr.6: 73762032-73762032 on Build GRCh38
rs370707246 Chr.6: 73762394-73762394 on Build GRCh38
rs565080852 Chr.6: 73761950-73761951 on Build GRCh38
rs903403710 Chr.6: 73761989-73761989 on Build GRCh38
rs942650206 Chr.6: 73762207-73762207 on Build GRCh38
rs557793736 Chr.6: 73762196-73762196 on Build GRCh38
rs959000370 Chr.6: 73762056-73762056 on Build GRCh38
rs977789918 Chr.6: 73762119-73762119 on Build GRCh38
rs574257892 Chr.6: 73762229-73762229 on Build GRCh38
rs374823973 Chr.6: 73762163-73762163 on Build GRCh38
rs767865919 Chr.6: 73762363-73762363 on Build GRCh38
rs189809778 Chr.6: 73762369-73762369 on Build GRCh38
rs185352568 Chr.6: 73762288-73762288 on Build GRCh38
rs894342692 Chr.6: 73761981-73761988 on Build GRCh38
rs537047461 Chr.6: 73762212-73762212 on Build GRCh38
rs543710368 Chr.6: 73762208-73762208 on Build GRCh38
rs565799755 Chr.6: 73762174-73762174 on Build GRCh38
rs182569842 Chr.6: 73762251-73762251 on Build GRCh38
rs759284700 Chr.6: 73762361-73762361 on Build GRCh38
rs1036234652 Chr.6: 73762050-73762050 on Build GRCh38
rs541291871 Chr.6: 73762250-73762250 on Build GRCh38
rs8180634 Chr.6: 73762189-73762189 on Build GRCh38
rs764569899 Chr.6: 73762384-73762384 on Build GRCh38
rs753896488 Chr.6: 73762388-73762388 on Build GRCh38
rs933350624 Chr.6: 73762157-73762157 on Build GRCh38
rs771964122 Chr.6: 73762185-73762185 on Build GRCh38
rs550455085 Chr.6: 73762213-73762213 on Build GRCh38
rs766106233 Chr.6: 73762345-73762345 on Build GRCh38
rs773167985 Chr.6: 73762240-73762240 on Build GRCh38
rs185828464 Chr.6: 73762075-73762075 on Build GRCh38
rs1011422313 Chr.6: 73762036-73762036 on Build GRCh38
rs555827315 Chr.6: 73762221-73762221 on Build GRCh38
rs1034169483 Chr.6: 73761980-73761980 on Build GRCh38

View Full Product Details