Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 117525252-117525771 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCBLD1
Entrez Gene ID
285761
Gene Name
discoidin, CUB and LCCL domain containing 1
Gene Aliases
dJ94G16.1
UniGene
Hs.658304
Gene Chromosome Location
Chr.6: 117480346-117569858 on Build GRCh38
Transcripts
NM_173674.2 XM_005266941.1 XM_006715463.1 XM_011535771.1 XM_011535773.2 XM_011535774.2 XM_017010775.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4949454 c.572A>G p.D191G Chr.6: 117525421-117525421 on Build GRCh38
1620815 c.581G>C p.R194T Chr.6: 117525430-117525430 on Build GRCh38
3662099 c.581G>C p.R194T Chr.6: 117525430-117525430 on Build GRCh38
4349757 c.513-4C>T p.? Chr.6: 117525358-117525358 on Build GRCh38
4349756 c.513-4C>T p.? Chr.6: 117525358-117525358 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1007014439 Chr.6: 117525503-117525503 on Build GRCh38
rs186209455 Chr.6: 117525547-117525547 on Build GRCh38
rs767838532 Chr.6: 117525406-117525406 on Build GRCh38
rs142155026 Chr.6: 117525429-117525429 on Build GRCh38
rs772196920 Chr.6: 117525520-117525520 on Build GRCh38
rs1000493269 Chr.6: 117525453-117525453 on Build GRCh38
rs951371202 Chr.6: 117525592-117525592 on Build GRCh38
rs187245896 Chr.6: 117525687-117525687 on Build GRCh38
rs749811592 Chr.6: 117525375-117525375 on Build GRCh38
rs750305020 Chr.6: 117525402-117525402 on Build GRCh38
rs199958002 Chr.6: 117525308-117525308 on Build GRCh38
rs917343817 Chr.6: 117525658-117525658 on Build GRCh38
rs1032415876 Chr.6: 117525455-117525455 on Build GRCh38
rs757249736 Chr.6: 117525442-117525442 on Build GRCh38
rs370148191 Chr.6: 117525417-117525417 on Build GRCh38
rs201112531 Chr.6: 117525279-117525279 on Build GRCh38
rs111772606 Chr.6: 117525633-117525633 on Build GRCh38
rs146408484 Chr.6: 117525617-117525618 on Build GRCh38
rs200777402 Chr.6: 117525452-117525452 on Build GRCh38
rs183124653 Chr.6: 117525684-117525684 on Build GRCh38
rs371948238 Chr.6: 117525369-117525369 on Build GRCh38
rs371632697 Chr.6: 117525641-117525641 on Build GRCh38
rs770062686 Chr.6: 117525318-117525318 on Build GRCh38
rs571711963 Chr.6: 117525616-117525616 on Build GRCh38
rs775191351 Chr.6: 117525413-117525413 on Build GRCh38
rs903385077 Chr.6: 117525702-117525702 on Build GRCh38
rs200146951 Chr.6: 117525431-117525431 on Build GRCh38
rs375411561 Chr.6: 117525389-117525389 on Build GRCh38
rs976571207 Chr.6: 117525521-117525521 on Build GRCh38
rs956149123 Chr.6: 117525647-117525647 on Build GRCh38
rs955428471 Chr.6: 117525283-117525283 on Build GRCh38
rs1003992806 Chr.6: 117525511-117525511 on Build GRCh38
rs766660535 Chr.6: 117525347-117525347 on Build GRCh38
rs191105900 Chr.6: 117525604-117525604 on Build GRCh38
rs924692694 Chr.6: 117525301-117525301 on Build GRCh38
rs986853877 Chr.6: 117525285-117525285 on Build GRCh38
rs538764350 Chr.6: 117525629-117525629 on Build GRCh38
rs140920753 Chr.6: 117525616-117525617 on Build GRCh38
rs761128608 Chr.6: 117525340-117525343 on Build GRCh38
rs991510372 Chr.6: 117525619-117525619 on Build GRCh38
rs1018057809 Chr.6: 117525506-117525506 on Build GRCh38
rs550136922 Chr.6: 117525614-117525614 on Build GRCh38
rs34254506 Chr.6: 117525618-117525619 on Build GRCh38
rs527934700 Chr.6: 117525282-117525282 on Build GRCh38
rs542506106 Chr.6: 117525287-117525287 on Build GRCh38
rs536515364 Chr.6: 117525693-117525693 on Build GRCh38
rs201692838 Chr.6: 117525306-117525306 on Build GRCh38
rs965180107 Chr.6: 117525515-117525515 on Build GRCh38
rs773111945 Chr.6: 117525337-117525337 on Build GRCh38
rs147911816 Chr.6: 117525597-117525597 on Build GRCh38
rs950165433 Chr.6: 117525677-117525677 on Build GRCh38
rs760306238 Chr.6: 117525416-117525416 on Build GRCh38
rs934783692 Chr.6: 117525309-117525309 on Build GRCh38
rs112073259 Chr.6: 117525655-117525655 on Build GRCh38
rs893480961 Chr.6: 117525492-117525492 on Build GRCh38
rs774801446 Chr.6: 117525390-117525390 on Build GRCh38
rs758306146 Chr.6: 117525476-117525476 on Build GRCh38

View Full Product Details