Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155087106-155087605 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFNA3
Entrez Gene ID
1944
Gene Name
ephrin A3
Gene Aliases
EFL2, EPLG3, Ehk1-L, LERK3
UniGene
Hs.516656
Gene Chromosome Location
Chr.1: 155078872-155087538 on Build GRCh38
Transcripts
NM_004952.4

SNP Detail

SNP ID Chromosome Location
rs780160882 Chr.1: 155087205-155087205 on Build GRCh38
rs901617645 Chr.1: 155087258-155087258 on Build GRCh38
rs1040093082 Chr.1: 155087225-155087225 on Build GRCh38
rs546342547 Chr.1: 155087364-155087364 on Build GRCh38
rs930848158 Chr.1: 155087401-155087401 on Build GRCh38
rs1015821084 Chr.1: 155087222-155087222 on Build GRCh38
rs112133541 Chr.1: 155087139-155087139 on Build GRCh38
rs977058851 Chr.1: 155087253-155087253 on Build GRCh38
rs568868065 Chr.1: 155087402-155087402 on Build GRCh38
rs1057089817 Chr.1: 155087380-155087380 on Build GRCh38
rs943034016 Chr.1: 155087426-155087426 on Build GRCh38
rs1022867844 Chr.1: 155087543-155087544 on Build GRCh38
rs572720799 Chr.1: 155087197-155087197 on Build GRCh38
rs910825579 Chr.1: 155087404-155087404 on Build GRCh38
rs1004353681 Chr.1: 155087455-155087455 on Build GRCh38
rs930898856 Chr.1: 155087167-155087168 on Build GRCh38
rs1011857646 Chr.1: 155087524-155087525 on Build GRCh38
rs957235761 Chr.1: 155087291-155087291 on Build GRCh38
rs143153278 Chr.1: 155087263-155087263 on Build GRCh38
rs888745785 Chr.1: 155087171-155087171 on Build GRCh38
rs1011489116 Chr.1: 155087301-155087301 on Build GRCh38
rs921244843 Chr.1: 155087279-155087279 on Build GRCh38
rs186168085 Chr.1: 155087377-155087377 on Build GRCh38
rs966026498 Chr.1: 155087224-155087224 on Build GRCh38
rs548488696 Chr.1: 155087518-155087518 on Build GRCh38
rs563747435 Chr.1: 155087353-155087354 on Build GRCh38
rs1049815388 Chr.1: 155087167-155087174 on Build GRCh38
rs1016162565 Chr.1: 155087562-155087562 on Build GRCh38
rs989514762 Chr.1: 155087292-155087292 on Build GRCh38
rs2306124 Chr.1: 155087375-155087375 on Build GRCh38
rs947198783 Chr.1: 155087342-155087342 on Build GRCh38
rs1004384713 Chr.1: 155087206-155087206 on Build GRCh38
rs972153770 Chr.1: 155087378-155087383 on Build GRCh38
rs998584852 Chr.1: 155087260-155087260 on Build GRCh38
rs1052805486 Chr.1: 155087473-155087473 on Build GRCh38
rs2306123 Chr.1: 155087330-155087330 on Build GRCh38
rs1023307325 Chr.1: 155087310-155087311 on Build GRCh38
rs11554834 Chr.1: 155087393-155087393 on Build GRCh38
rs1039994493 Chr.1: 155087427-155087427 on Build GRCh38
rs775006799 Chr.1: 155087202-155087202 on Build GRCh38
rs1026772143 Chr.1: 155087152-155087152 on Build GRCh38
rs527708452 Chr.1: 155087433-155087433 on Build GRCh38
rs139174640 Chr.1: 155087438-155087438 on Build GRCh38
rs954082946 Chr.1: 155087284-155087284 on Build GRCh38

View Full Product Details