Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 127330955-127331486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECHDC1
Entrez Gene ID
55862
Gene Name
ethylmalonyl-CoA decarboxylase 1
Gene Aliases
HEL-S-76, MMCD, dJ351K20.2
UniGene
Hs.486410
Gene Chromosome Location
Chr.6: 127288712-127343609 on Build GRCh38
Transcripts
NM_001002030.1 NM_001105544.1 NM_001105545.1 NM_001139510.1 NM_018479.3 XM_005267047.2 XM_005267048.1 XM_005267049.2 XM_005267050.2 XM_011535942.2 XM_011535943.2 XM_017011056.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6651336 c.43A>T p.T15S Chr.6: 127330986-127330986 on Build GRCh38
7146271 c.76C>G p.Q26E Chr.6: 127330971-127330971 on Build GRCh38
7146272 c.58C>G p.Q20E Chr.6: 127330971-127330971 on Build GRCh38
3429967 c.17-1G>A p.? Chr.6: 127331031-127331031 on Build GRCh38
1072990 c.6G>A p.A2A Chr.6: 127331023-127331023 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774841748 Chr.6: 127331053-127331053 on Build GRCh38
rs887769969 Chr.6: 127331141-127331141 on Build GRCh38
rs949865704 Chr.6: 127331092-127331092 on Build GRCh38
rs763787952 Chr.6: 127331000-127331000 on Build GRCh38
rs536758438 Chr.6: 127331155-127331155 on Build GRCh38
rs35842645 Chr.6: 127331152-127331152 on Build GRCh38
rs373335171 Chr.6: 127331054-127331054 on Build GRCh38
rs9375501 Chr.6: 127331394-127331394 on Build GRCh38
rs929244683 Chr.6: 127331118-127331118 on Build GRCh38
rs773445365 Chr.6: 127331039-127331039 on Build GRCh38
rs919087522 Chr.6: 127331447-127331447 on Build GRCh38
rs569775563 Chr.6: 127331146-127331146 on Build GRCh38
rs553737787 Chr.6: 127331292-127331292 on Build GRCh38
rs201511843 Chr.6: 127331023-127331023 on Build GRCh38
rs569839202 Chr.6: 127331303-127331303 on Build GRCh38
rs771283936 Chr.6: 127331047-127331047 on Build GRCh38
rs11556355 Chr.6: 127331001-127331001 on Build GRCh38
rs375748837 Chr.6: 127331077-127331077 on Build GRCh38
rs1021125301 Chr.6: 127331291-127331291 on Build GRCh38
rs1039112710 Chr.6: 127331208-127331208 on Build GRCh38
rs766032713 Chr.6: 127331327-127331327 on Build GRCh38
rs879164450 Chr.6: 127331153-127331153 on Build GRCh38
rs985403321 Chr.6: 127331258-127331258 on Build GRCh38
rs376685759 Chr.6: 127331034-127331034 on Build GRCh38
rs1049482457 Chr.6: 127331124-127331124 on Build GRCh38
rs1045555790 Chr.6: 127331098-127331098 on Build GRCh38
rs760772954 Chr.6: 127331069-127331069 on Build GRCh38
rs112681066 Chr.6: 127331225-127331225 on Build GRCh38
rs542713290 Chr.6: 127331346-127331346 on Build GRCh38
rs551187863 Chr.6: 127331010-127331010 on Build GRCh38
rs561514551 Chr.6: 127331347-127331347 on Build GRCh38
rs555951218 Chr.6: 127331173-127331173 on Build GRCh38
rs577936011 Chr.6: 127331293-127331293 on Build GRCh38
rs757132115 Chr.6: 127331005-127331005 on Build GRCh38
rs1033038590 Chr.6: 127331312-127331312 on Build GRCh38
rs535302868 Chr.6: 127331282-127331282 on Build GRCh38
rs955354929 Chr.6: 127331209-127331209 on Build GRCh38
rs768479816 Chr.6: 127331024-127331024 on Build GRCh38
rs991135776 Chr.6: 127331366-127331366 on Build GRCh38
rs749719664 Chr.6: 127331044-127331044 on Build GRCh38
rs1007587542 Chr.6: 127331181-127331181 on Build GRCh38
rs988925712 Chr.6: 127331148-127331148 on Build GRCh38
rs775592361 Chr.6: 127331055-127331055 on Build GRCh38
rs575652476 Chr.6: 127331304-127331304 on Build GRCh38
rs1003340670 Chr.6: 127331151-127331151 on Build GRCh38
rs142314959 Chr.6: 127331002-127331002 on Build GRCh38
rs957017609 Chr.6: 127331365-127331365 on Build GRCh38
rs370720855 Chr.6: 127331200-127331200 on Build GRCh38
rs369016435 Chr.6: 127331005-127331006 on Build GRCh38
rs752923726 Chr.6: 127330981-127330981 on Build GRCh38
rs76583416 Chr.6: 127331007-127331007 on Build GRCh38
rs918544243 Chr.6: 127331353-127331353 on Build GRCh38
rs879076082 Chr.6: 127331152-127331153 on Build GRCh38
rs915634388 Chr.6: 127331376-127331376 on Build GRCh38
rs758015854 Chr.6: 127331012-127331012 on Build GRCh38
rs752600705 Chr.6: 127331078-127331081 on Build GRCh38
rs919137108 Chr.6: 127331101-127331101 on Build GRCh38
rs755883205 Chr.6: 127330995-127330995 on Build GRCh38
rs767688651 Chr.6: 127330980-127330980 on Build GRCh38
rs765209150 Chr.6: 127331077-127331078 on Build GRCh38
rs142869825 Chr.6: 127331249-127331249 on Build GRCh38
rs901940449 Chr.6: 127331277-127331277 on Build GRCh38
rs759041437 Chr.6: 127331012-127331013 on Build GRCh38
rs981645426 Chr.6: 127331425-127331425 on Build GRCh38
rs12198208 Chr.6: 127331302-127331302 on Build GRCh38

View Full Product Details