Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 158637683-158638188 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNLT1
Entrez Gene ID
6993
Gene Name
dynein light chain Tctex-type 1
Gene Aliases
CW-1, TCTEL1, tctex-1
UniGene
Hs.445999
Gene Chromosome Location
Chr.6: 158636474-158644786 on Build GRCh38
Transcripts
NM_001291602.1 NM_001291603.1 NM_006519.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5420959 c.168G>T p.K56N Chr.6: 158637796-158637796 on Build GRCh38
6650672 c.97G>A p.A33T Chr.6: 158637867-158637867 on Build GRCh38
1621152 c.82G>A p.A28T Chr.6: 158637882-158637882 on Build GRCh38
4779414 c.74T>C p.I25T Chr.6: 158637890-158637890 on Build GRCh38
6272272 c.111C>A p.S37R Chr.6: 158637853-158637853 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1034222620 Chr.6: 158638075-158638075 on Build GRCh38
rs1048161457 Chr.6: 158637995-158638000 on Build GRCh38
rs777550322 Chr.6: 158638112-158638112 on Build GRCh38
rs976535957 Chr.6: 158638082-158638082 on Build GRCh38
rs1003007971 Chr.6: 158637957-158637957 on Build GRCh38
rs142843740 Chr.6: 158637882-158637882 on Build GRCh38
rs759596343 Chr.6: 158637866-158637866 on Build GRCh38
rs978825226 Chr.6: 158638142-158638142 on Build GRCh38
rs763464849 Chr.6: 158637929-158637929 on Build GRCh38
rs118095868 Chr.6: 158637850-158637850 on Build GRCh38
rs1014214217 Chr.6: 158638120-158638120 on Build GRCh38
rs201629862 Chr.6: 158637851-158637851 on Build GRCh38
rs932164038 Chr.6: 158637954-158637954 on Build GRCh38
rs1002358928 Chr.6: 158637711-158637711 on Build GRCh38
rs764576544 Chr.6: 158637750-158637750 on Build GRCh38
rs934478302 Chr.6: 158638091-158638091 on Build GRCh38
rs370399499 Chr.6: 158637721-158637721 on Build GRCh38
rs201782892 Chr.6: 158637741-158637741 on Build GRCh38
rs61738908 Chr.6: 158637859-158637859 on Build GRCh38
rs550087753 Chr.6: 158637852-158637852 on Build GRCh38
rs944972057 Chr.6: 158638160-158638160 on Build GRCh38
rs199990530 Chr.6: 158637718-158637718 on Build GRCh38
rs754505360 Chr.6: 158637829-158637829 on Build GRCh38
rs368367909 Chr.6: 158637902-158637902 on Build GRCh38
rs776071009 Chr.6: 158637743-158637743 on Build GRCh38
rs778533397 Chr.6: 158637889-158637889 on Build GRCh38
rs374363477 Chr.6: 158637793-158637793 on Build GRCh38
rs752499794 Chr.6: 158638139-158638141 on Build GRCh38
rs756628313 Chr.6: 158638088-158638089 on Build GRCh38
rs768420263 Chr.6: 158637737-158637737 on Build GRCh38
rs764466100 Chr.6: 158637870-158637870 on Build GRCh38
rs2306750 Chr.6: 158637735-158637735 on Build GRCh38
rs780340495 Chr.6: 158638014-158638014 on Build GRCh38
rs375535915 Chr.6: 158637879-158637879 on Build GRCh38
rs565774199 Chr.6: 158637967-158637967 on Build GRCh38
rs200276406 Chr.6: 158637745-158637745 on Build GRCh38
rs147467336 Chr.6: 158637832-158637832 on Build GRCh38
rs912109726 Chr.6: 158638154-158638154 on Build GRCh38
rs943471261 Chr.6: 158637819-158637819 on Build GRCh38
rs753472448 Chr.6: 158637945-158637945 on Build GRCh38
rs766508179 Chr.6: 158637757-158637757 on Build GRCh38
rs199887878 Chr.6: 158637918-158637918 on Build GRCh38
rs762060976 Chr.6: 158637868-158637868 on Build GRCh38
rs1044333407 Chr.6: 158638127-158638127 on Build GRCh38
rs952045872 Chr.6: 158637786-158637786 on Build GRCh38
rs762118691 Chr.6: 158637862-158637863 on Build GRCh38
rs557755774 Chr.6: 158637995-158637995 on Build GRCh38
rs774612118 Chr.6: 158637822-158637824 on Build GRCh38
rs372034593 Chr.6: 158637937-158637937 on Build GRCh38
rs554427309 Chr.6: 158638153-158638153 on Build GRCh38
rs746850467 Chr.6: 158637915-158637915 on Build GRCh38
rs765518781 Chr.6: 158637923-158637923 on Build GRCh38
rs538181440 Chr.6: 158638116-158638116 on Build GRCh38
rs964752537 Chr.6: 158638061-158638061 on Build GRCh38
rs750569509 Chr.6: 158637966-158637966 on Build GRCh38
rs953521301 Chr.6: 158638153-158638154 on Build GRCh38
rs186830023 Chr.6: 158637820-158637820 on Build GRCh38
rs767900629 Chr.6: 158637943-158637943 on Build GRCh38
rs200809602 Chr.6: 158637916-158637916 on Build GRCh38
rs893096713 Chr.6: 158637898-158637898 on Build GRCh38
rs199737567 Chr.6: 158637771-158637771 on Build GRCh38
rs914477870 Chr.6: 158638106-158638106 on Build GRCh38
rs776769712 Chr.6: 158637917-158637917 on Build GRCh38
rs753628954 Chr.6: 158637904-158637905 on Build GRCh38
rs751897953 Chr.6: 158638086-158638087 on Build GRCh38
rs748124506 Chr.6: 158637896-158637896 on Build GRCh38
rs755796004 Chr.6: 158637872-158637872 on Build GRCh38
rs770143300 Chr.6: 158637727-158637728 on Build GRCh38
rs776912098 Chr.6: 158637860-158637860 on Build GRCh38
rs767904503 Chr.6: 158637889-158637890 on Build GRCh38
rs769335215 Chr.6: 158637731-158637731 on Build GRCh38
rs926918416 Chr.6: 158638135-158638135 on Build GRCh38
rs1033136030 Chr.6: 158638153-158638153 on Build GRCh38
rs779630473 Chr.6: 158637817-158637817 on Build GRCh38
rs777719256 Chr.6: 158637913-158637913 on Build GRCh38
rs779633352 Chr.6: 158638144-158638146 on Build GRCh38
rs912184952 Chr.6: 158637964-158637964 on Build GRCh38
rs370636377 Chr.6: 158638092-158638095 on Build GRCh38
rs753807878 Chr.6: 158637766-158637766 on Build GRCh38
rs774835348 Chr.6: 158637938-158637938 on Build GRCh38
rs1020720749 Chr.6: 158638134-158638134 on Build GRCh38
rs769797202 Chr.6: 158637922-158637922 on Build GRCh38
rs755674058 Chr.6: 158637815-158637815 on Build GRCh38
rs191667256 Chr.6: 158638029-158638029 on Build GRCh38
rs539114471 Chr.6: 158637971-158637971 on Build GRCh38
rs923353350 Chr.6: 158638087-158638087 on Build GRCh38
rs762903281 Chr.6: 158637743-158637753 on Build GRCh38
rs750108639 Chr.6: 158637808-158637808 on Build GRCh38
rs984788700 Chr.6: 158638030-158638030 on Build GRCh38
rs929729531 Chr.6: 158637975-158637975 on Build GRCh38
rs751864431 Chr.6: 158637871-158637871 on Build GRCh38
rs766012904 Chr.6: 158637873-158637873 on Build GRCh38
rs527731074 Chr.6: 158638089-158638092 on Build GRCh38
rs111276200 Chr.6: 158637772-158637772 on Build GRCh38
rs778082262 Chr.6: 158637797-158637797 on Build GRCh38
rs752997251 Chr.6: 158637706-158637706 on Build GRCh38
rs763411609 Chr.6: 158637869-158637869 on Build GRCh38
rs988676958 Chr.6: 158638099-158638099 on Build GRCh38
rs761765295 Chr.6: 158637740-158637740 on Build GRCh38
rs368405907 Chr.6: 158637862-158637862 on Build GRCh38
rs372832664 Chr.6: 158637867-158637867 on Build GRCh38
rs376061466 Chr.6: 158637732-158637732 on Build GRCh38
rs376970070 Chr.6: 158637739-158637739 on Build GRCh38
rs768757152 Chr.6: 158637747-158637751 on Build GRCh38
rs756160552 Chr.6: 158638043-158638043 on Build GRCh38
rs758760975 Chr.6: 158637714-158637714 on Build GRCh38

View Full Product Details