Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 50467127-50467622 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDC
Entrez Gene ID
1644
Gene Name
dopa decarboxylase
Gene Aliases
AADC
UniGene
Hs.359698
Gene Chromosome Location
Chr.7: 50458436-50565460 on Build GRCh38
Transcripts
NM_000790.3 NM_001082971.1 NM_001242886.1 NM_001242887.1 NM_001242888.1 NM_001242889.1 XM_005271745.4 XM_011515161.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
379421 c.1222C>T p.L408F Chr.7: 50467234-50467234 on Build GRCh38
3412144 c.1177C>T p.R393C Chr.7: 50467279-50467279 on Build GRCh38
1090603 c.1238T>C p.L413P Chr.7: 50467218-50467218 on Build GRCh38
139704 c.1159G>A p.E387K Chr.7: 50467297-50467297 on Build GRCh38
7300751 c.1205_1206delTG p.V402fs*19 Chr.7: 50467250-50467251 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190898385 Chr.7: 50467353-50467353 on Build GRCh38
rs938327132 Chr.7: 50467162-50467162 on Build GRCh38
rs763479856 Chr.7: 50467180-50467180 on Build GRCh38
rs546683944 Chr.7: 50467451-50467451 on Build GRCh38
rs773297272 Chr.7: 50467271-50467271 on Build GRCh38
rs755400557 Chr.7: 50467202-50467202 on Build GRCh38
rs769942765 Chr.7: 50467270-50467270 on Build GRCh38
rs754952070 Chr.7: 50467345-50467345 on Build GRCh38
rs369370213 Chr.7: 50467292-50467292 on Build GRCh38
rs779977564 Chr.7: 50467236-50467236 on Build GRCh38
rs569139513 Chr.7: 50467546-50467546 on Build GRCh38
rs1051019447 Chr.7: 50467595-50467595 on Build GRCh38
rs1050979554 Chr.7: 50467151-50467151 on Build GRCh38
rs762003855 Chr.7: 50467164-50467164 on Build GRCh38
rs746931974 Chr.7: 50467343-50467343 on Build GRCh38
rs1001906305 Chr.7: 50467584-50467584 on Build GRCh38
rs757905175 Chr.7: 50467330-50467330 on Build GRCh38
rs932307101 Chr.7: 50467254-50467254 on Build GRCh38
rs542063660 Chr.7: 50467222-50467222 on Build GRCh38
rs951518629 Chr.7: 50467350-50467350 on Build GRCh38
rs756891812 Chr.7: 50467313-50467313 on Build GRCh38
rs1025143209 Chr.7: 50467363-50467363 on Build GRCh38
rs1025795093 Chr.7: 50467289-50467289 on Build GRCh38
rs564130357 Chr.7: 50467319-50467319 on Build GRCh38
rs144293557 Chr.7: 50467266-50467266 on Build GRCh38
rs549234079 Chr.7: 50467530-50467530 on Build GRCh38
rs980208404 Chr.7: 50467410-50467410 on Build GRCh38
rs935779704 Chr.7: 50467452-50467452 on Build GRCh38
rs753043261 Chr.7: 50467295-50467295 on Build GRCh38
rs768431322 Chr.7: 50467253-50467253 on Build GRCh38
rs752108612 Chr.7: 50467278-50467278 on Build GRCh38
rs773312663 Chr.7: 50467354-50467354 on Build GRCh38
rs753516133 Chr.7: 50467205-50467205 on Build GRCh38
rs779601267 Chr.7: 50467332-50467332 on Build GRCh38
rs1052919064 Chr.7: 50467331-50467331 on Build GRCh38
rs142777236 Chr.7: 50467529-50467529 on Build GRCh38
rs750512559 Chr.7: 50467177-50467177 on Build GRCh38
rs1040756254 Chr.7: 50467214-50467214 on Build GRCh38
rs926975198 Chr.7: 50467419-50467419 on Build GRCh38
rs765341094 Chr.7: 50467167-50467167 on Build GRCh38
rs771947566 Chr.7: 50467229-50467229 on Build GRCh38
rs898614956 Chr.7: 50467231-50467231 on Build GRCh38
rs185356649 Chr.7: 50467277-50467277 on Build GRCh38
rs113510841 Chr.7: 50467203-50467203 on Build GRCh38
rs1000120387 Chr.7: 50467300-50467300 on Build GRCh38
rs375309190 Chr.7: 50467279-50467279 on Build GRCh38
rs767018677 Chr.7: 50467190-50467190 on Build GRCh38
rs781005583 Chr.7: 50467351-50467351 on Build GRCh38
rs746737558 Chr.7: 50467238-50467238 on Build GRCh38
rs745400113 Chr.7: 50467227-50467227 on Build GRCh38
rs908066678 Chr.7: 50467199-50467199 on Build GRCh38
rs200295408 Chr.7: 50467275-50467275 on Build GRCh38
rs559651968 Chr.7: 50467192-50467192 on Build GRCh38
rs747920974 Chr.7: 50467352-50467352 on Build GRCh38
rs116739077 Chr.7: 50467515-50467515 on Build GRCh38
rs376967659 Chr.7: 50467209-50467209 on Build GRCh38
rs373529153 Chr.7: 50467267-50467267 on Build GRCh38
rs971094492 Chr.7: 50467386-50467386 on Build GRCh38
rs989870796 Chr.7: 50467467-50467467 on Build GRCh38
rs749887111 Chr.7: 50467325-50467325 on Build GRCh38
rs756907276 Chr.7: 50467221-50467221 on Build GRCh38

View Full Product Details