Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100091549-100092044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPS6
Entrez Gene ID
10980
Gene Name
COP9 signalosome subunit 6
Gene Aliases
CSN6, MOV34-34KD
UniGene
Hs.15591
Gene Chromosome Location
Chr.7: 100088960-100092200 on Build GRCh38
Transcripts
NM_006833.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6634097 c.905T>C p.M302T Chr.7: 100091710-100091710 on Build GRCh38
1093944 c.868G>A p.A290T Chr.7: 100091673-100091673 on Build GRCh38
6031331 c.870C>A p.A290A Chr.7: 100091675-100091675 on Build GRCh38
6634103 c.888_889insA p.T298fs*>31 Chr.7: 100091693-100091694 on Build GRCh38
1330422 c.944G>A p.R315Q Chr.7: 100091749-100091749 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765013312 Chr.7: 100091773-100091773 on Build GRCh38
rs752838502 Chr.7: 100091690-100091690 on Build GRCh38
rs572794934 Chr.7: 100091968-100091968 on Build GRCh38
rs761616256 Chr.7: 100091670-100091670 on Build GRCh38
rs973707596 Chr.7: 100091721-100091721 on Build GRCh38
rs1054592563 Chr.7: 100091601-100091601 on Build GRCh38
rs113803296 Chr.7: 100091908-100091908 on Build GRCh38
rs369196927 Chr.7: 100091570-100091570 on Build GRCh38
rs759619174 Chr.7: 100091777-100091777 on Build GRCh38
rs766604429 Chr.7: 100091673-100091673 on Build GRCh38
rs768730926 Chr.7: 100091619-100091619 on Build GRCh38
rs576238819 Chr.7: 100091926-100091927 on Build GRCh38
rs746244539 Chr.7: 100091831-100091831 on Build GRCh38
rs200743570 Chr.7: 100091645-100091645 on Build GRCh38
rs371103105 Chr.7: 100091644-100091644 on Build GRCh38
rs758547231 Chr.7: 100091612-100091612 on Build GRCh38
rs564389613 Chr.7: 100091969-100091969 on Build GRCh38
rs953919313 Chr.7: 100091635-100091635 on Build GRCh38
rs1029010152 Chr.7: 100091632-100091632 on Build GRCh38
rs901953261 Chr.7: 100091613-100091613 on Build GRCh38
rs757944640 Chr.7: 100091835-100091835 on Build GRCh38
rs1050378550 Chr.7: 100091912-100091912 on Build GRCh38
rs755086126 Chr.7: 100091682-100091682 on Build GRCh38
rs995553050 Chr.7: 100091887-100091887 on Build GRCh38
rs139971845 Chr.7: 100091660-100091660 on Build GRCh38
rs926966592 Chr.7: 100091798-100091798 on Build GRCh38
rs1801417 Chr.7: 100091713-100091713 on Build GRCh38
rs905479000 Chr.7: 100091852-100091852 on Build GRCh38
rs536502448 Chr.7: 100091970-100091970 on Build GRCh38
rs41280959 Chr.7: 100091871-100091871 on Build GRCh38
rs750331554 Chr.7: 100091824-100091824 on Build GRCh38
rs958612990 Chr.7: 100091987-100091987 on Build GRCh38
rs962679458 Chr.7: 100091932-100091932 on Build GRCh38
rs963028343 Chr.7: 100091884-100091884 on Build GRCh38
rs368365967 Chr.7: 100091810-100091810 on Build GRCh38
rs986030740 Chr.7: 100091907-100091907 on Build GRCh38
rs914869056 Chr.7: 100091817-100091817 on Build GRCh38
rs752787120 Chr.7: 100091790-100091790 on Build GRCh38
rs920559710 Chr.7: 100091722-100091722 on Build GRCh38
rs374625999 Chr.7: 100091948-100091948 on Build GRCh38
rs927081760 Chr.7: 100091976-100091976 on Build GRCh38
rs771460787 Chr.7: 100091717-100091717 on Build GRCh38
rs1035162212 Chr.7: 100091861-100091861 on Build GRCh38
rs774791666 Chr.7: 100091756-100091756 on Build GRCh38
rs747064885 Chr.7: 100091618-100091618 on Build GRCh38
rs911699485 Chr.7: 100091914-100091914 on Build GRCh38
rs994392006 Chr.7: 100091937-100091937 on Build GRCh38
rs372933685 Chr.7: 100091571-100091571 on Build GRCh38
rs778456150 Chr.7: 100091709-100091709 on Build GRCh38
rs780812046 Chr.7: 100091840-100091840 on Build GRCh38
rs971083554 Chr.7: 100091965-100091965 on Build GRCh38
rs775484271 Chr.7: 100091657-100091657 on Build GRCh38
rs76829163 Chr.7: 100091780-100091780 on Build GRCh38
rs575628804 Chr.7: 100091878-100091878 on Build GRCh38
rs779088504 Chr.7: 100091825-100091825 on Build GRCh38
rs760206278 Chr.7: 100091760-100091760 on Build GRCh38
rs906097475 Chr.7: 100091581-100091581 on Build GRCh38
rs555625764 Chr.7: 100091824-100091824 on Build GRCh38
rs889128097 Chr.7: 100091919-100091919 on Build GRCh38
rs143436923 Chr.7: 100091872-100091872 on Build GRCh38
rs1028748362 Chr.7: 100091897-100091897 on Build GRCh38
rs745563558 Chr.7: 100091735-100091735 on Build GRCh38
rs771547851 Chr.7: 100091656-100091656 on Build GRCh38
rs753640097 Chr.7: 100091609-100091609 on Build GRCh38
rs766355173 Chr.7: 100091839-100091839 on Build GRCh38
rs771766687 Chr.7: 100091744-100091744 on Build GRCh38
rs1006252722 Chr.7: 100091921-100091921 on Build GRCh38
rs778623027 Chr.7: 100091726-100091726 on Build GRCh38
rs751771635 Chr.7: 100091681-100091681 on Build GRCh38
rs557095125 Chr.7: 100091641-100091641 on Build GRCh38
rs546567557 Chr.7: 100092011-100092011 on Build GRCh38
rs1017259871 Chr.7: 100091926-100091926 on Build GRCh38
rs1045394766 Chr.7: 100091573-100091573 on Build GRCh38
rs192102771 Chr.7: 100091805-100091805 on Build GRCh38
rs989856851 Chr.7: 100091990-100091990 on Build GRCh38
rs1002466257 Chr.7: 100091861-100091862 on Build GRCh38
rs1011548794 Chr.7: 100091597-100091597 on Build GRCh38
rs781191573 Chr.7: 100091687-100091687 on Build GRCh38
rs574068792 Chr.7: 100091869-100091869 on Build GRCh38
rs953281569 Chr.7: 100091904-100091913 on Build GRCh38
rs748673921 Chr.7: 100091631-100091631 on Build GRCh38
rs764018352 Chr.7: 100091661-100091661 on Build GRCh38
rs188414718 Chr.7: 100091594-100091594 on Build GRCh38
rs756622620 Chr.7: 100091699-100091699 on Build GRCh38
rs781667527 Chr.7: 100091986-100091986 on Build GRCh38
rs190886795 Chr.7: 100091955-100091955 on Build GRCh38
rs61749908 Chr.7: 100091762-100091762 on Build GRCh38
rs761567342 Chr.7: 100091770-100091770 on Build GRCh38
rs1052151030 Chr.7: 100091793-100091793 on Build GRCh38
rs893305363 Chr.7: 100091604-100091604 on Build GRCh38
rs76623811 Chr.7: 100091992-100091992 on Build GRCh38
rs1052012443 Chr.7: 100091579-100091579 on Build GRCh38
rs749647179 Chr.7: 100091710-100091710 on Build GRCh38
rs754461318 Chr.7: 100091822-100091822 on Build GRCh38
rs1010437902 Chr.7: 100091605-100091605 on Build GRCh38
rs780282875 Chr.7: 100091614-100091614 on Build GRCh38
rs751797327 Chr.7: 100091774-100091774 on Build GRCh38
rs141560742 Chr.7: 100091608-100091608 on Build GRCh38
rs763076174 Chr.7: 100091639-100091642 on Build GRCh38
rs764128718 Chr.7: 100091816-100091817 on Build GRCh38
rs1044367011 Chr.7: 100091815-100091815 on Build GRCh38

View Full Product Details