Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 158182798-158183302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD1D
Entrez Gene ID
912
Gene Name
CD1d molecule
Gene Aliases
CD1A, R3, R3G1
UniGene
Hs.1799
Gene Chromosome Location
Chr.1: 158178038-158186427 on Build GRCh38
Transcripts
NM_001319145.1 NM_001766.3 XM_005245583.1 XM_006711621.1 XM_011510125.1 XM_011510127.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6564715 c.712T>G p.W238G Chr.1: 158182982-158182982 on Build GRCh38
5889465 c.802G>A p.D268N Chr.1: 158183072-158183072 on Build GRCh38
6541026 c.783G>T p.W261C Chr.1: 158183053-158183053 on Build GRCh38
1688834 c.704G>A p.W235* Chr.1: 158182974-158182974 on Build GRCh38
5802929 c.847C>G p.H283D Chr.1: 158183117-158183117 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529047977 Chr.1: 158183108-158183108 on Build GRCh38
rs777032434 Chr.1: 158183103-158183103 on Build GRCh38
rs775167221 Chr.1: 158182925-158182925 on Build GRCh38
rs763602241 Chr.1: 158183204-158183204 on Build GRCh38
rs769399031 Chr.1: 158183072-158183072 on Build GRCh38
rs369282658 Chr.1: 158183098-158183098 on Build GRCh38
rs750887347 Chr.1: 158182901-158182901 on Build GRCh38
rs765167318 Chr.1: 158182942-158182942 on Build GRCh38
rs1022444364 Chr.1: 158182986-158182986 on Build GRCh38
rs372757391 Chr.1: 158183172-158183172 on Build GRCh38
rs750404418 Chr.1: 158183121-158183121 on Build GRCh38
rs139209490 Chr.1: 158182988-158182988 on Build GRCh38
rs375390372 Chr.1: 158182918-158182918 on Build GRCh38
rs920387491 Chr.1: 158183073-158183073 on Build GRCh38
rs760058701 Chr.1: 158182981-158182981 on Build GRCh38
rs758827375 Chr.1: 158183043-158183043 on Build GRCh38
rs563918167 Chr.1: 158183274-158183274 on Build GRCh38
rs766516559 Chr.1: 158182852-158182852 on Build GRCh38
rs142766597 Chr.1: 158183151-158183151 on Build GRCh38
rs980136194 Chr.1: 158183272-158183272 on Build GRCh38
rs201516877 Chr.1: 158183192-158183192 on Build GRCh38
rs771645613 Chr.1: 158182924-158182924 on Build GRCh38
rs140583445 Chr.1: 158183141-158183141 on Build GRCh38
rs559158267 Chr.1: 158183102-158183102 on Build GRCh38
rs773245435 Chr.1: 158183010-158183010 on Build GRCh38
rs758837390 Chr.1: 158182991-158182991 on Build GRCh38
rs766418678 Chr.1: 158183179-158183179 on Build GRCh38
rs566569460 Chr.1: 158182892-158182892 on Build GRCh38
rs756299315 Chr.1: 158182889-158182889 on Build GRCh38
rs948414574 Chr.1: 158183006-158183006 on Build GRCh38
rs764933486 Chr.1: 158182843-158182844 on Build GRCh38
rs766250843 Chr.1: 158183019-158183019 on Build GRCh38
rs766059814 Chr.1: 158182952-158182952 on Build GRCh38
rs539075437 Chr.1: 158182917-158182917 on Build GRCh38
rs558792729 Chr.1: 158183226-158183226 on Build GRCh38
rs376750197 Chr.1: 158183173-158183173 on Build GRCh38
rs149772303 Chr.1: 158182996-158182996 on Build GRCh38
rs938614853 Chr.1: 158183014-158183014 on Build GRCh38
rs753062897 Chr.1: 158183006-158183007 on Build GRCh38
rs764352305 Chr.1: 158183027-158183027 on Build GRCh38
rs760028099 Chr.1: 158183188-158183188 on Build GRCh38
rs376697077 Chr.1: 158182833-158182833 on Build GRCh38
rs768212638 Chr.1: 158182929-158182929 on Build GRCh38
rs775143338 Chr.1: 158183170-158183170 on Build GRCh38
rs1019263699 Chr.1: 158183032-158183032 on Build GRCh38
rs182855664 Chr.1: 158183045-158183045 on Build GRCh38
rs144037659 Chr.1: 158183008-158183008 on Build GRCh38
rs779426050 Chr.1: 158182989-158182989 on Build GRCh38
rs774280958 Chr.1: 158183020-158183020 on Build GRCh38
rs766824137 Chr.1: 158183132-158183132 on Build GRCh38
rs569233577 Chr.1: 158182962-158182962 on Build GRCh38
rs767952763 Chr.1: 158183000-158183001 on Build GRCh38
rs143328267 Chr.1: 158183120-158183120 on Build GRCh38
rs763059094 Chr.1: 158182851-158182852 on Build GRCh38
rs201923868 Chr.1: 158183177-158183177 on Build GRCh38
rs750996933 Chr.1: 158183037-158183037 on Build GRCh38
rs575585006 Chr.1: 158183247-158183247 on Build GRCh38
rs144272399 Chr.1: 158183060-158183060 on Build GRCh38
rs200278882 Chr.1: 158183109-158183109 on Build GRCh38
rs371162796 Chr.1: 158182891-158182891 on Build GRCh38
rs773538241 Chr.1: 158182844-158182844 on Build GRCh38
rs138733189 Chr.1: 158182906-158182906 on Build GRCh38
rs1012408692 Chr.1: 158182972-158182972 on Build GRCh38
rs776115774 Chr.1: 158182936-158182936 on Build GRCh38
rs146500563 Chr.1: 158183229-158183229 on Build GRCh38
rs755802101 Chr.1: 158183200-158183200 on Build GRCh38
rs770703597 Chr.1: 158183086-158183086 on Build GRCh38
rs371913497 Chr.1: 158183033-158183033 on Build GRCh38
rs780403284 Chr.1: 158183044-158183044 on Build GRCh38
rs778952737 Chr.1: 158183154-158183154 on Build GRCh38
rs774299268 Chr.1: 158183097-158183097 on Build GRCh38
rs376559161 Chr.1: 158183080-158183080 on Build GRCh38
rs772906466 Chr.1: 158182944-158182944 on Build GRCh38
rs757319382 Chr.1: 158183030-158183030 on Build GRCh38
rs777339209 Chr.1: 158183149-158183149 on Build GRCh38
rs754650454 Chr.1: 158183049-158183049 on Build GRCh38
rs762903334 Chr.1: 158183178-158183178 on Build GRCh38
rs199831046 Chr.1: 158183062-158183062 on Build GRCh38
rs930404466 Chr.1: 158183089-158183089 on Build GRCh38
rs914299902 Chr.1: 158183209-158183209 on Build GRCh38
rs375741058 Chr.1: 158183144-158183144 on Build GRCh38
rs752114154 Chr.1: 158183185-158183185 on Build GRCh38
rs377573082 Chr.1: 158183206-158183206 on Build GRCh38
rs772985800 Chr.1: 158183129-158183129 on Build GRCh38
rs757831900 Chr.1: 158182902-158182902 on Build GRCh38
rs904013914 Chr.1: 158183157-158183157 on Build GRCh38
rs756569869 Chr.1: 158183112-158183112 on Build GRCh38
rs772041419 Chr.1: 158183162-158183162 on Build GRCh38
rs907604543 Chr.1: 158183276-158183276 on Build GRCh38
rs145809444 Chr.1: 158183148-158183148 on Build GRCh38
rs751598600 Chr.1: 158182865-158182867 on Build GRCh38
rs753014071 Chr.1: 158182957-158182957 on Build GRCh38
rs147417067 Chr.1: 158182923-158182923 on Build GRCh38
rs748004286 Chr.1: 158183024-158183024 on Build GRCh38
rs762773829 Chr.1: 158183013-158183013 on Build GRCh38
rs764333911 Chr.1: 158182983-158182983 on Build GRCh38
rs761745102 Chr.1: 158182937-158182937 on Build GRCh38
rs958230118 Chr.1: 158183189-158183189 on Build GRCh38
rs777129936 Chr.1: 158183077-158183077 on Build GRCh38
rs747612257 Chr.1: 158183002-158183002 on Build GRCh38
rs368229860 Chr.1: 158182856-158182856 on Build GRCh38
rs776133854 Chr.1: 158183001-158183001 on Build GRCh38
rs752934980 Chr.1: 158183196-158183196 on Build GRCh38
rs745768775 Chr.1: 158183159-158183159 on Build GRCh38
rs199860570 Chr.1: 158182926-158182926 on Build GRCh38
rs141805163 Chr.1: 158182945-158182945 on Build GRCh38
rs367746380 Chr.1: 158182992-158182992 on Build GRCh38
rs999650367 Chr.1: 158183161-158183161 on Build GRCh38

View Full Product Details