Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160216392-160216887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF8
Entrez Gene ID
50717
Gene Name
DDB1 and CUL4 associated factor 8
Gene Aliases
GAN2, H326, WDR42A
UniGene
Hs.632447
Gene Chromosome Location
Chr.1: 160215715-160262560 on Build GRCh38
Transcripts
NM_015726.3 NR_028103.1 NR_028104.1

SNP Detail

SNP ID Chromosome Location
rs990597378 Chr.1: 160216485-160216485 on Build GRCh38
rs543203001 Chr.1: 160216425-160216425 on Build GRCh38
rs573295388 Chr.1: 160216585-160216585 on Build GRCh38
rs938004901 Chr.1: 160216501-160216501 on Build GRCh38
rs886299981 Chr.1: 160216577-160216577 on Build GRCh38
rs1132881 Chr.1: 160216505-160216505 on Build GRCh38
rs762862285 Chr.1: 160216516-160216516 on Build GRCh38
rs532351636 Chr.1: 160216761-160216761 on Build GRCh38
rs751338030 Chr.1: 160216809-160216809 on Build GRCh38
rs967772699 Chr.1: 160216541-160216541 on Build GRCh38
rs923505694 Chr.1: 160216621-160216621 on Build GRCh38
rs1048265191 Chr.1: 160216556-160216556 on Build GRCh38
rs1001381055 Chr.1: 160216864-160216864 on Build GRCh38
rs993883496 Chr.1: 160216428-160216428 on Build GRCh38
rs572525144 Chr.1: 160216767-160216767 on Build GRCh38
rs896488365 Chr.1: 160216524-160216524 on Build GRCh38
rs1006444331 Chr.1: 160216602-160216602 on Build GRCh38
rs1029991808 Chr.1: 160216841-160216841 on Build GRCh38
rs1001310811 Chr.1: 160216454-160216455 on Build GRCh38
rs971310072 Chr.1: 160216445-160216445 on Build GRCh38
rs564960698 Chr.1: 160216698-160216698 on Build GRCh38
rs1024638827 Chr.1: 160216432-160216432 on Build GRCh38
rs1034116548 Chr.1: 160216458-160216458 on Build GRCh38
rs756621717 Chr.1: 160216682-160216682 on Build GRCh38
rs932195324 Chr.1: 160216626-160216626 on Build GRCh38
rs906890492 Chr.1: 160216845-160216845 on Build GRCh38
rs189272552 Chr.1: 160216633-160216633 on Build GRCh38
rs896942833 Chr.1: 160216717-160216717 on Build GRCh38
rs957217500 Chr.1: 160216483-160216484 on Build GRCh38
rs544135623 Chr.1: 160216844-160216844 on Build GRCh38
rs1045430567 Chr.1: 160216834-160216835 on Build GRCh38
rs909341883 Chr.1: 160216659-160216659 on Build GRCh38
rs1013712832 Chr.1: 160216539-160216539 on Build GRCh38
rs964565813 Chr.1: 160216695-160216695 on Build GRCh38
rs995800921 Chr.1: 160216798-160216798 on Build GRCh38
rs994219006 Chr.1: 160216786-160216786 on Build GRCh38
rs763651499 Chr.1: 160216563-160216563 on Build GRCh38

View Full Product Details