Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 105996170-105996667 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDHR3
Entrez Gene ID
222256
Gene Name
cadherin related family member 3
Gene Aliases
CDH28
UniGene
Hs.150120
Gene Chromosome Location
Chr.7: 105876773-106036431 on Build GRCh38
Transcripts
NM_001301161.1 NM_152750.4 XM_005250224.4 XM_005250225.2 XM_006715905.2 XM_011515955.2 XM_011515956.2 XM_011515957.2 XM_011515958.2 XM_017011862.1 XM_017011863.1 XM_017011864.1 XM_017011865.1 XM_017011866.1 XM_017011867.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6898343 c.670G>A p.V224M Chr.7: 105996311-105996311 on Build GRCh38
5406155 c.621C>T p.I207I Chr.7: 105996262-105996262 on Build GRCh38
3631690 c.618C>T p.L206L Chr.7: 105996259-105996259 on Build GRCh38
5406157 c.621C>T p.I207I Chr.7: 105996262-105996262 on Build GRCh38
3745453 c.704G>A p.R235H Chr.7: 105996345-105996345 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs552936182 Chr.7: 105996241-105996241 on Build GRCh38
rs958525623 Chr.7: 105996487-105996487 on Build GRCh38
rs779530979 Chr.7: 105996336-105996336 on Build GRCh38
rs929528816 Chr.7: 105996617-105996617 on Build GRCh38
rs576023062 Chr.7: 105996396-105996396 on Build GRCh38
rs925964035 Chr.7: 105996543-105996543 on Build GRCh38
rs765032086 Chr.7: 105996395-105996395 on Build GRCh38
rs574877411 Chr.7: 105996409-105996409 on Build GRCh38
rs571961979 Chr.7: 105996414-105996414 on Build GRCh38
rs982938333 Chr.7: 105996435-105996435 on Build GRCh38
rs529844085 Chr.7: 105996465-105996465 on Build GRCh38
rs146976293 Chr.7: 105996540-105996540 on Build GRCh38
rs1047067602 Chr.7: 105996515-105996515 on Build GRCh38
rs771493524 Chr.7: 105996314-105996314 on Build GRCh38
rs1005282009 Chr.7: 105996442-105996442 on Build GRCh38
rs761786153 Chr.7: 105996392-105996392 on Build GRCh38
rs762421624 Chr.7: 105996216-105996216 on Build GRCh38
rs777649960 Chr.7: 105996262-105996262 on Build GRCh38
rs765300014 Chr.7: 105996407-105996407 on Build GRCh38
rs200623269 Chr.7: 105996345-105996345 on Build GRCh38
rs756987349 Chr.7: 105996272-105996272 on Build GRCh38
rs561176599 Chr.7: 105996423-105996423 on Build GRCh38
rs887326153 Chr.7: 105996270-105996270 on Build GRCh38
rs986230735 Chr.7: 105996558-105996558 on Build GRCh38
rs1010056397 Chr.7: 105996582-105996582 on Build GRCh38
rs766709153 Chr.7: 105996237-105996237 on Build GRCh38
rs942407207 Chr.7: 105996221-105996221 on Build GRCh38
rs759677955 Chr.7: 105996246-105996246 on Build GRCh38
rs751944987 Chr.7: 105996539-105996539 on Build GRCh38
rs371882137 Chr.7: 105996402-105996402 on Build GRCh38
rs760874555 Chr.7: 105996383-105996383 on Build GRCh38
rs775517689 Chr.7: 105996379-105996379 on Build GRCh38
rs771544793 Chr.7: 105996366-105996366 on Build GRCh38
rs768470118 Chr.7: 105996206-105996206 on Build GRCh38
rs767587815 Chr.7: 105996328-105996328 on Build GRCh38
rs746452146 Chr.7: 105996298-105996298 on Build GRCh38
rs772345425 Chr.7: 105996370-105996370 on Build GRCh38
rs396600 Chr.7: 105996561-105996561 on Build GRCh38
rs746180577 Chr.7: 105996369-105996369 on Build GRCh38
rs375566112 Chr.7: 105996320-105996320 on Build GRCh38
rs73412250 Chr.7: 105996415-105996415 on Build GRCh38
rs925966801 Chr.7: 105996389-105996389 on Build GRCh38
rs774805085 Chr.7: 105996367-105996367 on Build GRCh38
rs749802725 Chr.7: 105996308-105996308 on Build GRCh38
rs771109526 Chr.7: 105996269-105996269 on Build GRCh38
rs1046679744 Chr.7: 105996288-105996288 on Build GRCh38
rs535238500 Chr.7: 105996332-105996332 on Build GRCh38
rs574450144 Chr.7: 105996315-105996315 on Build GRCh38
rs1019461220 Chr.7: 105996616-105996616 on Build GRCh38
rs945976205 Chr.7: 105996368-105996368 on Build GRCh38
rs778665117 Chr.7: 105996277-105996277 on Build GRCh38
rs745598665 Chr.7: 105996279-105996279 on Build GRCh38
rs946092313 Chr.7: 105996196-105996196 on Build GRCh38
rs200019471 Chr.7: 105996263-105996263 on Build GRCh38
rs1002229230 Chr.7: 105996313-105996313 on Build GRCh38
rs764605734 Chr.7: 105996510-105996510 on Build GRCh38
rs76140210 Chr.7: 105996591-105996591 on Build GRCh38
rs774550441 Chr.7: 105996319-105996319 on Build GRCh38
rs113955296 Chr.7: 105996229-105996229 on Build GRCh38
rs372809296 Chr.7: 105996289-105996289 on Build GRCh38
rs544571491 Chr.7: 105996408-105996408 on Build GRCh38
rs563612969 Chr.7: 105996550-105996550 on Build GRCh38
rs1049799282 Chr.7: 105996624-105996624 on Build GRCh38
rs944774200 Chr.7: 105996563-105996563 on Build GRCh38
rs1024097633 Chr.7: 105996534-105996534 on Build GRCh38
rs957625655 Chr.7: 105996555-105996555 on Build GRCh38
rs963666842 Chr.7: 105996407-105996408 on Build GRCh38
rs572514563 Chr.7: 105996334-105996334 on Build GRCh38
rs1034550869 Chr.7: 105996469-105996469 on Build GRCh38
rs368906068 Chr.7: 105996436-105996436 on Build GRCh38
rs770451107 Chr.7: 105996301-105996301 on Build GRCh38
rs760682075 Chr.7: 105996330-105996330 on Build GRCh38
rs77856426 Chr.7: 105996388-105996388 on Build GRCh38
rs754369922 Chr.7: 105996379-105996379 on Build GRCh38
rs1008088187 Chr.7: 105996596-105996596 on Build GRCh38
rs371586125 Chr.7: 105996335-105996335 on Build GRCh38
rs746528834 Chr.7: 105996342-105996342 on Build GRCh38
rs1041777166 Chr.7: 105996198-105996198 on Build GRCh38
rs368294260 Chr.7: 105996331-105996331 on Build GRCh38
rs764225043 Chr.7: 105996387-105996387 on Build GRCh38
rs374571701 Chr.7: 105996230-105996230 on Build GRCh38
rs752700057 Chr.7: 105996250-105996250 on Build GRCh38
rs566350612 Chr.7: 105996253-105996253 on Build GRCh38
rs888136210 Chr.7: 105996424-105996424 on Build GRCh38
rs775672486 Chr.7: 105996329-105996329 on Build GRCh38
rs73195651 Chr.7: 105996628-105996628 on Build GRCh38
rs201595088 Chr.7: 105996344-105996344 on Build GRCh38
rs773784444 Chr.7: 105996304-105996304 on Build GRCh38
rs757955102 Chr.7: 105996284-105996284 on Build GRCh38
rs770399813 Chr.7: 105996227-105996227 on Build GRCh38
rs34616261 Chr.7: 105996240-105996241 on Build GRCh38
rs566389124 Chr.7: 105996595-105996595 on Build GRCh38
rs1050593119 Chr.7: 105996236-105996236 on Build GRCh38

View Full Product Details