Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 117530821-117531337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFTR
Entrez Gene ID
1080
Gene Name
cystic fibrosis transmembrane conductance regulator
Gene Aliases
ABC35, ABCC7, CF, CFTR/MRP, MRP7, TNR-CFTR, dJ760C5.1
UniGene
Hs.489786
Gene Chromosome Location
Chr.7: 117478367-117668665 on Build GRCh38
Transcripts
NM_000492.3 XM_011515751.2 XM_011515753.1 XM_011515754.2 XM_017011699.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3726295 c.379T>G p.L127V Chr.7: 117531004-117531004 on Build GRCh38
6489588 c.431G>A p.G144D Chr.7: 117531056-117531056 on Build GRCh38
4405635 c.299T>G p.L100R Chr.7: 117530924-117530924 on Build GRCh38
6175950 c.419C>T p.P140L Chr.7: 117531044-117531044 on Build GRCh38
4635465 c.456G>A p.M152I Chr.7: 117531081-117531081 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017175032 Chr.7: 117531308-117531308 on Build GRCh38
rs397508537 Chr.7: 117530955-117530955 on Build GRCh38
rs79660178 Chr.7: 117530991-117530991 on Build GRCh38
rs397508425 Chr.7: 117530889-117530889 on Build GRCh38
rs759310470 Chr.7: 117531066-117531066 on Build GRCh38
rs575711764 Chr.7: 117530890-117530890 on Build GRCh38
rs77834169 Chr.7: 117530974-117530974 on Build GRCh38
rs397508725 Chr.7: 117531098-117531098 on Build GRCh38
rs397508679 Chr.7: 117531037-117531038 on Build GRCh38
rs751674090 Chr.7: 117531159-117531159 on Build GRCh38
rs397508520 Chr.7: 117530948-117530948 on Build GRCh38
rs78655421 Chr.7: 117530975-117530975 on Build GRCh38
rs397508727 Chr.7: 117531101-117531101 on Build GRCh38
rs1001491596 Chr.7: 117531302-117531302 on Build GRCh38
rs397508551 Chr.7: 117530963-117530963 on Build GRCh38
rs397508718 Chr.7: 117531070-117531070 on Build GRCh38
rs770241677 Chr.7: 117530960-117530960 on Build GRCh38
rs78756941 Chr.7: 117531115-117531115 on Build GRCh38
rs397508554 Chr.7: 117530965-117530965 on Build GRCh38
rs193922518 Chr.7: 117530980-117530980 on Build GRCh38
rs757136340 Chr.7: 117531094-117531094 on Build GRCh38
rs397508486 Chr.7: 117530928-117530929 on Build GRCh38
rs568263792 Chr.7: 117531215-117531215 on Build GRCh38
rs397508647 Chr.7: 117531018-117531018 on Build GRCh38
rs193922519 Chr.7: 117530995-117530995 on Build GRCh38
rs397508526 Chr.7: 117530951-117530952 on Build GRCh38
rs397508733 Chr.7: 117531114-117531114 on Build GRCh38
rs377729736 Chr.7: 117531117-117531117 on Build GRCh38
rs749801869 Chr.7: 117531019-117531019 on Build GRCh38
rs1024092547 Chr.7: 117530941-117530941 on Build GRCh38
rs397508672 Chr.7: 117531034-117531034 on Build GRCh38
rs557602723 Chr.7: 117530847-117530847 on Build GRCh38
rs905760728 Chr.7: 117531278-117531278 on Build GRCh38
rs397508449 Chr.7: 117530912-117530912 on Build GRCh38
rs535616800 Chr.7: 117531245-117531245 on Build GRCh38
rs764227419 Chr.7: 117531077-117531077 on Build GRCh38
rs397508686 Chr.7: 117531039-117531040 on Build GRCh38
rs397508571 Chr.7: 117530971-117530971 on Build GRCh38
rs397508717 Chr.7: 117531069-117531070 on Build GRCh38
rs397508609 Chr.7: 117531002-117531002 on Build GRCh38
rs761423802 Chr.7: 117530904-117530904 on Build GRCh38
rs397508583 Chr.7: 117530982-117530982 on Build GRCh38
rs397508719 Chr.7: 117531071-117531071 on Build GRCh38
rs778204477 Chr.7: 117530946-117530946 on Build GRCh38
rs1024941632 Chr.7: 117531194-117531194 on Build GRCh38
rs397508522 Chr.7: 117530950-117530950 on Build GRCh38
rs397508627 Chr.7: 117531012-117531012 on Build GRCh38
rs758675549 Chr.7: 117530938-117530938 on Build GRCh38
rs397508694 Chr.7: 117531044-117531044 on Build GRCh38
rs773684025 Chr.7: 117530853-117530853 on Build GRCh38
rs397508484 Chr.7: 117530927-117530927 on Build GRCh38
rs776878618 Chr.7: 117530958-117530958 on Build GRCh38
rs1021474992 Chr.7: 117530886-117530886 on Build GRCh38
rs1014161183 Chr.7: 117530936-117530936 on Build GRCh38
rs397508541 Chr.7: 117530956-117530956 on Build GRCh38
rs397508530 Chr.7: 117530953-117530953 on Build GRCh38
rs764174585 Chr.7: 117531146-117531146 on Build GRCh38
rs762950441 Chr.7: 117531140-117531140 on Build GRCh38
rs769423740 Chr.7: 117531136-117531136 on Build GRCh38
rs767204327 Chr.7: 117530906-117530906 on Build GRCh38
rs747429689 Chr.7: 117530865-117530865 on Build GRCh38
rs397508592 Chr.7: 117530989-117530989 on Build GRCh38
rs908247802 Chr.7: 117531261-117531261 on Build GRCh38
rs149706251 Chr.7: 117530969-117530969 on Build GRCh38
rs996641773 Chr.7: 117530867-117530867 on Build GRCh38
rs397508426 Chr.7: 117530897-117530897 on Build GRCh38
rs950028791 Chr.7: 117531267-117531267 on Build GRCh38
rs397508516 Chr.7: 117530944-117530951 on Build GRCh38
rs113993958 Chr.7: 117530953-117530953 on Build GRCh38
rs397508712 Chr.7: 117531059-117531059 on Build GRCh38
rs1039221071 Chr.7: 117531018-117531018 on Build GRCh38
rs145900055 Chr.7: 117531043-117531043 on Build GRCh38
rs121908798 Chr.7: 117530950-117530952 on Build GRCh38
rs780492177 Chr.7: 117531016-117531016 on Build GRCh38
rs397508674 Chr.7: 117531035-117531035 on Build GRCh38
rs1025401020 Chr.7: 117531162-117531162 on Build GRCh38
rs397508729 Chr.7: 117531106-117531106 on Build GRCh38
rs371315549 Chr.7: 117530893-117530893 on Build GRCh38
rs121909031 Chr.7: 117530951-117530951 on Build GRCh38
rs397508606 Chr.7: 117531001-117531001 on Build GRCh38
rs397508728 Chr.7: 117531105-117531105 on Build GRCh38
rs121908792 Chr.7: 117530898-117530898 on Build GRCh38
rs943659024 Chr.7: 117531196-117531196 on Build GRCh38
rs397508671 Chr.7: 117531034-117531037 on Build GRCh38
rs769183771 Chr.7: 117531100-117531100 on Build GRCh38
rs749836021 Chr.7: 117531058-117531058 on Build GRCh38
rs193922521 Chr.7: 117531007-117531008 on Build GRCh38
rs397508726 Chr.7: 117531098-117531099 on Build GRCh38
rs531830892 Chr.7: 117531139-117531139 on Build GRCh38
rs121908751 Chr.7: 117530899-117530899 on Build GRCh38
rs770608125 Chr.7: 117531121-117531121 on Build GRCh38
rs397508509 Chr.7: 117530939-117530939 on Build GRCh38
rs141723617 Chr.7: 117530999-117530999 on Build GRCh38
rs574654063 Chr.7: 117530986-117530986 on Build GRCh38
rs397508432 Chr.7: 117530901-117530901 on Build GRCh38
rs986478134 Chr.7: 117531193-117531193 on Build GRCh38
rs56094102 Chr.7: 117531205-117531205 on Build GRCh38
rs397508720 Chr.7: 117531076-117531076 on Build GRCh38
rs761864917 Chr.7: 117531160-117531160 on Build GRCh38
rs752619770 Chr.7: 117531075-117531075 on Build GRCh38
rs387906364 Chr.7: 117531054-117531054 on Build GRCh38
rs761370893 Chr.7: 117530972-117530972 on Build GRCh38
rs387906371 Chr.7: 117531084-117531101 on Build GRCh38
rs35516286 Chr.7: 117531068-117531068 on Build GRCh38
rs397508713 Chr.7: 117531062-117531062 on Build GRCh38
rs781327181 Chr.7: 117531086-117531086 on Build GRCh38
rs1800078 Chr.7: 117531038-117531038 on Build GRCh38
rs771512600 Chr.7: 117531028-117531028 on Build GRCh38
rs954990448 Chr.7: 117531164-117531164 on Build GRCh38
rs397508700 Chr.7: 117531047-117531047 on Build GRCh38
rs771557069 Chr.7: 117530887-117530887 on Build GRCh38
rs397508464 Chr.7: 117530918-117530918 on Build GRCh38
rs397508490 Chr.7: 117530930-117530930 on Build GRCh38
rs397508721 Chr.7: 117531079-117531079 on Build GRCh38
rs397508731 Chr.7: 117531109-117531109 on Build GRCh38
rs201958172 Chr.7: 117530983-117530983 on Build GRCh38
rs397508528 Chr.7: 117530952-117530952 on Build GRCh38
rs943677386 Chr.7: 117531067-117531067 on Build GRCh38
rs754993212 Chr.7: 117530887-117530887 on Build GRCh38
rs397508687 Chr.7: 117531040-117531041 on Build GRCh38
rs760281820 Chr.7: 117531042-117531042 on Build GRCh38
rs397508732 Chr.7: 117531116-117531116 on Build GRCh38
rs1031657153 Chr.7: 117530920-117530920 on Build GRCh38
rs76371115 Chr.7: 117531041-117531041 on Build GRCh38
rs909596175 Chr.7: 117530961-117530961 on Build GRCh38
rs121908801 Chr.7: 117530938-117530938 on Build GRCh38
rs397508723 Chr.7: 117531088-117531088 on Build GRCh38
rs397508611 Chr.7: 117531005-117531005 on Build GRCh38
rs397508722 Chr.7: 117531080-117531080 on Build GRCh38
rs397508461 Chr.7: 117530917-117530917 on Build GRCh38
rs35871908 Chr.7: 117530978-117530979 on Build GRCh38
rs972514866 Chr.7: 117531155-117531155 on Build GRCh38
rs727504712 Chr.7: 117531122-117531122 on Build GRCh38
rs397508632 Chr.7: 117531013-117531013 on Build GRCh38
rs397508730 Chr.7: 117531107-117531107 on Build GRCh38
rs397508563 Chr.7: 117530968-117530970 on Build GRCh38
rs149197463 Chr.7: 117531083-117531083 on Build GRCh38
rs397508724 Chr.7: 117531097-117531097 on Build GRCh38
rs746344714 Chr.7: 117530959-117530959 on Build GRCh38
rs377295859 Chr.7: 117530990-117530990 on Build GRCh38
rs374689323 Chr.7: 117531091-117531091 on Build GRCh38
rs121908770 Chr.7: 117531067-117531067 on Build GRCh38
rs969399514 Chr.7: 117531201-117531201 on Build GRCh38
rs397508467 Chr.7: 117530921-117530921 on Build GRCh38
rs965163927 Chr.7: 117531299-117531299 on Build GRCh38
rs375543675 Chr.7: 117530923-117530923 on Build GRCh38
rs397508698 Chr.7: 117531045-117531046 on Build GRCh38
rs898290568 Chr.7: 117531311-117531311 on Build GRCh38
rs397508499 Chr.7: 117530935-117530935 on Build GRCh38
rs982987179 Chr.7: 117531260-117531260 on Build GRCh38
rs1045750610 Chr.7: 117531269-117531269 on Build GRCh38
rs387906363 Chr.7: 117531049-117531049 on Build GRCh38
rs772733538 Chr.7: 117531030-117531030 on Build GRCh38
rs140502196 Chr.7: 117530957-117530957 on Build GRCh38
rs1800077 Chr.7: 117530985-117530985 on Build GRCh38
rs369715785 Chr.7: 117530935-117530935 on Build GRCh38

View Full Product Details