Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 122345434-122345973 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CADPS2
Entrez Gene ID
93664
Gene Name
calcium dependent secretion activator 2
Gene Aliases
CAPS2
UniGene
Hs.649459
Gene Chromosome Location
Chr.7: 122318424-122886759 on Build GRCh38
Transcripts
NM_001009571.3 NM_001167940.1 NM_017954.10 XM_005250695.4 XM_005250696.4 XM_005250697.4 XM_005250698.4 XM_005250699.4 XM_005250700.4 XM_005250701.4 XM_005250702.4 XM_005250703.4 XM_005250704.4 XM_005250705.4 XM_005250706.4 XM_005250707.4 XM_005250708.3 XM_011516695.2 XM_011516696.2 XM_011516697.2 XM_017012794.1 XM_017012795.1 XM_017012796.1 XM_017012797.1 XM_017012798.1 XM_017012799.1 XM_017012800.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5928901 c.1098G>A p.M366I Chr.7: 122345669-122345669 on Build GRCh38
1447647 c.3554delA p.N1185fs*15 Chr.7: 122345632-122345632 on Build GRCh38
6251344 c.3508C>A p.P1170T Chr.7: 122345678-122345678 on Build GRCh38
6108320 c.3418T>C p.F1140L Chr.7: 122345645-122345645 on Build GRCh38
744822 c.3548G>C p.R1183P Chr.7: 122345638-122345638 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763748395 Chr.7: 122345657-122345657 on Build GRCh38
rs889022645 Chr.7: 122345717-122345717 on Build GRCh38
rs201104778 Chr.7: 122345579-122345579 on Build GRCh38
rs780021942 Chr.7: 122345591-122345591 on Build GRCh38
rs17144272 Chr.7: 122345728-122345728 on Build GRCh38
rs774661345 Chr.7: 122345606-122345606 on Build GRCh38
rs187675901 Chr.7: 122345802-122345802 on Build GRCh38
rs961510682 Chr.7: 122345856-122345856 on Build GRCh38
rs748345306 Chr.7: 122345723-122345723 on Build GRCh38
rs528585432 Chr.7: 122345590-122345590 on Build GRCh38
rs798685 Chr.7: 122345487-122345487 on Build GRCh38
rs766259642 Chr.7: 122345694-122345694 on Build GRCh38
rs116019460 Chr.7: 122345933-122345933 on Build GRCh38
rs545215047 Chr.7: 122345685-122345685 on Build GRCh38
rs398006076 Chr.7: 122345932-122345933 on Build GRCh38
rs951762341 Chr.7: 122345917-122345917 on Build GRCh38
rs1025211598 Chr.7: 122345747-122345747 on Build GRCh38
rs798684 Chr.7: 122345502-122345502 on Build GRCh38
rs71159791 Chr.7: 122345918-122345919 on Build GRCh38
rs138695640 Chr.7: 122345539-122345539 on Build GRCh38
rs372248816 Chr.7: 122345844-122345844 on Build GRCh38
rs957149507 Chr.7: 122345758-122345758 on Build GRCh38
rs773731270 Chr.7: 122345730-122345730 on Build GRCh38
rs910474606 Chr.7: 122345523-122345523 on Build GRCh38
rs386411171 Chr.7: 122345928-122345929 on Build GRCh38
rs920007758 Chr.7: 122345905-122345905 on Build GRCh38
rs749350918 Chr.7: 122345595-122345595 on Build GRCh38
rs558395938 Chr.7: 122345939-122345939 on Build GRCh38
rs756987418 Chr.7: 122345559-122345559 on Build GRCh38
rs200528100 Chr.7: 122345618-122345618 on Build GRCh38
rs757705782 Chr.7: 122345503-122345503 on Build GRCh38
rs762458379 Chr.7: 122345650-122345650 on Build GRCh38
rs751209797 Chr.7: 122345659-122345659 on Build GRCh38
rs566384561 Chr.7: 122345903-122345903 on Build GRCh38
rs377072400 Chr.7: 122345688-122345688 on Build GRCh38
rs776309308 Chr.7: 122345552-122345552 on Build GRCh38
rs974234512 Chr.7: 122345861-122345861 on Build GRCh38
rs13232404 Chr.7: 122345472-122345472 on Build GRCh38
rs762329407 Chr.7: 122345614-122345614 on Build GRCh38
rs768767842 Chr.7: 122345597-122345597 on Build GRCh38
rs781174376 Chr.7: 122345571-122345571 on Build GRCh38
rs753741025 Chr.7: 122345916-122345917 on Build GRCh38
rs908810714 Chr.7: 122345849-122345850 on Build GRCh38
rs780944935 Chr.7: 122345663-122345663 on Build GRCh38
rs1050278197 Chr.7: 122345911-122345911 on Build GRCh38
rs761061448 Chr.7: 122345639-122345639 on Build GRCh38
rs149384835 Chr.7: 122345722-122345722 on Build GRCh38
rs1026022636 Chr.7: 122345908-122345908 on Build GRCh38
rs182511823 Chr.7: 122345738-122345738 on Build GRCh38
rs565464129 Chr.7: 122345690-122345690 on Build GRCh38
rs140351965 Chr.7: 122345511-122345511 on Build GRCh38
rs759168115 Chr.7: 122345682-122345682 on Build GRCh38
rs756040315 Chr.7: 122345686-122345686 on Build GRCh38
rs796722862 Chr.7: 122345724-122345724 on Build GRCh38
rs781679582 Chr.7: 122345560-122345560 on Build GRCh38
rs538712262 Chr.7: 122345909-122345909 on Build GRCh38
rs191222311 Chr.7: 122345460-122345460 on Build GRCh38
rs912541305 Chr.7: 122345930-122345930 on Build GRCh38
rs150197608 Chr.7: 122345514-122345514 on Build GRCh38
rs764619203 Chr.7: 122345530-122345530 on Build GRCh38
rs932771692 Chr.7: 122345910-122345910 on Build GRCh38
rs772556975 Chr.7: 122345623-122345623 on Build GRCh38
rs759147878 Chr.7: 122345527-122345527 on Build GRCh38
rs902990808 Chr.7: 122345736-122345738 on Build GRCh38
rs546184701 Chr.7: 122345891-122345891 on Build GRCh38
rs79928621 Chr.7: 122345945-122345945 on Build GRCh38
rs971503838 Chr.7: 122345775-122345775 on Build GRCh38
rs554876689 Chr.7: 122345515-122345515 on Build GRCh38
rs940209011 Chr.7: 122345550-122345550 on Build GRCh38
rs768847859 Chr.7: 122345705-122345705 on Build GRCh38
rs766859626 Chr.7: 122345647-122345647 on Build GRCh38
rs1032745568 Chr.7: 122345740-122345743 on Build GRCh38
rs377738414 Chr.7: 122345918-122345918 on Build GRCh38
rs903938649 Chr.7: 122345692-122345692 on Build GRCh38
rs75984157 Chr.7: 122345535-122345535 on Build GRCh38
rs971471259 Chr.7: 122345814-122345814 on Build GRCh38
rs568679701 Chr.7: 122345500-122345500 on Build GRCh38
rs1049791633 Chr.7: 122345713-122345713 on Build GRCh38
rs754851028 Chr.7: 122345638-122345638 on Build GRCh38
rs981690986 Chr.7: 122345836-122345836 on Build GRCh38
rs758008164 Chr.7: 122345532-122345532 on Build GRCh38
rs200631630 Chr.7: 122345534-122345534 on Build GRCh38

View Full Product Details