Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 152852036-152852541 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTR3B
Entrez Gene ID
57180
Gene Name
ARP3 actin related protein 3 homolog B
Gene Aliases
ARP11, ARP3BETA
UniGene
Hs.647117
Gene Chromosome Location
Chr.7: 152759749-153070979 on Build GRCh38
Transcripts
NM_001040135.2 NM_020445.5 NR_073000.1 NR_073001.1 XM_006716065.2 XM_011516425.2 XM_011516426.2 XM_017012460.1 XM_017012461.1 XM_017012462.1 XM_017012463.1 XM_017012464.1 XM_017012465.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1181740 c.1066G>A p.G356R Chr.7: 152852240-152852240 on Build GRCh38
6593108 c.1032T>C p.D344D Chr.7: 152852206-152852206 on Build GRCh38
5014775 c.1070G>A p.R357K Chr.7: 152852244-152852244 on Build GRCh38
4937889 c.963C>G p.L321L Chr.7: 152852137-152852137 on Build GRCh38
6593112 c.994G>A p.G332R Chr.7: 152852168-152852168 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367772022 Chr.7: 152852294-152852294 on Build GRCh38
rs947984444 Chr.7: 152852225-152852225 on Build GRCh38
rs769937667 Chr.7: 152852149-152852149 on Build GRCh38
rs769349115 Chr.7: 152852293-152852293 on Build GRCh38
rs1015013310 Chr.7: 152852468-152852468 on Build GRCh38
rs531521474 Chr.7: 152852464-152852464 on Build GRCh38
rs1013309996 Chr.7: 152852470-152852470 on Build GRCh38
rs896129074 Chr.7: 152852469-152852469 on Build GRCh38
rs757533169 Chr.7: 152852140-152852140 on Build GRCh38
rs944133063 Chr.7: 152852374-152852374 on Build GRCh38
rs951671715 Chr.7: 152852098-152852098 on Build GRCh38
rs973397082 Chr.7: 152852356-152852356 on Build GRCh38
rs569167262 Chr.7: 152852322-152852322 on Build GRCh38
rs112613087 Chr.7: 152852328-152852328 on Build GRCh38
rs1023442607 Chr.7: 152852329-152852329 on Build GRCh38
rs866194211 Chr.7: 152852315-152852315 on Build GRCh38
rs139229319 Chr.7: 152852236-152852236 on Build GRCh38
rs766273957 Chr.7: 152852111-152852111 on Build GRCh38
rs202105401 Chr.7: 152852273-152852273 on Build GRCh38
rs545115430 Chr.7: 152852360-152852360 on Build GRCh38
rs760198211 Chr.7: 152852105-152852105 on Build GRCh38
rs180686003 Chr.7: 152852372-152852372 on Build GRCh38
rs1001781499 Chr.7: 152852327-152852327 on Build GRCh38
rs1039788473 Chr.7: 152852385-152852385 on Build GRCh38
rs746900743 Chr.7: 152852098-152852098 on Build GRCh38
rs762259780 Chr.7: 152852224-152852224 on Build GRCh38
rs753833874 Chr.7: 152852113-152852113 on Build GRCh38
rs1008802131 Chr.7: 152852058-152852058 on Build GRCh38
rs751560555 Chr.7: 152852475-152852475 on Build GRCh38
rs764553139 Chr.7: 152852087-152852087 on Build GRCh38
rs969224710 Chr.7: 152852489-152852489 on Build GRCh38
rs553681103 Chr.7: 152852366-152852366 on Build GRCh38
rs750688445 Chr.7: 152852277-152852277 on Build GRCh38
rs780192495 Chr.7: 152852287-152852287 on Build GRCh38
rs920592354 Chr.7: 152852375-152852375 on Build GRCh38
rs115650402 Chr.7: 152852407-152852407 on Build GRCh38
rs772591461 Chr.7: 152852298-152852298 on Build GRCh38
rs770666784 Chr.7: 152852281-152852282 on Build GRCh38
rs571378039 Chr.7: 152852102-152852102 on Build GRCh38
rs752529156 Chr.7: 152852270-152852270 on Build GRCh38
rs904797641 Chr.7: 152852312-152852312 on Build GRCh38
rs745983721 Chr.7: 152852146-152852146 on Build GRCh38
rs370586367 Chr.7: 152852295-152852295 on Build GRCh38
rs1052562824 Chr.7: 152852253-152852253 on Build GRCh38
rs369637549 Chr.7: 152852263-152852263 on Build GRCh38
rs773411016 Chr.7: 152852299-152852299 on Build GRCh38
rs757601219 Chr.7: 152852500-152852500 on Build GRCh38
rs373998353 Chr.7: 152852370-152852370 on Build GRCh38
rs753221447 Chr.7: 152852248-152852248 on Build GRCh38
rs760382238 Chr.7: 152852301-152852301 on Build GRCh38
rs146208789 Chr.7: 152852221-152852221 on Build GRCh38
rs764191568 Chr.7: 152852427-152852427 on Build GRCh38
rs1023326502 Chr.7: 152852486-152852486 on Build GRCh38
rs139903799 Chr.7: 152852190-152852190 on Build GRCh38
rs150589116 Chr.7: 152852131-152852131 on Build GRCh38
rs371487908 Chr.7: 152852483-152852483 on Build GRCh38
rs887356772 Chr.7: 152852456-152852456 on Build GRCh38
rs749311514 Chr.7: 152852290-152852290 on Build GRCh38
rs979232586 Chr.7: 152852491-152852491 on Build GRCh38
rs369887594 Chr.7: 152852264-152852283 on Build GRCh38
rs776044411 Chr.7: 152852083-152852083 on Build GRCh38
rs953473068 Chr.7: 152852381-152852381 on Build GRCh38
rs1014341452 Chr.7: 152852345-152852345 on Build GRCh38
rs758166655 Chr.7: 152852272-152852272 on Build GRCh38
rs765947348 Chr.7: 152852244-152852244 on Build GRCh38
rs113145340 Chr.7: 152852305-152852305 on Build GRCh38
rs983247136 Chr.7: 152852121-152852121 on Build GRCh38
rs756221022 Chr.7: 152852285-152852285 on Build GRCh38
rs144915052 Chr.7: 152852239-152852239 on Build GRCh38
rs925194478 Chr.7: 152852365-152852365 on Build GRCh38
rs375488425 Chr.7: 152852175-152852175 on Build GRCh38
rs141763187 Chr.7: 152852210-152852210 on Build GRCh38
rs577779823 Chr.7: 152852348-152852348 on Build GRCh38
rs781361308 Chr.7: 152852145-152852145 on Build GRCh38
rs149943653 Chr.7: 152852084-152852084 on Build GRCh38
rs960236395 Chr.7: 152852341-152852341 on Build GRCh38
rs375397151 Chr.7: 152852302-152852302 on Build GRCh38
rs779954922 Chr.7: 152852183-152852183 on Build GRCh38
rs1027223962 Chr.7: 152852309-152852309 on Build GRCh38
rs951777611 Chr.7: 152852314-152852314 on Build GRCh38
rs146450910 Chr.7: 152852347-152852347 on Build GRCh38
rs77620966 Chr.7: 152852088-152852088 on Build GRCh38
rs960740646 Chr.7: 152852325-152852325 on Build GRCh38
rs900026432 Chr.7: 152852393-152852393 on Build GRCh38
rs561186926 Chr.7: 152852445-152852445 on Build GRCh38
rs956842437 Chr.7: 152852516-152852516 on Build GRCh38
rs772980591 Chr.7: 152852237-152852237 on Build GRCh38
rs772764869 Chr.7: 152852167-152852167 on Build GRCh38
rs752433041 Chr.7: 152852132-152852132 on Build GRCh38
rs1008271346 Chr.7: 152852269-152852269 on Build GRCh38
rs561116208 Chr.7: 152852071-152852071 on Build GRCh38
rs558174301 Chr.7: 152852348-152852348 on Build GRCh38

View Full Product Details