Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 90018635-90019168 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DECR1
Entrez Gene ID
1666
Gene Name
2,4-dienoyl-CoA reductase 1, mitochondrial
Gene Aliases
DECR, NADPH, SDR18C1
UniGene
Hs.492212
Gene Chromosome Location
Chr.8: 90001337-90052004 on Build GRCh38
Transcripts
NM_001359.1 XM_005250808.4 XM_011516880.2 XM_017013147.1 XM_017013148.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1193453 c.320C>G p.T107S Chr.8: 90018956-90018956 on Build GRCh38
7231598 c.273-2A>T p.? Chr.8: 90018907-90018907 on Build GRCh38
5529658 c.316C>T p.Q106* Chr.8: 90018952-90018952 on Build GRCh38
3413155 c.352G>A p.V118M Chr.8: 90019107-90019107 on Build GRCh38
3901981 c.408A>T p.G136G Chr.8: 90019163-90019163 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778496133 Chr.8: 90018939-90018939 on Build GRCh38
rs72671850 Chr.8: 90018782-90018782 on Build GRCh38
rs371896281 Chr.8: 90018969-90018969 on Build GRCh38
rs772249642 Chr.8: 90019040-90019040 on Build GRCh38
rs764382840 Chr.8: 90018869-90018869 on Build GRCh38
rs777833695 Chr.8: 90018885-90018885 on Build GRCh38
rs202002079 Chr.8: 90018680-90018680 on Build GRCh38
rs1805867 Chr.8: 90018846-90018846 on Build GRCh38
rs773153749 Chr.8: 90019146-90019146 on Build GRCh38
rs748575351 Chr.8: 90018860-90018860 on Build GRCh38
rs774230213 Chr.8: 90018891-90018891 on Build GRCh38
rs745535271 Chr.8: 90018841-90018841 on Build GRCh38
rs371840859 Chr.8: 90019068-90019068 on Build GRCh38
rs753515489 Chr.8: 90019058-90019058 on Build GRCh38
rs759758137 Chr.8: 90019035-90019035 on Build GRCh38
rs749672502 Chr.8: 90019089-90019089 on Build GRCh38
rs777959797 Chr.8: 90018981-90018981 on Build GRCh38
rs776731536 Chr.8: 90019143-90019143 on Build GRCh38
rs774900035 Chr.8: 90019125-90019125 on Build GRCh38
rs1055314326 Chr.8: 90018708-90018708 on Build GRCh38
rs907801937 Chr.8: 90018688-90018688 on Build GRCh38
rs895022305 Chr.8: 90018724-90018724 on Build GRCh38
rs767910567 Chr.8: 90018917-90018917 on Build GRCh38
rs151187801 Chr.8: 90019114-90019114 on Build GRCh38
rs756844777 Chr.8: 90018813-90018813 on Build GRCh38
rs1009178042 Chr.8: 90018866-90018866 on Build GRCh38
rs779718328 Chr.8: 90018962-90018962 on Build GRCh38
rs752965042 Chr.8: 90018919-90018919 on Build GRCh38
rs757107112 Chr.8: 90018938-90018938 on Build GRCh38
rs770884634 Chr.8: 90019044-90019044 on Build GRCh38
rs551418507 Chr.8: 90019060-90019060 on Build GRCh38
rs184650731 Chr.8: 90018789-90018789 on Build GRCh38
rs189154625 Chr.8: 90019034-90019034 on Build GRCh38
rs778240464 Chr.8: 90018819-90018819 on Build GRCh38
rs760589420 Chr.8: 90018930-90018930 on Build GRCh38
rs983346856 Chr.8: 90018684-90018684 on Build GRCh38
rs763777845 Chr.8: 90018933-90018933 on Build GRCh38
rs754519979 Chr.8: 90019084-90019084 on Build GRCh38
rs918097217 Chr.8: 90019139-90019139 on Build GRCh38
rs751242810 Chr.8: 90019082-90019082 on Build GRCh38
rs1056212951 Chr.8: 90018713-90018713 on Build GRCh38
rs759851485 Chr.8: 90018895-90018895 on Build GRCh38
rs867532362 Chr.8: 90018935-90018935 on Build GRCh38
rs779605523 Chr.8: 90019100-90019100 on Build GRCh38
rs1015961901 Chr.8: 90019007-90019007 on Build GRCh38
rs202215448 Chr.8: 90019107-90019107 on Build GRCh38
rs763984023 Chr.8: 90019056-90019056 on Build GRCh38
rs757896983 Chr.8: 90018948-90018948 on Build GRCh38
rs370716901 Chr.8: 90018877-90018877 on Build GRCh38
rs374362672 Chr.8: 90018913-90018913 on Build GRCh38
rs964907980 Chr.8: 90018912-90018912 on Build GRCh38
rs529586632 Chr.8: 90018940-90018940 on Build GRCh38
rs775675495 Chr.8: 90019112-90019112 on Build GRCh38
rs749805745 Chr.8: 90018995-90018995 on Build GRCh38
rs746896193 Chr.8: 90018937-90018937 on Build GRCh38
rs138725804 Chr.8: 90019121-90019121 on Build GRCh38
rs540780043 Chr.8: 90018812-90018812 on Build GRCh38
rs755961415 Chr.8: 90018802-90018802 on Build GRCh38
rs568855857 Chr.8: 90018967-90018967 on Build GRCh38
rs376164923 Chr.8: 90019003-90019003 on Build GRCh38
rs554490600 Chr.8: 90018749-90018749 on Build GRCh38
rs776506271 Chr.8: 90019077-90019077 on Build GRCh38
rs896705406 Chr.8: 90018722-90018722 on Build GRCh38
rs939169318 Chr.8: 90018704-90018704 on Build GRCh38
rs766510339 Chr.8: 90018890-90018890 on Build GRCh38
rs757630614 Chr.8: 90019002-90019003 on Build GRCh38
rs376239724 Chr.8: 90018876-90018876 on Build GRCh38
rs771323617 Chr.8: 90019094-90019094 on Build GRCh38
rs778214175 Chr.8: 90019086-90019086 on Build GRCh38
rs369033482 Chr.8: 90019051-90019051 on Build GRCh38
rs948800133 Chr.8: 90018739-90018739 on Build GRCh38
rs751881956 Chr.8: 90018869-90018873 on Build GRCh38
rs192012718 Chr.8: 90019123-90019123 on Build GRCh38
rs771944438 Chr.8: 90018856-90018856 on Build GRCh38
rs1805878 Chr.8: 90018699-90018699 on Build GRCh38
rs374823946 Chr.8: 90018881-90018881 on Build GRCh38
rs755210500 Chr.8: 90018775-90018775 on Build GRCh38
rs760560056 Chr.8: 90018795-90018799 on Build GRCh38
rs757950234 Chr.8: 90019067-90019067 on Build GRCh38
rs545868900 Chr.8: 90018757-90018757 on Build GRCh38
rs774909417 Chr.8: 90019013-90019013 on Build GRCh38
rs753840184 Chr.8: 90018934-90018934 on Build GRCh38
rs761550038 Chr.8: 90019133-90019133 on Build GRCh38
rs750327194 Chr.8: 90019066-90019066 on Build GRCh38
rs1805866 Chr.8: 90018682-90018682 on Build GRCh38
rs551338571 Chr.8: 90018944-90018944 on Build GRCh38
rs765152387 Chr.8: 90019065-90019065 on Build GRCh38
rs765056874 Chr.8: 90019135-90019135 on Build GRCh38
rs575267470 Chr.8: 90018710-90018710 on Build GRCh38

View Full Product Details