Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 167918075-167918591 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MPC2
Entrez Gene ID
25874
Gene Name
mitochondrial pyruvate carrier 2
Gene Aliases
BRP44
UniGene
Hs.517768
Gene Chromosome Location
Chr.1: 167916675-167937069 on Build GRCh38
Transcripts
NM_001143674.3 NM_015415.3 NR_026550.2 XM_006711266.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3477853 c.378C>T p.H126H Chr.1: 167918329-167918329 on Build GRCh38
5414561 c.353A>G p.N118S Chr.1: 167918354-167918354 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs374633504 Chr.1: 167918256-167918256 on Build GRCh38
rs931778440 Chr.1: 167918262-167918262 on Build GRCh38
rs944491252 Chr.1: 167918390-167918390 on Build GRCh38
rs938135790 Chr.1: 167918173-167918173 on Build GRCh38
rs750036351 Chr.1: 167918103-167918103 on Build GRCh38
rs773579672 Chr.1: 167918278-167918278 on Build GRCh38
rs371012330 Chr.1: 167918363-167918363 on Build GRCh38
rs1048421538 Chr.1: 167918414-167918414 on Build GRCh38
rs1007316171 Chr.1: 167918408-167918408 on Build GRCh38
rs745590496 Chr.1: 167918361-167918361 on Build GRCh38
rs189565081 Chr.1: 167918248-167918248 on Build GRCh38
rs919246864 Chr.1: 167918105-167918105 on Build GRCh38
rs376142767 Chr.1: 167918281-167918281 on Build GRCh38
rs146964973 Chr.1: 167918469-167918469 on Build GRCh38
rs770308563 Chr.1: 167918385-167918385 on Build GRCh38
rs551307466 Chr.1: 167918189-167918189 on Build GRCh38
rs1050037 Chr.1: 167918178-167918178 on Build GRCh38
rs375408238 Chr.1: 167918113-167918113 on Build GRCh38
rs567720057 Chr.1: 167918394-167918394 on Build GRCh38
rs1050036 Chr.1: 167918198-167918198 on Build GRCh38
rs774678637 Chr.1: 167918286-167918286 on Build GRCh38
rs926874842 Chr.1: 167918158-167918158 on Build GRCh38
rs533244024 Chr.1: 167918188-167918188 on Build GRCh38
rs982127110 Chr.1: 167918489-167918489 on Build GRCh38
rs1026427115 Chr.1: 167918447-167918447 on Build GRCh38
rs10918798 Chr.1: 167918404-167918404 on Build GRCh38
rs754875111 Chr.1: 167918440-167918440 on Build GRCh38
rs181568773 Chr.1: 167918428-167918428 on Build GRCh38
rs539338523 Chr.1: 167918444-167918444 on Build GRCh38
rs949435410 Chr.1: 167918143-167918143 on Build GRCh38
rs764070318 Chr.1: 167918317-167918317 on Build GRCh38
rs960462662 Chr.1: 167918144-167918144 on Build GRCh38
rs989788116 Chr.1: 167918520-167918520 on Build GRCh38
rs746463976 Chr.1: 167918364-167918364 on Build GRCh38
rs1050580422 Chr.1: 167918263-167918263 on Build GRCh38
rs572663249 Chr.1: 167918495-167918495 on Build GRCh38
rs755214300 Chr.1: 167918406-167918406 on Build GRCh38
rs543212360 Chr.1: 167918551-167918551 on Build GRCh38
rs773673277 Chr.1: 167918389-167918389 on Build GRCh38
rs982346135 Chr.1: 167918150-167918150 on Build GRCh38
rs148820068 Chr.1: 167918433-167918433 on Build GRCh38
rs767766833 Chr.1: 167918301-167918301 on Build GRCh38
rs780359074 Chr.1: 167918305-167918305 on Build GRCh38
rs759657831 Chr.1: 167918294-167918294 on Build GRCh38
rs1034550353 Chr.1: 167918142-167918142 on Build GRCh38
rs959619016 Chr.1: 167918508-167918508 on Build GRCh38
rs756129812 Chr.1: 167918306-167918306 on Build GRCh38
rs778548749 Chr.1: 167918357-167918357 on Build GRCh38
rs879461954 Chr.1: 167918193-167918193 on Build GRCh38
rs757115050 Chr.1: 167918356-167918356 on Build GRCh38
rs763328907 Chr.1: 167918279-167918279 on Build GRCh38
rs879066892 Chr.1: 167918337-167918337 on Build GRCh38
rs770984315 Chr.1: 167918498-167918498 on Build GRCh38
rs915376961 Chr.1: 167918547-167918547 on Build GRCh38
rs761684083 Chr.1: 167918308-167918309 on Build GRCh38
rs536794472 Chr.1: 167918338-167918338 on Build GRCh38
rs771378476 Chr.1: 167918391-167918391 on Build GRCh38
rs548782562 Chr.1: 167918254-167918254 on Build GRCh38
rs879495211 Chr.1: 167918430-167918430 on Build GRCh38

View Full Product Details