Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 171185169-171185675 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMO2
Entrez Gene ID
2327
Gene Name
flavin containing monooxygenase 2
Gene Aliases
FMO1B1
UniGene
Hs.144912
Gene Chromosome Location
Chr.1: 171185208-171212683 on Build GRCh38
Transcripts
NM_001301347.1 NM_001460.4

SNP Detail

SNP ID Chromosome Location
rs542998756 Chr.1: 171185287-171185287 on Build GRCh38
rs781117214 Chr.1: 171185434-171185434 on Build GRCh38
rs181710264 Chr.1: 171185467-171185467 on Build GRCh38
rs28369797 Chr.1: 171185279-171185279 on Build GRCh38
rs1056136642 Chr.1: 171185575-171185575 on Build GRCh38
rs190249141 Chr.1: 171185450-171185450 on Build GRCh38
rs1037136122 Chr.1: 171185522-171185522 on Build GRCh38
rs543885917 Chr.1: 171185298-171185298 on Build GRCh38
rs926343984 Chr.1: 171185195-171185195 on Build GRCh38
rs150129010 Chr.1: 171185515-171185515 on Build GRCh38
rs770072774 Chr.1: 171185587-171185587 on Build GRCh38
rs565457647 Chr.1: 171185410-171185410 on Build GRCh38
rs58582514 Chr.1: 171185306-171185306 on Build GRCh38
rs555049310 Chr.1: 171185332-171185332 on Build GRCh38
rs76414019 Chr.1: 171185506-171185506 on Build GRCh38
rs769427859 Chr.1: 171185558-171185558 on Build GRCh38
rs3215452 Chr.1: 171185505-171185505 on Build GRCh38
rs1032776321 Chr.1: 171185624-171185624 on Build GRCh38
rs1008309996 Chr.1: 171185347-171185347 on Build GRCh38
rs748155803 Chr.1: 171185546-171185546 on Build GRCh38
rs1043386714 Chr.1: 171185644-171185644 on Build GRCh38
rs146530714 Chr.1: 171185232-171185232 on Build GRCh38
rs186189927 Chr.1: 171185337-171185337 on Build GRCh38
rs897317978 Chr.1: 171185530-171185530 on Build GRCh38
rs961350854 Chr.1: 171185414-171185414 on Build GRCh38
rs531458466 Chr.1: 171185357-171185361 on Build GRCh38
rs998483803 Chr.1: 171185552-171185552 on Build GRCh38
rs970356116 Chr.1: 171185495-171185495 on Build GRCh38
rs945030407 Chr.1: 171185611-171185611 on Build GRCh38
rs867317528 Chr.1: 171185227-171185227 on Build GRCh38
rs997054828 Chr.1: 171185311-171185311 on Build GRCh38
rs1044585456 Chr.1: 171185194-171185194 on Build GRCh38
rs28369796 Chr.1: 171185239-171185239 on Build GRCh38
rs867125925 Chr.1: 171185240-171185240 on Build GRCh38
rs559143716 Chr.1: 171185475-171185475 on Build GRCh38
rs751684079 Chr.1: 171185468-171185468 on Build GRCh38
rs777638948 Chr.1: 171185573-171185573 on Build GRCh38
rs957200352 Chr.1: 171185280-171185280 on Build GRCh38
rs553943994 Chr.1: 171185252-171185252 on Build GRCh38
rs755165390 Chr.1: 171185497-171185497 on Build GRCh38
rs1006934325 Chr.1: 171185608-171185608 on Build GRCh38
rs949971205 Chr.1: 171185390-171185390 on Build GRCh38
rs985269390 Chr.1: 171185263-171185263 on Build GRCh38
rs1015726841 Chr.1: 171185372-171185372 on Build GRCh38
rs780968699 Chr.1: 171185533-171185533 on Build GRCh38
rs576800559 Chr.1: 171185335-171185335 on Build GRCh38
rs1046046512 Chr.1: 171185403-171185403 on Build GRCh38

View Full Product Details