Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 171209085-171209582 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMO2
Entrez Gene ID
2327
Gene Name
flavin containing monooxygenase 2
Gene Aliases
FMO1B1
UniGene
Hs.144912
Gene Chromosome Location
Chr.1: 171185208-171212683 on Build GRCh38
Transcripts
NM_001301347.1 NM_001460.4

SNP Detail

SNP ID Chromosome Location
rs976291823 Chr.1: 171209527-171209527 on Build GRCh38
rs900474285 Chr.1: 171209554-171209554 on Build GRCh38
rs961109546 Chr.1: 171209254-171209254 on Build GRCh38
rs200428987 Chr.1: 171209126-171209126 on Build GRCh38
rs1004577840 Chr.1: 171209485-171209485 on Build GRCh38
rs550656602 Chr.1: 171209319-171209319 on Build GRCh38
rs926062397 Chr.1: 171209264-171209264 on Build GRCh38
rs545794963 Chr.1: 171209132-171209132 on Build GRCh38
rs891663098 Chr.1: 171209416-171209416 on Build GRCh38
rs891954745 Chr.1: 171209304-171209304 on Build GRCh38
rs28369914 Chr.1: 171209351-171209351 on Build GRCh38
rs532956235 Chr.1: 171209377-171209377 on Build GRCh38
rs772162212 Chr.1: 171209433-171209433 on Build GRCh38
rs938623915 Chr.1: 171209365-171209365 on Build GRCh38
rs371841312 Chr.1: 171209207-171209207 on Build GRCh38
rs948430040 Chr.1: 171209310-171209310 on Build GRCh38
rs983288979 Chr.1: 171209110-171209110 on Build GRCh38
rs955161847 Chr.1: 171209143-171209143 on Build GRCh38
rs2234889 Chr.1: 171209118-171209119 on Build GRCh38
rs1045998026 Chr.1: 171209344-171209344 on Build GRCh38
rs745908121 Chr.1: 171209352-171209352 on Build GRCh38
rs1028919278 Chr.1: 171209536-171209536 on Build GRCh38
rs763946864 Chr.1: 171209438-171209438 on Build GRCh38
rs1027692306 Chr.1: 171209406-171209406 on Build GRCh38
rs564621161 Chr.1: 171209161-171209161 on Build GRCh38
rs397824010 Chr.1: 171209125-171209126 on Build GRCh38
rs1015142758 Chr.1: 171209242-171209242 on Build GRCh38
rs760606831 Chr.1: 171209439-171209439 on Build GRCh38
rs28369915 Chr.1: 171209487-171209487 on Build GRCh38
rs540720849 Chr.1: 171209402-171209402 on Build GRCh38
rs573522092 Chr.1: 171209209-171209209 on Build GRCh38
rs188310906 Chr.1: 171209517-171209517 on Build GRCh38
rs540449232 Chr.1: 171209265-171209265 on Build GRCh38
rs184768578 Chr.1: 171209281-171209281 on Build GRCh38
rs1014735863 Chr.1: 171209486-171209486 on Build GRCh38
rs1001214700 Chr.1: 171209363-171209363 on Build GRCh38
rs866288114 Chr.1: 171209405-171209405 on Build GRCh38
rs529527458 Chr.1: 171209283-171209283 on Build GRCh38
rs566641271 Chr.1: 171209498-171209498 on Build GRCh38
rs985831111 Chr.1: 171209253-171209253 on Build GRCh38
rs564866845 Chr.1: 171209197-171209197 on Build GRCh38
rs907708541 Chr.1: 171209116-171209116 on Build GRCh38
rs879012132 Chr.1: 171209196-171209197 on Build GRCh38
rs192580205 Chr.1: 171209124-171209124 on Build GRCh38

View Full Product Details