Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 171987401-171987940 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNM3
Entrez Gene ID
26052
Gene Name
dynamin 3
Gene Aliases
Dyna III
UniGene
Hs.654775
Gene Chromosome Location
Chr.1: 171841478-172418466 on Build GRCh38
Transcripts
NM_001136127.2 NM_001278252.1 NM_015569.4 XM_005245079.1 XM_005245080.1 XM_006711268.2 XM_017000976.1 XM_017000977.1 XM_017000978.1 XM_017000979.1 XM_017000980.1 XM_017000981.1 XM_017000982.1 XM_017000983.1 XM_017000984.1 XM_017000985.1 XM_017000986.1 XM_017000987.1 XM_017000988.1 XM_017000989.1 XM_017000990.1 XM_017000991.1 XM_017000992.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1320747 c.367C>T p.R123* Chr.1: 171987787-171987787 on Build GRCh38
224642 c.277G>A p.D93N Chr.1: 171987697-171987697 on Build GRCh38
2075001 c.292C>T p.R98C Chr.1: 171987712-171987712 on Build GRCh38
3478354 c.333G>A p.M111I Chr.1: 171987753-171987753 on Build GRCh38
6647644 c.269A>C p.K90T Chr.1: 171987689-171987689 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs944693334 Chr.1: 171987868-171987868 on Build GRCh38
rs917538663 Chr.1: 171987555-171987555 on Build GRCh38
rs374793664 Chr.1: 171987783-171987783 on Build GRCh38
rs761593512 Chr.1: 171987735-171987735 on Build GRCh38
rs540025554 Chr.1: 171987548-171987548 on Build GRCh38
rs748364887 Chr.1: 171987749-171987749 on Build GRCh38
rs575621768 Chr.1: 171987903-171987903 on Build GRCh38
rs767058578 Chr.1: 171987468-171987468 on Build GRCh38
rs183183173 Chr.1: 171987608-171987608 on Build GRCh38
rs774067894 Chr.1: 171987794-171987794 on Build GRCh38
rs371617456 Chr.1: 171987805-171987805 on Build GRCh38
rs767017496 Chr.1: 171987799-171987799 on Build GRCh38
rs754976492 Chr.1: 171987827-171987827 on Build GRCh38
rs910658547 Chr.1: 171987838-171987838 on Build GRCh38
rs865780128 Chr.1: 171987700-171987700 on Build GRCh38
rs764142675 Chr.1: 171987477-171987477 on Build GRCh38
rs749268391 Chr.1: 171987641-171987641 on Build GRCh38
rs200490605 Chr.1: 171987804-171987804 on Build GRCh38
rs571773722 Chr.1: 171987729-171987729 on Build GRCh38
rs778071141 Chr.1: 171987763-171987763 on Build GRCh38
rs779779066 Chr.1: 171987627-171987627 on Build GRCh38
rs773516911 Chr.1: 171987677-171987677 on Build GRCh38
rs771360366 Chr.1: 171987770-171987770 on Build GRCh38
rs1006201715 Chr.1: 171987438-171987438 on Build GRCh38
rs201168352 Chr.1: 171987704-171987704 on Build GRCh38
rs528001323 Chr.1: 171987581-171987581 on Build GRCh38
rs901122107 Chr.1: 171987717-171987717 on Build GRCh38
rs190375910 Chr.1: 171987571-171987571 on Build GRCh38
rs992521621 Chr.1: 171987788-171987788 on Build GRCh38
rs572275619 Chr.1: 171987850-171987850 on Build GRCh38
rs113625155 Chr.1: 171987465-171987465 on Build GRCh38
rs538785687 Chr.1: 171987761-171987761 on Build GRCh38
rs201273492 Chr.1: 171987751-171987751 on Build GRCh38
rs758769077 Chr.1: 171987871-171987871 on Build GRCh38
rs1030088718 Chr.1: 171987478-171987478 on Build GRCh38
rs938931814 Chr.1: 171987695-171987695 on Build GRCh38
rs527384140 Chr.1: 171987712-171987712 on Build GRCh38
rs374618528 Chr.1: 171987491-171987491 on Build GRCh38
rs755001015 Chr.1: 171987742-171987742 on Build GRCh38
rs901071139 Chr.1: 171987878-171987878 on Build GRCh38
rs952514669 Chr.1: 171987445-171987445 on Build GRCh38
rs542533865 Chr.1: 171987888-171987888 on Build GRCh38
rs748391175 Chr.1: 171987668-171987668 on Build GRCh38
rs752717562 Chr.1: 171987849-171987849 on Build GRCh38
rs778258725 Chr.1: 171987539-171987539 on Build GRCh38
rs376217186 Chr.1: 171987811-171987811 on Build GRCh38
rs372756678 Chr.1: 171987737-171987737 on Build GRCh38
rs752295847 Chr.1: 171987494-171987494 on Build GRCh38
rs761542182 Chr.1: 171987798-171987798 on Build GRCh38
rs567650154 Chr.1: 171987640-171987640 on Build GRCh38
rs1017458195 Chr.1: 171987552-171987552 on Build GRCh38
rs747540202 Chr.1: 171987687-171987687 on Build GRCh38
rs756070921 Chr.1: 171987635-171987635 on Build GRCh38
rs200882657 Chr.1: 171987722-171987722 on Build GRCh38
rs897914371 Chr.1: 171987808-171987808 on Build GRCh38
rs541085879 Chr.1: 171987778-171987778 on Build GRCh38
rs766159947 Chr.1: 171987739-171987739 on Build GRCh38
rs894442045 Chr.1: 171987433-171987433 on Build GRCh38
rs747273591 Chr.1: 171987768-171987768 on Build GRCh38
rs369476188 Chr.1: 171987845-171987845 on Build GRCh38
rs77868630 Chr.1: 171987902-171987902 on Build GRCh38
rs1017974112 Chr.1: 171987859-171987859 on Build GRCh38
rs954642463 Chr.1: 171987498-171987498 on Build GRCh38
rs772395153 Chr.1: 171987672-171987672 on Build GRCh38
rs926006045 Chr.1: 171987588-171987588 on Build GRCh38
rs755949799 Chr.1: 171987738-171987738 on Build GRCh38
rs936042513 Chr.1: 171987815-171987815 on Build GRCh38
rs1007334429 Chr.1: 171987857-171987857 on Build GRCh38
rs768247468 Chr.1: 171987789-171987789 on Build GRCh38
rs1050529525 Chr.1: 171987832-171987832 on Build GRCh38
rs771286083 Chr.1: 171987697-171987697 on Build GRCh38
rs755583246 Chr.1: 171987645-171987645 on Build GRCh38
rs966446156 Chr.1: 171987661-171987661 on Build GRCh38
rs753751868 Chr.1: 171987813-171987813 on Build GRCh38
rs984164579 Chr.1: 171987464-171987464 on Build GRCh38
rs1025491474 Chr.1: 171987723-171987723 on Build GRCh38
rs3736790 Chr.1: 171987663-171987663 on Build GRCh38
rs753844227 Chr.1: 171987741-171987741 on Build GRCh38

View Full Product Details